This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02744463_cn
See other CHD1L CNV Assays ›
Gene Symbol
CHD1L
Assay Reference Genome
Location

Chr.1:147201312 on build GRCh38
Cytoband
1q21.1
Assay ID Hs02744463_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9557

Gene Name:

chromodomain helicase DNA binding protein 1 like

Gene Aliases:

ALC1, CHDL

Location:

Chr.1:147172804-147295766 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CHD1L XM_017002858.1 XP_016858347.1

Target Gene Details

Entrez Gene ID:

2330

Gene Name:

flavin containing monooxygenase 5

Gene Aliases:

-

Location:

Chr.1:147183963-147225798 on Build GRCh38

Assay Gene Location:

Within Exon 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FMO5 NM_001144829.2 7 1422 NP_001138301.1
NM_001144830.2 NP_001138302.1
NM_001461.3 7 1412 NP_001452.2
XM_005272946.4 7 1386 XP_005273003.1
XM_005272947.4 7 1130 XP_005273004.1
XM_005272948.4 7 1113 XP_005273005.1
XM_006711245.3 XP_006711308.1
XM_011509350.2 7 1417 XP_011507652.1
XM_011509351.2 6 1149 XP_011507653.1
XM_017000801.1 6 1161 XP_016856290.1
XM_017000802.1 6 931 XP_016856291.1
AK222728.1 7 1127 BAD96448.1
AK225739.1 7 1137
AK314647.1 7 1340
BC035687.1 AAH35687.1
L37080.1 7 1098 AAA67849.1
Z47553.1 7 1104 CAA87633.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv327n100 Chr.1:146028214 - 148860637 on Build GRCh38 Gain LINC01138 RNU1-13P LOC728989 PDE4DIP LINC00624 PDZK1P1 LOC105371226 LOC100132057 NBPF10 RNVU1-7 MIR6077 LOC101928979 LOC100505824 NBPF14 MIR5087 PDIA3P1 LOC101927468 NUDT4P2 ACP6 BCL9 CHD1L LOC105371242 PPIAL4G PRKAB2 RNVU1-6 RNVU1-8 NUDT4P1 NBPF13P CH17-408M7.1 FMO5 NBPF11 GPR89B NBPF12 RNVU1-3 RNVU1-1 GJA8 NOTCH2NL LOC100996763 HYDIN2 GJA5 LOC100996517 LOC284561
nsv1005055 Chr.1:145642988 - 148991068 on Build GRCh38 Loss LINC01138 LOC728989 PDE4DIP LOC105371226 LOC100132057 NUDT17 LOC101928979 NBPF14 TXNIP MIR5087 PDIA3P1 GNRHR2 LOC105371242 PPIAL4G PRKAB2 ANKRD35 NUDT4P1 NBPF13P NBPF11 GPR89B NBPF12 MIR6736 RNVU1-1 PIAS3 LOC100996763 LOC284561 RNU1-13P CD160 LINC00624 PDZK1P1 NBPF10 RNVU1-7 HFE2 PDZK1 MIR6077 GPR89A LOC100505824 RNF115 ITGA10 LOC101927468 NUDT4P2 ACP6 BCL9 LIX1L CHD1L RNVU1-6 RNVU1-8 POLR3C PEX11B CH17-408M7.1 POLR3GL FMO5 RNVU1-3 ANKRD34A GJA8 NOTCH2NL HYDIN2 GJA5 LOC100996517 RBM8A
nsv1121713 Chr.1:147010816 - 147207577 on Build GRCh38 Deletion LOC728989 FMO5 CHD1L PRKAB2 PDIA3P1 RNVU1-8 NBPF13P
esv2761299 Chr.1:146068341 - 148926801 on Build GRCh38 Gain+Loss LINC01138 RNU1-13P LOC728989 PDE4DIP LINC00624 PDZK1P1 LOC105371226 LOC100132057 NBPF10 RNVU1-7 MIR6077 LOC100505824 NBPF14 MIR5087 PDIA3P1 LOC101927468 NUDT4P2 ACP6 BCL9 CHD1L LOC105371242 PPIAL4G PRKAB2 RNVU1-8 NUDT4P1 NBPF13P CH17-408M7.1 FMO5 NBPF11 GPR89B NBPF12 RNVU1-3 RNVU1-1 GJA8 NOTCH2NL LOC100996763 HYDIN2 GJA5 LOC100996517 LOC284561
nsv509457 Chr.1:145948024 - 149021821 on Build GRCh38 Insertion LINC01138 RNU1-13P LOC728989 PDE4DIP LINC00624 PDZK1P1 LOC105371226 LOC100132057 NBPF10 RNVU1-7 HFE2 MIR6077 LOC101928979 LOC100505824 NBPF14 TXNIP MIR5087 PDIA3P1 LOC101927468 NUDT4P2 ACP6 BCL9 LIX1L CHD1L LOC105371242 PPIAL4G PRKAB2 RNVU1-6 RNVU1-8 NUDT4P1 NBPF13P CH17-408M7.1 POLR3GL FMO5 NBPF11 GPR89B NBPF12 RNVU1-3 RNVU1-1 ANKRD34A GJA8 NOTCH2NL LOC100996763 HYDIN2 GJA5 LOC100996517 LOC284561
nsv1132304 Chr.1:146186094 - 148977674 on Build GRCh38 Duplication LINC01138 RNU1-13P LOC728989 PDE4DIP LINC00624 PDZK1P1 LOC105371226 LOC100132057 RNVU1-7 MIR6077 LOC100505824 NBPF14 MIR5087 PDIA3P1 LOC101927468 NUDT4P2 ACP6 BCL9 CHD1L LOC105371242 PPIAL4G PRKAB2 RNVU1-8 NUDT4P1 NBPF13P CH17-408M7.1 FMO5 NBPF11 GPR89B NBPF12 RNVU1-3 RNVU1-1 GJA8 NOTCH2NL LOC100996763 HYDIN2 GJA5 LOC100996517 LOC284561
nsv1003688 Chr.1:146036817 - 148934526 on Build GRCh38 Gain LINC01138 RNU1-13P LOC728989 PDE4DIP LINC00624 PDZK1P1 LOC105371226 LOC100132057 NBPF10 RNVU1-7 MIR6077 LOC101928979 LOC100505824 NBPF14 MIR5087 PDIA3P1 LOC101927468 NUDT4P2 ACP6 BCL9 CHD1L LOC105371242 PPIAL4G PRKAB2 RNVU1-6 RNVU1-8 NUDT4P1 NBPF13P CH17-408M7.1 FMO5 NBPF11 GPR89B NBPF12 RNVU1-3 RNVU1-1 GJA8 NOTCH2NL LOC100996763 HYDIN2 GJA5 LOC100996517 LOC284561
dgv66e199 Chr.1:146055970 - 149056514 on Build GRCh38 Deletion LINC01138 RNU1-13P LOC728989 PDE4DIP LINC00624 PDZK1P1 LOC105371226 LOC100132057 NBPF10 RNVU1-7 MIR6077 LOC101928979 LOC100505824 NBPF9 NBPF14 MIR5087 PDIA3P1 LOC101927468 NUDT4P2 ACP6 BCL9 CHD1L LOC105371242 PPIAL4G PRKAB2 RNVU1-8 NUDT4P1 NBPF13P CH17-408M7.1 FMO5 NBPF11 GPR89B NBPF12 RNVU1-3 RNVU1-1 GJA8 NOTCH2NL LOC100996763 HYDIN2 GJA5 LOC100996517 LOC284561
nsv437047 Chr.1:147171976 - 147369886 on Build GRCh38 Loss FMO5 CHD1L PRKAB2 PDIA3P1

Back To Top

More Information


Set Membership:

Intragenic Exonic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
chromatin remodeling
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
global genome nucleotide-excision repair
biological_process
drug metabolic process
oxidation-reduction process
nucleotide binding
nucleic acid binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
ATPase activity
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline