This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02765193_cn
See other NDRG2 CNV Assays ›
Gene Symbol
NDRG2
Assay Reference Genome
Location

Chr.14:21016901 on build GRCh38
Cytoband
14q11.2
Assay ID Hs02765193_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

57447

Gene Name:

NDRG family member 2

Gene Aliases:

SYLD

Location:

Chr.14:21016763-21070872 on Build GRCh38

Assay Gene Location:

Within Exon 21
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NDRG2 NM_001282211.1 15 1820 NP_001269140.1
NM_001282212.1 15 2046 NP_001269141.1
NM_001282213.1 16 2099 NP_001269142.1
NM_001282214.1 16 2087 NP_001269143.1
NM_001282215.1 16 1884 NP_001269144.1
NM_001282216.1 15 1835 NP_001269145.1
NM_001320329.1 16 2752 NP_001307258.1
NM_016250.2 16 1942 NP_057334.1
NM_201535.1 17 1984 NP_963293.1
NM_201536.1 15 1875 NP_963294.1
NM_201537.1 16 1917 NP_963831.1
NM_201538.1 15 1852 NP_963832.1
NM_201539.1 16 1894 NP_963833.1
NM_201540.1 17 1961 NP_963834.1
NM_201541.1 16 1919 NP_963835.1
XM_011536996.2 17 1971 XP_011535298.1
XM_011536997.1 17 1972 XP_011535299.1
XM_011536998.1 17 1962 XP_011535300.1
XM_011536999.1 15 2367 XP_011535301.1
XM_011537001.1 16 1925 XP_011535303.1
XM_011537002.1 16 2071 XP_011535304.1
XM_017021480.1 17 2073 XP_016876969.1
XM_017021481.1 17 2112 XP_016876970.1
XM_017021482.1 17 2121 XP_016876971.1
XM_017021483.1 16 2005 XP_016876972.1
XM_017021484.1 16 2047 XP_016876973.1
XM_017021485.1 17 2112 XP_016876974.1
XM_017021486.1 17 2122 XP_016876975.1
XM_017021487.1 16 2031 XP_016876976.1
XM_017021488.1 16 2072 XP_016876977.1
XM_017021489.1 16 2177 XP_016876978.1
XM_017021490.1 15 1969 XP_016876979.1
XM_017021491.1 15 2004 XP_016876980.1
XM_017021492.1 16 2075 XP_016876981.1
XM_017021493.1 16 2221 XP_016876982.1
XM_017021494.1 18 1977 XP_016876983.1
XM_017021495.1 17 1909 XP_016876984.1
XM_017021496.1 16 1908 XP_016876985.1
XM_017021497.1 11 1602 XP_016876986.1
AB033074.1 15 1804
AF159092.3 16 1865 AAD43131.2
AF304051.1 17 1984 AAL08624.1
AK024521.1 17 1917
AK057843.1 16 1888
AK057847.1 15 1852
AK090657.1 15 1791
AK096999.1
AK130029.1 15 1790
AL136574.1 15 1829 CAB66509.1
BC010458.1 15 1823 AAH10458.1
BC011240.1 16 1844 AAH11240.1
BC093038.1 16 1918 AAH93038.1
BF341160.1 1 348
BX247987.1 15 1905 CAD62321.1
BX248031.1 13 1694 CAD62350.1
BX647091.1 13 2017
BX647748.1 16 2034
CR749252.1 16 2087 CAH18108.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv428293 Chr.14:20747949 - 21063624 on Build GRCh38 Gain+Loss SLC39A2 RNASE8 RNASE7 RNASE13 NDRG2 EDDM3A RNASE3 LOC101929718 RNASE2 TPPP2 RNASE1 ECRP ARHGEF40 RNASE6 MIR6717 METTL17 EDDM3B
nsv832740 Chr.14:20863901 - 21076893 on Build GRCh38 Gain SLC39A2 RNASE8 RNASE7 RNASE13 NDRG2 RNASE3 LOC101929718 RNASE2 TPPP2 ECRP ARHGEF40 MIR6717 METTL17

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cytokine production
signal transduction
regulation of vascular endothelial growth factor production
Wnt signaling pathway
substantia nigra development
cell differentiation
negative regulation of smooth muscle cell proliferation
negative regulation of ERK1 and ERK2 cascade
regulation of platelet-derived growth factor production
molecular_function
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline