This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02796347_cn
See other ESRRA CNV Assays ›
Gene Symbol
ESRRA
Assay Reference Genome
Location

Chr.11:64307178 on build GRCh38
Cytoband
11q13.1
Assay ID Hs02796347_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2101

Gene Name:

estrogen related receptor alpha

Gene Aliases:

ERR1, ERRa, ERRalpha, ESRL1, NR3B1

Location:

Chr.11:64305528-64316743 on Build GRCh38

Assay Gene Location:

Within Exon 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ESRRA NM_001282450.1 NP_001269379.1
NM_001282451.1 NP_001269380.1
NM_004451.4 NP_004442.3
XM_017017313.1 1 724 XP_016872802.1
BC063795.1
BC073956.1 AAH73956.1
BC092470.1 AAH92470.1
BC093720.1 AAH93720.1
BC093722.1 AAH93722.1
DB478332.1
HQ692867.1
L38487.1 AAB17015.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv951010 Chr.11:63942829 - 64352128 on Build GRCh38 Deletion DNAJC4 TEX40 COX8A STIP1 PLCB3 CCDC88B NUDT22 GPR137 LOC105369340 FERMT3 MIR7155 MACROD1 PRDX5 BAD KCNK4 KCNK4-TEX40 TRMT112 ESRRA TRPT1 FKBP2 VEGFB OTUB1 NAA40 PPP1R14B FLRT1
dgv1963n54 Chr.11:64174262 - 64329761 on Build GRCh38 Loss DNAJC4 TEX40 BAD KCNK4 KCNK4-TEX40 TRMT112 STIP1 PLCB3 ESRRA TRPT1 NUDT22 GPR137 FKBP2 VEGFB LOC105369340 FERMT3 PPP1R14B PRDX5
nsv832186 Chr.11:64204788 - 64402587 on Build GRCh38 Loss DNAJC4 TEX40 BAD KCNK4 KCNK4-TEX40 TRMT112 PLCB3 ESRRA TRPT1 CCDC88B NUDT22 GPR137 FKBP2 MIR1237 VEGFB LOC105369340 FERMT3 MIR7155 PPP1R14B PRDX5 RPS6KA4
nsv469961 Chr.11:64055468 - 64318495 on Build GRCh38 Loss DNAJC4 TEX40 BAD KCNK4 KCNK4-TEX40 TRMT112 STIP1 PLCB3 ESRRA TRPT1 NUDT22 GPR137 FKBP2 VEGFB LOC105369340 FERMT3 PPP1R14B FLRT1 MACROD1 PRDX5
nsv357 Chr.11:64284335 - 64308789 on Build GRCh38 Insertion TEX40 BAD KCNK4 KCNK4-TEX40 ESRRA GPR137

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
mitochondrion organization
intracellular receptor signaling pathway
response to estradiol
regulation of cell proliferation
steroid hormone mediated signaling pathway
regulation of osteoblast differentiation
regulation of osteoclast differentiation
positive regulation of transcription from RNA polymerase II promoter
cartilage development
positive regulation of cellular response to insulin stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
steroid binding
protein binding
zinc ion binding
protein domain specific binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline