This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02841063_cn
See other MUTYH CNV Assays ›
Gene Symbol
MUTYH
Assay Reference Genome
Location

Chr.1:45331230 on build GRCh38
Cytoband
1p34.1
Assay ID Hs02841063_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

4595

Gene Name:

mutY DNA glycosylase

Gene Aliases:

MYH

Location:

Chr.1:45329242-45340925 on Build GRCh38

Assay Gene Location:

Within Exon 20
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MUTYH NM_001048171.1 14 1602 NP_001041636.1
NM_001048172.1 14 1493 NP_001041637.1
NM_001048173.1 14 1490 NP_001041638.1
NM_001048174.1 14 1409 NP_001041639.1
NM_001128425.1 14 1644 NP_001121897.1
NM_001293190.1 14 1605 NP_001280119.1
NM_001293191.1 14 1442 NP_001280120.1
NM_001293192.1 14 1538 NP_001280121.1
NM_001293195.1 15 1559 NP_001280124.1
NM_001293196.1 14 1427 NP_001280125.1
NM_012222.2 14 1635 NP_036354.1
XM_011541497.2 14 1701 XP_011539799.1
XM_011541498.1 14 1479 XP_011539800.1
XM_011541499.1 14 1465 XP_011539801.1
XM_011541500.2 14 1660 XP_011539802.1
XM_011541501.2 15 1574 XP_011539803.1
XM_011541502.2 14 1479 XP_011539804.1
XM_011541503.2 13 1447 XP_011539805.1
XM_011541504.2 14 1528 XP_011539806.1
XM_011541505.2 10 1148 XP_011539807.1
XM_011541506.1 10 1132 XP_011539808.1
XM_011541507.2 9 944 XP_011539809.2
XM_017001331.1 14 1478 XP_016856820.1
XM_017001332.1 14 1480 XP_016856821.1
XM_017001333.1 14 1533 XP_016856822.1
XM_017001334.1 14 1426 XP_016856823.1
XM_017001335.1 14 1372 XP_016856824.1
XM_017001336.1 13 1456 XP_016856825.1
XM_017001337.1 13 1396 XP_016856826.1
XM_017001338.1 13 1983 XP_016856827.1
XM_017001339.1 13 1434 XP_016856828.1
AB032920.1 14 1553 BAA89336.1
AB032921.1 14 1523 BAA89337.1
AB032922.1 14 1520 BAA89338.1
AB032923.1 14 1456 BAA89339.1
AB032924.1 14 1420 BAA89340.1
AB032925.1 14 1387 BAA89341.1
AB032926.1 15 1537 BAA89342.1
AB032927.1 14 1458 BAA89343.1
AB032928.1 14 1455 BAA89344.1
AB032929.1 14 1391 BAA89345.1
AK293830.1 5 664
BC003178.1 14 1540 AAH03178.1
KU178203.1 14 1386
KU178204.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2750837 Chr.1:45330443 - 45442591 on Build GRCh38 Loss TOE1 TESK2 MUTYH

Back To Top

More Information


Additional Information:

For this assay, SNP(s) [rs74318065] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

DNA glycosylase

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
mismatch repair
response to oxidative stress
depurination
DNA binding
protein binding
DNA N-glycosylase activity
MutLalpha complex binding
MutLbeta complex binding
MutSalpha complex binding
MutSbeta complex binding
metal ion binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-t8jrk:80/100.66.77.131:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0