This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02852661_cn
See other UCHL5 CNV Assays ›
Gene Symbol
UCHL5
Assay Reference Genome
Location

Chr.1:193015748 on build GRCh38
Cytoband
1q31.2
Assay ID Hs02852661_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

51377

Gene Name:

ubiquitin C-terminal hydrolase L5

Gene Aliases:

CGI-70, INO80R, UCH-L5, UCH37

Location:

Chr.1:193012366-193060140 on Build GRCh38

Assay Gene Location:

Within Exon 15
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UCHL5 NM_001199261.1 11 1723 NP_001186190.1
NM_015984.3 11 1726 NP_057068.1
XM_005245244.4 12 1887 XP_005245301.1
XM_005245246.4 12 1881 XP_005245303.1
XM_005245247.4 11 1661 XP_005245304.1
XM_005245248.4 11 1681 XP_005245305.1
XM_005245249.4 11 1580 XP_005245306.1
XM_006711366.3 12 1890 XP_006711429.1
XM_006711367.3 13 1965 XP_006711430.1
XM_006711368.3 12 4315 XP_006711431.1
XM_006711370.3 12 1843 XP_006711433.3
XM_006711371.3 12 1661 XP_006711434.1
XM_011509607.2 12 1701 XP_011507909.1
XM_011509608.2 13 2062 XP_011507910.1
XM_011509609.2 13 1966 XP_011507911.1
XM_017001429.1 13 1962 XP_016856918.1
XM_017001431.1 12 1884 XP_016856920.1
XM_017001434.1 12 1840 XP_016856923.1
XM_017001435.1 11 1759 XP_016856924.1
XM_017001436.1 12 1981 XP_016856925.1
XM_017001437.1 12 1978 XP_016856926.1
XM_017001438.1 12 1685 XP_016856927.1
XM_017001439.1 12 2103 XP_016856928.1
XM_017001440.1 11 1989 XP_016856929.1
XM_017001441.1 11 1962 XP_016856930.1
XM_017001442.1 11 1577 XP_016856931.1
AF147717.1 AAD31528.1
AK024751.1
AK225794.1
BC015521.1 AAH15521.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv498n100 Chr.1:192992568 - 193114915 on Build GRCh38 Loss TROVE2 UCHL5 GLRX2 SCARNA18B
nsv498699 Chr.1:193001306 - 193024494 on Build GRCh38 Loss UCHL5
nsv466994 Chr.1:192394185 - 193552205 on Build GRCh38 Loss TROVE2 MIR1278 MIR4426 B3GALT2 CDC73 LINC01031 SCARNA18B RGS13 RGS1 RGS2 UCHL5 GLRX2 LINC01032
nsv3966 Chr.1:193007327 - 193035756 on Build GRCh38 Deletion UCHL5

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
DNA recombination
transcription, DNA-templated
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
negative regulation of endopeptidase activity
protein deubiquitination
lateral ventricle development
midbrain development
forebrain morphogenesis
regulation of proteasomal protein catabolic process
thiol-dependent ubiquitin-specific protease activity
endopeptidase inhibitor activity
protein binding
poly(A) RNA binding
proteasome binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lzcpb:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0