This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02881214_cn
See other HES2 CNV Assays ›
Gene Symbol
HES2
Assay Reference Genome
Location

Chr.1:6418152 on build GRCh38
Cytoband
1p36.31
Assay ID Hs02881214_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

54626

Gene Name:

hes family bHLH transcription factor 2

Gene Aliases:

bHLHb40

Location:

Chr.1:6415232-6419919 on Build GRCh38

Assay Gene Location:

Within Exon 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
HES2 NM_019089.4 4 1342 NP_061962.2
AK023754.1 1 527
AK091122.1 4 1342
BC012091.1 AAH12091.1
BC142687.1 4 1322

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1009548 Chr.1:6342976 - 6517352 on Build GRCh38 Loss PLEKHG5 MIR4252 TNFRSF25 ACOT7 ESPN HES2
dgv2n27 Chr.1:6396412 - 6512115 on Build GRCh38 Loss PLEKHG5 MIR4252 TNFRSF25 ESPN HES2
nsv954048 Chr.1:6414241 - 6500740 on Build GRCh38 Deletion PLEKHG5 MIR4252 TNFRSF25 ESPN HES2
nsv7880 Chr.1:6417968 - 6611914 on Build GRCh38 Gain+Loss PLEKHG5 MIR4252 KLHL21 TNFRSF25 TAS1R1 NOL9 ESPN ZBTB48 HES2
nsv545240 Chr.1:6351154 - 6512115 on Build GRCh38 Loss PLEKHG5 MIR4252 TNFRSF25 ACOT7 ESPN HES2
nsv508836 Chr.1:6406896 - 6501029 on Build GRCh38 Insertion PLEKHG5 MIR4252 TNFRSF25 ESPN HES2
esv25412 Chr.1:6416568 - 6461456 on Build GRCh38 Gain+Loss MIR4252 TNFRSF25 ESPN HES2

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
negative regulation of transcription, DNA-templated
double-stranded DNA binding
transcription factor binding
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline