This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02931772_cn
See other ESRRA CNV Assays ›
Gene Symbol
ESRRA
Assay Reference Genome
Location

Chr.11:64307504 on build GRCh38
Cytoband
11q13.1
Assay ID Hs02931772_cn
Size
Availability Made To Order
Catalog # 4400291
Price 475.00
Your Price
Online offer: 441.65
441.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2101

Gene Name:

estrogen related receptor alpha

Gene Aliases:

ERR1, ERRa, ERRalpha, ESRL1, NR3B1

Location:

Chr.11:64305528-64316743 on Build GRCh38

Assay Gene Location:

Overlaps Exon 3 - Intron 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ESRRA NM_001282450.1 NP_001269379.1
NM_001282451.1 NP_001269380.1
NM_004451.4 NP_004442.3
XM_017017313.1 XP_016872802.1
BC007915.2 AAH07915.2
BC063795.1
BC073956.1 AAH73956.1
BC092470.1 AAH92470.1
BC093720.1 AAH93720.1
BC093722.1 AAH93722.1
HQ692867.1
L38487.1 AAB17015.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv951010 Chr.11:63942829 - 64352128 on Build GRCh38 Deletion TRMT112 GPR137 PLCB3 LOC105369340 KCNK4-TEX40 MIR7155 MACROD1 FKBP2 COX8A PPP1R14B TRPT1 NUDT22 FLRT1 KCNK4 CCDC88B FERMT3 ESRRA BAD VEGFB NAA40 STIP1 PRDX5 DNAJC4 TEX40 OTUB1
dgv1963n54 Chr.11:64174262 - 64329761 on Build GRCh38 Loss KCNK4 TRMT112 FERMT3 ESRRA GPR137 PLCB3 BAD LOC105369340 KCNK4-TEX40 VEGFB STIP1 PRDX5 DNAJC4 TEX40 FKBP2 PPP1R14B TRPT1 NUDT22
nsv832186 Chr.11:64204788 - 64402587 on Build GRCh38 Loss KCNK4 CCDC88B TRMT112 FERMT3 ESRRA GPR137 PLCB3 BAD LOC105369340 KCNK4-TEX40 MIR1237 VEGFB PRDX5 MIR7155 DNAJC4 TEX40 FKBP2 PPP1R14B RPS6KA4 TRPT1 NUDT22
nsv469961 Chr.11:64055468 - 64318495 on Build GRCh38 Loss KCNK4 TRMT112 FERMT3 ESRRA GPR137 PLCB3 BAD LOC105369340 KCNK4-TEX40 VEGFB STIP1 PRDX5 DNAJC4 TEX40 MACROD1 FKBP2 PPP1R14B TRPT1 NUDT22 FLRT1
nsv357 Chr.11:64284335 - 64308789 on Build GRCh38 Insertion KCNK4 TEX40 ESRRA GPR137 BAD KCNK4-TEX40

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
mitochondrion organization
intracellular receptor signaling pathway
response to estradiol
regulation of cell proliferation
steroid hormone mediated signaling pathway
regulation of osteoblast differentiation
regulation of osteoclast differentiation
positive regulation of transcription from RNA polymerase II promoter
cartilage development
positive regulation of cellular response to insulin stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
steroid binding
protein binding
zinc ion binding
protein domain specific binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline