This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02945807_cn
See other EIF4G3 CNV Assays ›
Gene Symbol
EIF4G3
Assay Reference Genome
Location

Chr.1:20807324 on build GRCh38
Cytoband
1p36.12
Assay ID Hs02945807_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

8672

Gene Name:

eukaryotic translation initiation factor 4 gamma 3

Gene Aliases:

eIF-4G 3, eIF4G 3, eIF4GII

Location:

Chr.1:20806292-21177320 on Build GRCh38

Assay Gene Location:

Within Exon 43
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
EIF4G3 NM_001198801.1 35 5483 NP_001185730.1
NM_001198802.1 35 5393 NP_001185731.1
NM_003760.4 32 5009 NP_003751.2
XM_011542356.2 38 6083 XP_011540658.1
XM_011542361.2 37 5972 XP_011540663.1
XM_011542370.1 38 6086 XP_011540672.1
XM_011542371.1 36 5828 XP_011540673.1
XM_011542374.1 35 5795 XP_011540676.1
XM_011542375.2 35 7383 XP_011540677.1
XM_011542381.2 25 3469 XP_011540683.1
XM_017002672.1 39 7621 XP_016858161.1
XM_017002673.1 37 6065 XP_016858162.1
XM_017002674.1 37 6062 XP_016858163.1
XM_017002675.1 37 6053 XP_016858164.1
XM_017002676.1 36 6032 XP_016858165.1
XM_017002677.1 36 5909 XP_016858166.1
XM_017002678.1 37 5975 XP_016858167.1
XM_017002679.1 37 5972 XP_016858168.1
XM_017002680.1 34 5002 XP_016858169.1
XM_017002681.1 37 5969 XP_016858170.1
XM_017002682.1 36 5951 XP_016858171.1
XM_017002683.1 36 5951 XP_016858172.1
XM_017002684.1 37 7477 XP_016858173.1
XM_017002685.1 36 5942 XP_016858174.1
XM_017002686.1 36 5939 XP_016858175.1
XM_017002687.1 36 5939 XP_016858176.1
XM_017002688.1 33 4968 XP_016858177.1
XM_017002689.1 37 5939 XP_016858178.1
XM_017002690.1 36 5936 XP_016858179.1
XM_017002691.1 35 5921 XP_016858180.1
XM_017002692.1 35 5918 XP_016858181.1
XM_017002693.1 34 5789 XP_016858182.1
XM_017002694.1 36 5861 XP_016858183.1
XM_017002695.1 36 5858 XP_016858184.1
XM_017002696.1 35 5840 XP_016858185.1
XM_017002697.1 35 5837 XP_016858186.1
XM_017002698.1 35 5828 XP_016858187.1
XM_017002699.1 36 5828 XP_016858188.1
XM_017002700.1 34 5807 XP_016858189.1
XM_017002701.1 34 5804 XP_016858190.1
XM_017002702.1 34 5738 XP_016858191.1
XM_017002703.1 32 4899 XP_016858192.1
XM_017002704.1 36 5951 XP_016858193.1
XM_017002705.1 37 5246 XP_016858194.1
XM_017002706.1 36 5135 XP_016858195.1
XM_017002707.1 34 5078 XP_016858196.1
XM_017002708.1 33 4967 XP_016858197.1
XM_017002709.1 34 5683 XP_016858198.1
AB209119.1 35 5352 BAD92356.1
AF012072.2 32 5009 AAC02903.2
AK294883.1
AK302873.1 23 3250
BC094683.1 30 4016 AAH94683.1
BC136643.1 32 4932
Z34918.1 CAA84397.1

Back To Top

More Information


Set Membership:

Intragenic Exonic non-DGV Variation

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
regulation of translational initiation
viral process
RNA cap binding
translation initiation factor activity
translation factor activity, RNA binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline