This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02969677_cn
See other CHD1L CNV Assays ›
Gene Symbol
CHD1L
Assay Reference Genome
Location

Chr.1:147186291 on build GRCh38
Cytoband
1q21.1
Assay ID Hs02969677_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9557

Gene Name:

chromodomain helicase DNA binding protein 1 like

Gene Aliases:

ALC1, CHDL

Location:

Chr.1:147172804-147295766 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CHD1L XM_017002858.1 XP_016858347.1

Target Gene Details

Entrez Gene ID:

2330

Gene Name:

flavin containing monooxygenase 5

Gene Aliases:

-

Location:

Chr.1:147183963-147225798 on Build GRCh38

Assay Gene Location:

Within Exon 10
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FMO5 NM_001144829.2 NP_001138301.1
NM_001144830.2 NP_001138302.1
NM_001461.3 NP_001452.2
XM_005272946.4 9 2574 XP_005273003.1
XM_005272947.4 9 2318 XP_005273004.1
XM_005272948.4 9 2301 XP_005273005.1
XM_006711245.3 8 2237 XP_006711308.1
XM_011509350.2 9 2605 XP_011507652.1
XM_011509351.2 8 2337 XP_011507653.1
XM_017000801.1 8 2349 XP_016856290.1
XM_017000802.1 8 2119 XP_016856291.1
AK222728.1 BAD96448.1
AK225739.1
BC035687.1 AAH35687.1
L37080.1 AAA67849.1
Z47553.1 CAA87633.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv327n100 Chr.1:146028214 - 148860637 on Build GRCh38 Gain RNVU1-3 PRKAB2 CH17-408M7.1 MIR6077 CHD1L ACP6 LOC101928979 NBPF13P LOC101927468 NBPF10 PDE4DIP PDIA3P1 LOC105371242 RNVU1-7 GJA5 LOC100996517 PPIAL4G RNVU1-8 GPR89B NUDT4P1 RNU1-13P LOC100132057 PDZK1P1 LOC284561 NUDT4P2 FMO5 RNVU1-1 HYDIN2 LOC728989 NBPF14 BCL9 MIR5087 LOC100505824 NBPF12 LOC105371226 LINC01138 NOTCH2NL LINC00624 RNVU1-6 GJA8 LOC100996763 NBPF11
nsv1005055 Chr.1:145642988 - 148991068 on Build GRCh38 Loss RNVU1-3 ANKRD34A PRKAB2 ACP6 ITGA10 MIR6736 PEX11B NBPF10 POLR3C PDE4DIP PDIA3P1 LOC105371242 HFE2 RNVU1-7 PPIAL4G RNVU1-8 PDZK1 GPR89B RNU1-13P PDZK1P1 LOC284561 NUDT4P2 FMO5 RNVU1-1 HYDIN2 POLR3GL NUDT17 NBPF14 GPR89A LOC105371226 GJA8 LOC100996763 NBPF11 CH17-408M7.1 MIR6077 CHD1L LOC101928979 NBPF13P LOC101927468 PIAS3 GJA5 LOC100996517 ANKRD35 NUDT4P1 LOC100132057 GNRHR2 LIX1L LOC728989 BCL9 MIR5087 LOC100505824 NBPF12 LINC01138 NOTCH2NL CD160 TXNIP LINC00624 RNVU1-6 RNF115 RBM8A
nsv1121713 Chr.1:147010816 - 147207577 on Build GRCh38 Deletion PRKAB2 LOC728989 CHD1L PDIA3P1 FMO5 NBPF13P RNVU1-8
esv2761299 Chr.1:146068341 - 148926801 on Build GRCh38 Gain+Loss RNVU1-3 PRKAB2 CH17-408M7.1 MIR6077 CHD1L ACP6 NBPF13P LOC101927468 NBPF10 PDE4DIP PDIA3P1 LOC105371242 RNVU1-7 GJA5 LOC100996517 PPIAL4G RNVU1-8 GPR89B NUDT4P1 RNU1-13P LOC100132057 PDZK1P1 LOC284561 NUDT4P2 FMO5 RNVU1-1 HYDIN2 LOC728989 NBPF14 BCL9 MIR5087 LOC100505824 NBPF12 LOC105371226 LINC01138 NOTCH2NL LINC00624 GJA8 LOC100996763 NBPF11
nsv509457 Chr.1:145948024 - 149021821 on Build GRCh38 Insertion RNVU1-3 ANKRD34A PRKAB2 CH17-408M7.1 MIR6077 CHD1L ACP6 LOC101928979 NBPF13P LOC101927468 NBPF10 PDE4DIP PDIA3P1 LOC105371242 HFE2 RNVU1-7 GJA5 LOC100996517 PPIAL4G RNVU1-8 GPR89B NUDT4P1 RNU1-13P LOC100132057 PDZK1P1 LOC284561 NUDT4P2 FMO5 RNVU1-1 LIX1L HYDIN2 POLR3GL LOC728989 NBPF14 BCL9 MIR5087 LOC100505824 NBPF12 LOC105371226 LINC01138 NOTCH2NL TXNIP LINC00624 RNVU1-6 GJA8 LOC100996763 NBPF11
nsv1132304 Chr.1:146186094 - 148977674 on Build GRCh38 Duplication RNVU1-3 PRKAB2 CH17-408M7.1 MIR6077 CHD1L ACP6 NBPF13P LOC101927468 PDE4DIP PDIA3P1 LOC105371242 RNVU1-7 GJA5 LOC100996517 PPIAL4G RNVU1-8 GPR89B NUDT4P1 RNU1-13P LOC100132057 PDZK1P1 LOC284561 NUDT4P2 FMO5 RNVU1-1 HYDIN2 LOC728989 NBPF14 BCL9 MIR5087 LOC100505824 NBPF12 LOC105371226 LINC01138 NOTCH2NL LINC00624 GJA8 LOC100996763 NBPF11
nsv1003688 Chr.1:146036817 - 148934526 on Build GRCh38 Gain RNVU1-3 PRKAB2 CH17-408M7.1 MIR6077 CHD1L ACP6 LOC101928979 NBPF13P LOC101927468 NBPF10 PDE4DIP PDIA3P1 LOC105371242 RNVU1-7 GJA5 LOC100996517 PPIAL4G RNVU1-8 GPR89B NUDT4P1 RNU1-13P LOC100132057 PDZK1P1 LOC284561 NUDT4P2 FMO5 RNVU1-1 HYDIN2 LOC728989 NBPF14 BCL9 MIR5087 LOC100505824 NBPF12 LOC105371226 LINC01138 NOTCH2NL LINC00624 RNVU1-6 GJA8 LOC100996763 NBPF11
dgv66e199 Chr.1:146055970 - 149056514 on Build GRCh38 Deletion RNVU1-3 PRKAB2 CH17-408M7.1 MIR6077 CHD1L ACP6 LOC101928979 NBPF13P LOC101927468 NBPF9 NBPF10 PDE4DIP PDIA3P1 LOC105371242 RNVU1-7 GJA5 LOC100996517 PPIAL4G RNVU1-8 GPR89B NUDT4P1 RNU1-13P LOC100132057 PDZK1P1 LOC284561 NUDT4P2 FMO5 RNVU1-1 HYDIN2 LOC728989 NBPF14 BCL9 MIR5087 LOC100505824 NBPF12 LOC105371226 LINC01138 NOTCH2NL LINC00624 GJA8 LOC100996763 NBPF11
nsv437047 Chr.1:147171976 - 147369886 on Build GRCh38 Loss PRKAB2 CHD1L PDIA3P1 FMO5

Back To Top

More Information


Set Membership:

Intragenic Exonic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
chromatin remodeling
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
global genome nucleotide-excision repair
biological_process
drug metabolic process
oxidation-reduction process
nucleotide binding
nucleic acid binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
ATPase activity
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline