This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02991152_cn
See other SLC2A4RG CNV Assays ›
Gene Symbol
SLC2A4RG
Assay Reference Genome
Location

Chr.20:63742992 on build GRCh38
Cytoband
20q13.33
Assay ID Hs02991152_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

56731

Gene Name:

SLC2A4 regulator

Gene Aliases:

GEF, HDBP-1, HDBP1, Si-1-2, Si-1-2-19

Location:

Chr.20:63739858-63744050 on Build GRCh38

Assay Gene Location:

Within Exon 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SLC2A4RG NM_020062.3 8 1221 NP_064446.2
AB044777.1 7 1112 BAE71373.1
AB044786.1 6 782 BAD29732.1
AB052777.1 8 1265 BAD29733.1
AF249267.3 8 1166 AAF97516.2
AK298304.1 6 1144
BC001402.2 7 992 AAH01402.2
BC017446.1 5 815 AAH17446.1
BC028349.2 3 416 AAH28349.2
BC052306.1 8 1266 AAH52306.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2722959 Chr.20:63715664 - 63806918 on Build GRCh38 Deletion ZGPAT SLC2A4RG LIME1 ZBTB46
nsv953308 Chr.20:63650148 - 63831847 on Build GRCh38 Deletion ZBTB46-AS1 ZGPAT RTEL1-TNFRSF6B ARFRP1 TNFRSF6B STMN3 SLC2A4RG RTEL1 LIME1 ZBTB46
nsv1160690 Chr.20:63639552 - 63744287 on Build GRCh38 Deletion ZGPAT RTEL1-TNFRSF6B ARFRP1 TNFRSF6B STMN3 SLC2A4RG RTEL1 LIME1 ZBTB46
nsv428381 Chr.20:63583375 - 64334167 on Build GRCh38 Gain+Loss MYT1 ABHD16B ARFRP1 GMEB2 UCKL1-AS1 STMN3 MIR941-2 ZBTB46 MIR647 UCKL1 ZGPAT SAMD10 RTEL1-TNFRSF6B TNFRSF6B SLC2A4RG SOX18 MIR941-3 MIR941-5 PCMTD2 LINC00266-1 MIR941-1 MIR1914 MIR6813 OPRL1 LOC100505771 LIME1 RGS19 C20orf181 ZNF512B LKAAEAR1 TPD52L2 MIR941-4 ZBTB46-AS1 DNAJC5 PRPF6 NPBWR2 LINC00176 RTEL1 TCEA2
esv7227 Chr.20:63715550 - 63807161 on Build GRCh38 Loss ZGPAT SLC2A4RG LIME1 ZBTB46
nsv1059142 Chr.20:63731978 - 63798885 on Build GRCh38 Gain ZGPAT SLC2A4RG LIME1 ZBTB46
nsv470563 Chr.20:63486715 - 63793308 on Build GRCh38 Loss FNDC11 PPDPF ARFRP1 GMEB2 STMN3 LOC100505771 LIME1 EEF1A2 HELZ2 ZBTB46 PTK6 ZGPAT RTEL1-TNFRSF6B SRMS TNFRSF6B SLC2A4RG RTEL1
dgv7684n54 Chr.20:63738644 - 63789659 on Build GRCh38 Loss SLC2A4RG LIME1 ZBTB46
esv2758806 Chr.20:63470996 - 63745014 on Build GRCh38 Gain+Loss FNDC11 PPDPF ARFRP1 KCNQ2 GMEB2 STMN3 LOC100505771 LIME1 EEF1A2 HELZ2 ZBTB46 PTK6 ZGPAT RTEL1-TNFRSF6B SRMS TNFRSF6B SLC2A4RG RTEL1
dgv7680n54 Chr.20:63737451 - 63743088 on Build GRCh38 Gain SLC2A4RG LIME1

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline