This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02997018_cn
See other NUDT22 CNV Assays ›
Gene Symbol
NUDT22
Assay Reference Genome
Location

Chr.11:64226488 on build GRCh38
Cytoband
11q13.1
Assay ID Hs02997018_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

84304

Gene Name:

nudix hydrolase 22

Gene Aliases:

-

Location:

Chr.11:64226258-64230016 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NUDT22 NM_001128612.2 NP_001122084.1
NM_001128613.2 NP_001122085.1
NM_001271831.1 NP_001258760.1
NM_032344.3 NP_115720.1
AK308555.1
BC006129.1 AAH06129.1
BG705568.1
BM045237.1
BM923060.1
HY075773.1
HY160553.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1159904 Chr.11:64150129 - 64279169 on Build GRCh38 Deletion TRPT1 FKBP2 FERMT3 DNAJC4 STIP1 PLCB3 LOC105369340 NUDT22 PPP1R14B BAD VEGFB GPR137 MACROD1
esv992196 Chr.11:64225062 - 64234883 on Build GRCh38 Deletion TRPT1 DNAJC4 NUDT22 VEGFB
nsv951010 Chr.11:63942829 - 64352128 on Build GRCh38 Deletion FLRT1 FERMT3 DNAJC4 STIP1 KCNK4 PRDX5 TEX40 LOC105369340 TRMT112 NUDT22 PPP1R14B GPR137 MACROD1 CCDC88B TRPT1 FKBP2 PLCB3 MIR7155 ESRRA COX8A NAA40 OTUB1 BAD VEGFB KCNK4-TEX40
dgv1963n54 Chr.11:64174262 - 64329761 on Build GRCh38 Loss TRPT1 FKBP2 FERMT3 DNAJC4 STIP1 PLCB3 KCNK4 ESRRA PRDX5 TEX40 LOC105369340 TRMT112 NUDT22 PPP1R14B BAD VEGFB KCNK4-TEX40 GPR137
nsv555187 Chr.11:64185649 - 64260416 on Build GRCh38 Loss TRPT1 FKBP2 FERMT3 DNAJC4 STIP1 PLCB3 NUDT22 PPP1R14B VEGFB LOC105369340
nsv555184 Chr.11:64156974 - 64275525 on Build GRCh38 Loss TRPT1 FKBP2 FERMT3 DNAJC4 STIP1 PLCB3 LOC105369340 NUDT22 PPP1R14B BAD VEGFB GPR137 MACROD1
nsv832186 Chr.11:64204788 - 64402587 on Build GRCh38 Loss CCDC88B TRPT1 FKBP2 FERMT3 DNAJC4 PLCB3 KCNK4 MIR7155 ESRRA PRDX5 TEX40 MIR1237 LOC105369340 TRMT112 NUDT22 RPS6KA4 PPP1R14B BAD VEGFB KCNK4-TEX40 GPR137
nsv469961 Chr.11:64055468 - 64318495 on Build GRCh38 Loss FLRT1 TRPT1 FKBP2 FERMT3 DNAJC4 STIP1 PLCB3 KCNK4 ESRRA PRDX5 TEX40 LOC105369340 TRMT112 NUDT22 PPP1R14B BAD VEGFB KCNK4-TEX40 GPR137 MACROD1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Process(es)

protein binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-5xwv5:80/100.66.76.242:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0