This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03035959_cn
See other HES2 CNV Assays ›
Gene Symbol
HES2
Assay Reference Genome
Location

Chr.1:6419133 on build GRCh38
Cytoband
1p36.31
Assay ID Hs03035959_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

54626

Gene Name:

hes family bHLH transcription factor 2

Gene Aliases:

bHLHb40

Location:

Chr.1:6415232-6419919 on Build GRCh38

Assay Gene Location:

Overlaps Intron 3 - Exon 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
HES2 NM_019089.4 NP_061962.2
AK091122.1
BC012091.1 AAH12091.1
BC132698.1
BC136963.1
BC142687.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1009548 Chr.1:6342976 - 6517352 on Build GRCh38 Loss ESPN HES2 TNFRSF25 ACOT7 PLEKHG5 MIR4252
dgv80n54 Chr.1:6396412 - 6512115 on Build GRCh38 Loss ESPN HES2 TNFRSF25 PLEKHG5 MIR4252
nsv954048 Chr.1:6414241 - 6500740 on Build GRCh38 Deletion ESPN HES2 TNFRSF25 PLEKHG5 MIR4252
nsv7880 Chr.1:6417968 - 6611914 on Build GRCh38 Gain+Loss ESPN HES2 TNFRSF25 ZBTB48 KLHL21 TAS1R1 PLEKHG5 MIR4252 NOL9
nsv545240 Chr.1:6351154 - 6512115 on Build GRCh38 Loss ESPN HES2 TNFRSF25 ACOT7 PLEKHG5 MIR4252
esv25412 Chr.1:6416568 - 6461456 on Build GRCh38 Gain+Loss ESPN HES2 TNFRSF25 MIR4252
nsv508836 Chr.1:6406896 - 6501029 on Build GRCh38 Insertion ESPN HES2 TNFRSF25 PLEKHG5 MIR4252

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
negative regulation of transcription, DNA-templated
double-stranded DNA binding
transcription factor binding
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0