This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03039144_cn
See other EIF4G3 CNV Assays ›
Gene Symbol
EIF4G3
Assay Reference Genome
Location

Chr.1:20849505 on build GRCh38
Cytoband
1p36.12
Assay ID Hs03039144_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

8672

Gene Name:

eukaryotic translation initiation factor 4 gamma 3

Gene Aliases:

eIF-4G 3, eIF4G 3, eIF4GII

Location:

Chr.1:20806292-21177320 on Build GRCh38

Assay Gene Location:

Overlaps Intron 34 - Exon 35
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
EIF4G3 NM_001198801.1 NP_001185730.1
NM_001198802.1 NP_001185731.1
NM_003760.4 NP_003751.2
XM_011542356.2 XP_011540658.1
XM_011542361.2 XP_011540663.1
XM_011542370.1 XP_011540672.1
XM_011542371.1 XP_011540673.1
XM_011542374.1 XP_011540676.1
XM_011542375.2 XP_011540677.1
XM_011542381.2 XP_011540683.1
XM_017002672.1 XP_016858161.1
XM_017002673.1 XP_016858162.1
XM_017002674.1 XP_016858163.1
XM_017002675.1 XP_016858164.1
XM_017002676.1 XP_016858165.1
XM_017002677.1 XP_016858166.1
XM_017002678.1 XP_016858167.1
XM_017002679.1 XP_016858168.1
XM_017002680.1 XP_016858169.1
XM_017002681.1 XP_016858170.1
XM_017002682.1 XP_016858171.1
XM_017002683.1 XP_016858172.1
XM_017002684.1 XP_016858173.1
XM_017002685.1 XP_016858174.1
XM_017002686.1 XP_016858175.1
XM_017002687.1 XP_016858176.1
XM_017002688.1 XP_016858177.1
XM_017002689.1 XP_016858178.1
XM_017002690.1 XP_016858179.1
XM_017002691.1 XP_016858180.1
XM_017002692.1 XP_016858181.1
XM_017002693.1 XP_016858182.1
XM_017002694.1 XP_016858183.1
XM_017002695.1 XP_016858184.1
XM_017002696.1 XP_016858185.1
XM_017002697.1 XP_016858186.1
XM_017002698.1 XP_016858187.1
XM_017002699.1 XP_016858188.1
XM_017002700.1 XP_016858189.1
XM_017002701.1 XP_016858190.1
XM_017002702.1 XP_016858191.1
XM_017002703.1 XP_016858192.1
XM_017002704.1 XP_016858193.1
XM_017002705.1 XP_016858194.1
XM_017002706.1 XP_016858195.1
XM_017002707.1 XP_016858196.1
XM_017002708.1 XP_016858197.1
XM_017002709.1 XP_016858198.1
AB209119.1 BAD92356.1
AF012072.2 AAC02903.2
AK294883.1
AK302873.1
BC094683.1 AAH94683.1
BC136643.1
Z34918.1 CAA84397.1

Back To Top

More Information


Set Membership:

Intragenic Non-exonic non-DGV Variation

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
regulation of translational initiation
viral process
RNA cap binding
translation initiation factor activity
translation factor activity, RNA binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0