Assay Details
Target Gene Details
Entrez Gene ID: | 104564225 |
Gene Name: | myosin heavy chain-associated RNA transcript |
Gene Aliases: |
Myheart |
Location: |
Chr.14:23415450-23417595 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MHRT | NR_126491.1 |
Target Gene Details
Entrez Gene ID: | 4625 |
Gene Name: | myosin, heavy chain 7, cardiac muscle, beta |
Gene Aliases: |
CMD1S, CMH1, MPD1, MYHCB, SPMD, SPMM |
Location: |
Chr.14:23412738-23435686 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 31 - Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYH7 | NM_000257.3 | NP_000248.2 | ||
| XM_017021340.1 | XP_016876829.1 | |||
| AB209708.1 | BAD92945.1 | |||
| AY518538.1 | AAT44534.1 | |||
| BC112171.1 | AAI12172.1 | |||
| BC112173.1 | AAI12174.1 | |||
| DQ248310.1 | ABB55265.1 | |||
| EF560725.1 | ||||
| EU747717.1 | ||||
| M17712.1 | AAA36343.1 | |||
| M21665.1 | AAA36345.1 | |||
| M58018.1 | AAA62830.1 | |||
| X03741.1 | CAA27381.1 | |||
| X06976.1 | CAA30039.1 | |||
| X51591.1 | CAA35940.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470623 | Chr.14:23391499 - 23418656 on Build GRCh38 | Gain |
|
| esv2748480 | Chr.14:23390535 - 23420438 on Build GRCh38 | Deletion |
|
| nsv456166 | Chr.14:23392602 - 23418656 on Build GRCh38 | Gain |
|
| esv3584149 | Chr.14:23395407 - 23427942 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map