This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03061653_cn
See other GNB1 CNV Assays ›
Gene Symbol
GNB1
Assay Reference Genome
Location

Chr.1:1786966 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03061653_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2782

Gene Name:

G protein subunit beta 1

Gene Aliases:

MRD42

Location:

Chr.1:1785285-1891117 on Build GRCh38

Assay Gene Location:

Within Exon 13
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GNB1 NM_001282538.1 10 1367 NP_001269467.1
NM_001282539.1 11 1464 NP_001269468.1
NM_002074.4 12 1513 NP_002065.1
XM_017001059.1 11 1417 XP_016856548.1
XM_017001060.1 12 1283 XP_016856549.1
XM_017001061.1 11 1456 XP_016856550.1
AF070597.1 4 482 AAC28652.1
AF070603.1 6 721 AAC28655.1
AK122949.1 10 1151
BC004186.2 12 1396 AAH04186.1
BC005888.2 10 1164 AAH05888.1
BC008991.1 12 1452 AAH08991.1
M36430.1 6 841 AAA63265.1
X04526.1 12 1400 CAA28207.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion SLC35E2 SLC35E2B MMP23A MMP23B MIB2 NADK CDK11A FNDC10 CDK11B GNB1
nsv834091 Chr.1:1701359 - 1880638 on Build GRCh38 Loss SLC35E2 MMP23A NADK CDK11A GNB1
nsv951380 Chr.1:1785162 - 1791261 on Build GRCh38 Deletion GNB1
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication SLC35E2 SLC35E2B MMP23A MMP23B MIB2 NADK SSU72 CDK11A FNDC10 CDK11B GNB1
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss MIR6859-2 LOC100133331 FAM132A FAM138A CPSF3L TNFRSF18 MIR1302-2 KLHL17 SLC35E2 LOC107985728 TMEM52 ISG15 ATAD3A PLEKHN1 TMEM88B TTLL10 UBE2J2 OR4F5 DVL1 MIR6727 LINC01342 TNFRSF4 LOC105378948 TMEM240 LOC102725121 SLC35E2B MIR6723 LOC100132287 LINC01128 LOC107985729 LOC105378947 SDF4 AGRN ATAD3C MIB2 AURKAIP1 RNF223 NOC2L LOC100288175 PRKCZ MRPL20 MIR200B SKI CDK11A LOC100288069 MIR6726 SAMD11 MTND2P28 WASH7P CPTP LOC100130417 LOC105378592 LOC100506504 LINC00115 LOC284600 LOC105378591 LOC102724312 FAM87B LOC100287934 MIR429 MIR6859-1 MORN1 FNDC10 OR4F29 FAM41C FAAP20 ANKRD65 NADK VWA1 MXRA8 LOC107984841 LOC729737 MTND1P23 CFAP74 LOC100134822 TTLL10-AS1 LOC105378589 CALML6 GABRD TAS1R3 C1orf159 B3GALT6 PERM1 MIR6808 CDK11B OR4F16 LOC148413 PUSL1 MMP23B CCNL2 HES4 SSU72 GNB1 LOC101928626 MIR200A SCNN1D LOC105378949 MMP23A ATAD3B ACAP3 DDX11L1 LOC100129534
nsv1160926 Chr.1:1772699 - 1905488 on Build GRCh38 Duplication LOC105378949 NADK GNB1
dgv10n100 Chr.1:1779975 - 1884542 on Build GRCh38 Gain NADK GNB1
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss SLC35E2B LOC105378591 LOC102724312 GABRD SLC35E2 LOC107985729 ATAD3C TMEM52 MIB2 ATAD3A FNDC10 CDK11B TMEM88B PRKCZ MRPL20 LOC148413 MMP23B ANKRD65 CCNL2 NADK SSU72 CDK11A VWA1 GNB1 LOC105378949 CFAP74 MMP23A ATAD3B LOC105378589 TMEM240 CALML6

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

protein folding
signal transduction
adenylate cyclase-activating dopamine receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
G-protein coupled acetylcholine receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
Ras protein signal transduction
cell proliferation
cardiac muscle cell apoptotic process
rhodopsin mediated signaling pathway
platelet activation
sensory perception of taste
retina development in camera-type eye
cellular response to glucagon stimulus
cellular response to prostaglandin E stimulus
cellular response to hypoxia
cellular response to catecholamine stimulus
GTPase activity
signal transducer activity
protein binding
spectrin binding
type 1 angiotensin receptor binding
protein complex binding
GTPase binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fbxsq:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline