This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03327894_cn
See other CHD1L CNV Assays ›
Gene Symbol
CHD1L
Assay Reference Genome
Location

Chr.1:147174692 on build GRCh38
Cytoband
1q21.1
Assay ID Hs03327894_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,505.00
Your Price
Online offer:
4505.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9557

Gene Name:

chromodomain helicase DNA binding protein 1 like

Gene Aliases:

ALC1, CHDL

Location:

Chr.1:147172804-147295766 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CHD1L XM_017002858.1 XP_016858347.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv327n100 Chr.1:146028214 - 148860637 on Build GRCh38 Gain NBPF11 MIR5087 PDZK1P1 MIR6077 NUDT4P2 NBPF14 LOC100996517 NBPF10 ACP6 HYDIN2 LOC284561 CH17-408M7.1 NOTCH2NL LOC105371226 PDIA3P1 RNVU1-7 RNVU1-8 RNVU1-6 RNVU1-1 RNVU1-3 LINC00624 NBPF12 FMO5 LOC100132057 RNU1-13P LOC728989 CHD1L PRKAB2 BCL9 GJA8 LOC101928979 GPR89B LOC100505824 PPIAL4G GJA5 PDE4DIP LOC105371242 LOC101927468 NBPF13P LOC100996763 NUDT4P1 LINC01138
nsv1005055 Chr.1:145642988 - 148991068 on Build GRCh38 Loss NBPF11 ANKRD35 LIX1L MIR6077 NUDT4P2 PEX11B ITGA10 POLR3C LOC100996517 RBM8A NBPF10 ACP6 CD160 LOC284561 CH17-408M7.1 NOTCH2NL PDIA3P1 RNVU1-8 LINC00624 NBPF12 LOC100132057 RNU1-13P PRKAB2 BCL9 GJA8 LOC101928979 GPR89B GJA5 LOC105371242 MIR6736 NBPF13P PDZK1 MIR5087 PDZK1P1 NBPF14 NUDT17 HYDIN2 LOC105371226 HFE2 RNVU1-7 GPR89A RNVU1-6 RNVU1-1 POLR3GL RNVU1-3 RNF115 GNRHR2 FMO5 ANKRD34A LOC728989 CHD1L LOC100505824 PPIAL4G PDE4DIP PIAS3 LOC101927468 TXNIP LOC100996763 NUDT4P1 LINC01138
nsv1121713 Chr.1:147010816 - 147207577 on Build GRCh38 Deletion FMO5 PDIA3P1 LOC728989 NBPF13P CHD1L PRKAB2 RNVU1-8
esv2761299 Chr.1:146068341 - 148926801 on Build GRCh38 Gain+Loss NBPF11 MIR5087 PDZK1P1 MIR6077 NUDT4P2 NBPF14 LOC100996517 NBPF10 ACP6 HYDIN2 LOC284561 CH17-408M7.1 NOTCH2NL LOC105371226 PDIA3P1 RNVU1-7 RNVU1-8 RNVU1-1 RNVU1-3 LINC00624 NBPF12 FMO5 LOC100132057 RNU1-13P LOC728989 CHD1L PRKAB2 BCL9 GJA8 GPR89B LOC100505824 PPIAL4G GJA5 PDE4DIP LOC105371242 LOC101927468 NBPF13P LOC100996763 NUDT4P1 LINC01138
nsv509457 Chr.1:145948024 - 149021821 on Build GRCh38 Insertion NBPF11 LIX1L MIR5087 PDZK1P1 MIR6077 NUDT4P2 NBPF14 LOC100996517 NBPF10 ACP6 HYDIN2 LOC284561 CH17-408M7.1 NOTCH2NL LOC105371226 PDIA3P1 HFE2 RNVU1-7 RNVU1-8 RNVU1-6 RNVU1-1 POLR3GL RNVU1-3 LINC00624 NBPF12 FMO5 ANKRD34A LOC100132057 RNU1-13P LOC728989 CHD1L PRKAB2 BCL9 GJA8 LOC101928979 GPR89B LOC100505824 PPIAL4G GJA5 PDE4DIP LOC105371242 LOC101927468 NBPF13P TXNIP LOC100996763 NUDT4P1 LINC01138
nsv1132304 Chr.1:146186094 - 148977674 on Build GRCh38 Duplication NBPF11 MIR5087 PDZK1P1 MIR6077 NUDT4P2 NBPF14 LOC100996517 ACP6 HYDIN2 LOC284561 CH17-408M7.1 NOTCH2NL LOC105371226 PDIA3P1 RNVU1-7 RNVU1-8 RNVU1-1 RNVU1-3 LINC00624 NBPF12 FMO5 LOC100132057 RNU1-13P LOC728989 CHD1L PRKAB2 BCL9 GJA8 GPR89B LOC100505824 PPIAL4G GJA5 PDE4DIP LOC105371242 LOC101927468 NBPF13P LOC100996763 NUDT4P1 LINC01138
nsv1003688 Chr.1:146036817 - 148934526 on Build GRCh38 Gain NBPF11 MIR5087 PDZK1P1 MIR6077 NUDT4P2 NBPF14 LOC100996517 NBPF10 ACP6 HYDIN2 LOC284561 CH17-408M7.1 NOTCH2NL LOC105371226 PDIA3P1 RNVU1-7 RNVU1-8 RNVU1-6 RNVU1-1 RNVU1-3 LINC00624 NBPF12 FMO5 LOC100132057 RNU1-13P LOC728989 CHD1L PRKAB2 BCL9 GJA8 LOC101928979 GPR89B LOC100505824 PPIAL4G GJA5 PDE4DIP LOC105371242 LOC101927468 NBPF13P LOC100996763 NUDT4P1 LINC01138
dgv66e199 Chr.1:146055970 - 149056514 on Build GRCh38 Deletion NBPF11 NBPF9 MIR5087 PDZK1P1 MIR6077 NUDT4P2 NBPF14 LOC100996517 NBPF10 ACP6 HYDIN2 LOC284561 CH17-408M7.1 NOTCH2NL LOC105371226 PDIA3P1 RNVU1-7 RNVU1-8 RNVU1-1 RNVU1-3 LINC00624 NBPF12 FMO5 LOC100132057 RNU1-13P LOC728989 CHD1L PRKAB2 BCL9 GJA8 LOC101928979 GPR89B LOC100505824 PPIAL4G GJA5 PDE4DIP LOC105371242 LOC101927468 NBPF13P LOC100996763 NUDT4P1 LINC01138
nsv437047 Chr.1:147171976 - 147369886 on Build GRCh38 Loss FMO5 PDIA3P1 CHD1L PRKAB2

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
chromatin remodeling
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
global genome nucleotide-excision repair
nucleotide binding
nucleic acid binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
ATPase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0