This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03328706_cn
See other H6PD CNV Assays ›
Gene Symbol
H6PD
Assay Reference Genome
Location

Chr.1:9255865 on build GRCh38
Cytoband
1p36.22
Assay ID Hs03328706_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9563

Gene Name:

hexose-6-phosphate dehydrogenase/glucose 1-dehydrogenase

Gene Aliases:

CORTRD1, G6PDH, GDH

Location:

Chr.1:9234767-9271337 on Build GRCh38

Assay Gene Location:

Within Intron 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
H6PD NM_001282587.1 NP_001269516.1
NM_004285.3 NP_004276.2
XM_005263540.4 XP_005263597.1
XM_006711052.3 XP_006711115.1
XM_017002865.1 XP_016858354.1
XM_017002866.1 XP_016858355.1
AJ012590.1 CAA10071.1
BC081559.1 AAH81559.1
CR749282.1 CAH18137.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv2e55 Chr.1:9255788 - 9344634 on Build GRCh38 Gain SPSB1 H6PD
dgv16n100 Chr.1:9237601 - 9351410 on Build GRCh38 Gain SPSB1 H6PD
nsv832092 Chr.1:9126509 - 9304543 on Build GRCh38 Loss SPSB1 MIR34AHG MIR34A GPR157 H6PD LOC102724571
dgv97n54 Chr.1:9249687 - 9352728 on Build GRCh38 Gain SPSB1 H6PD
dgv12e212 Chr.1:9243684 - 9344908 on Build GRCh38 Gain SPSB1 H6PD
esv2763409 Chr.1:9247978 - 9344958 on Build GRCh38 Gain SPSB1 H6PD

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

glucose metabolic process
pentose-phosphate shunt
oxidation-reduction process
glucose-6-phosphate dehydrogenase activity
6-phosphogluconolactonase activity
glucose 1-dehydrogenase [NAD(P)] activity
NADP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0