This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03329774_cn
See other SLC35E2B CNV Assays ›
Gene Symbol
SLC35E2B
Assay Reference Genome
Location

Chr.1:1671919 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03329774_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

728661

Gene Name:

solute carrier family 35 member E2B

Gene Aliases:

SLC35E2

Location:

Chr.1:1661478-1692804 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SLC35E2B NM_001110781.2 NP_001104251.1
NM_001290264.1 NP_001277193.1
XM_011542072.1 XP_011540374.1
AB007916.3
AK125619.1
AK307886.1
BC110653.1 AAI10654.1
CR936618.1 CAI56761.1
DA794179.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv37n54 Chr.1:1663759 - 1696548 on Build GRCh38 Loss SLC35E2B
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion MMP23B GNB1 CDK11A MIB2 SLC35E2 NADK MMP23A CDK11B FNDC10 SLC35E2B
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss MMP23B CDK11A MIB2 SLC35E2 MMP23A CDK11B FNDC10 SSU72 SLC35E2B
esv3575582 Chr.1:1624086 - 1721336 on Build GRCh38 Gain MMP23B CDK11A MIB2 MMP23A CDK11B SLC35E2B
esv3585010 Chr.1:1670280 - 1751138 on Build GRCh38 Loss CDK11A SLC35E2 MMP23A SLC35E2B
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication MMP23B GNB1 CDK11A MIB2 SLC35E2 NADK MMP23A CDK11B FNDC10 SSU72 SLC35E2B
dgv2e199 Chr.1:1653096 - 1721890 on Build GRCh38 Deletion CDK11A MMP23A CDK11B SLC35E2B
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss PLEKHN1 LOC105378947 DDX11L1 SSU72 LOC101928626 AGRN FAM87B CDK11A MIR200A LINC00115 FAM41C C1orf159 VWA1 LOC100134822 TTLL10-AS1 LOC107985729 TMEM240 DVL1 MIR6726 MTND2P28 FAM132A LOC284600 GNB1 MIR429 LOC107984841 MMP23A SDF4 LOC100287934 LINC01128 FAM138A ATAD3B LOC102725121 SLC35E2 CCNL2 LOC100129534 MORN1 LINC01342 HES4 SLC35E2B MXRA8 PRKCZ ISG15 MIR6727 ATAD3C LOC105378948 AURKAIP1 MIR6859-1 TAS1R3 OR4F5 UBE2J2 MTND1P23 MIR1302-2 LOC105378949 NOC2L WASH7P OR4F29 LOC100132287 LOC105378591 TTLL10 LOC105378589 CPSF3L TMEM52 PUSL1 LOC100288069 SAMD11 CDK11B MIR6859-2 ACAP3 TMEM88B LOC100288175 MMP23B LOC105378592 OR4F16 LOC102724312 LOC100133331 TNFRSF4 LOC100506504 MIR6723 LOC729737 CPTP ANKRD65 LOC100130417 TNFRSF18 NADK RNF223 KLHL17 FAAP20 PERM1 B3GALT6 FNDC10 MIR6808 MIR200B CFAP74 SCNN1D LOC148413 MRPL20 MIB2 GABRD SKI CALML6 LOC107985728 ATAD3A
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss MMP23B CDK11A MIB2 SLC35E2 NADK MMP23A CDK11B FNDC10 SSU72 SLC35E2B
esv3585008 Chr.1:1663929 - 1681002 on Build GRCh38 Loss SLC35E2B
nsv945745 Chr.1:1671930 - 1674224 on Build GRCh38 Duplication SLC35E2B
esv3585007 Chr.1:1647187 - 1714507 on Build GRCh38 Loss CDK11A MMP23A CDK11B SLC35E2B
nsv950516 Chr.1:1663141 - 1715961 on Build GRCh38 Deletion CDK11A MMP23A SLC35E2B
esv27786 Chr.1:1632502 - 1752267 on Build GRCh38 Gain MMP23B CDK11A SLC35E2 NADK MMP23A CDK11B SLC35E2B
nsv545022 Chr.1:1665581 - 1683909 on Build GRCh38 Loss SLC35E2B
nsv545026 Chr.1:1671348 - 1706566 on Build GRCh38 Loss CDK11A MMP23A SLC35E2B
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss TAS1R3 PLEKHN1 UBE2J2 SSU72 AGRN FAM87B NOC2L CDK11A MIR200A LINC00115 FAM41C TTLL10 C1orf159 VWA1 CPSF3L PUSL1 TTLL10-AS1 LOC107985729 TMEM240 DVL1 MIR6726 LOC100288069 SAMD11 FAM132A LOC284600 MIR429 MMP23A CDK11B ACAP3 TMEM88B SDF4 LOC100288175 LOC100287934 MMP23B LINC01128 ATAD3B SLC35E2 LOC102724312 CCNL2 TNFRSF4 CPTP ANKRD65 LOC100130417 TNFRSF18 LINC01342 NADK RNF223 HES4 KLHL17 SLC35E2B MXRA8 PERM1 B3GALT6 ISG15 FNDC10 MIR6727 ATAD3C MIR6808 MIR200B SCNN1D LOC148413 LOC105378948 MRPL20 MIB2 LOC107985728 ATAD3A AURKAIP1
nsv545018 Chr.1:1638499 - 1688102 on Build GRCh38 Loss CDK11B SLC35E2B
dgv3n67 Chr.1:1653920 - 1735718 on Build GRCh38 Gain CDK11A SLC35E2 MMP23A CDK11B SLC35E2B
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss MMP23B ATAD3B SLC35E2 SSU72 LOC102724312 CCNL2 LOC105378949 ANKRD65 CDK11A NADK LOC105378591 VWA1 SLC35E2B LOC105378589 TMEM52 LOC107985729 PRKCZ TMEM240 FNDC10 ATAD3C CFAP74 GNB1 LOC148413 MRPL20 MIB2 GABRD MMP23A CDK11B CALML6 TMEM88B ATAD3A

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

secondary carrier transporter

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0