This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03331012_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1265024 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03331012_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Exon 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 1 408 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss UBE2J2 SCNN1D ACAP3 MIR6726
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication FAM132A CPSF3L TTLL10 PUSL1 TTLL10-AS1 MIR429 MIR6726 AGRN CPTP UBE2J2 B3GALT6 MIR6727 ACAP3 LOC100288175 C1orf159 LINC01342 MIR200A TNFRSF4 TNFRSF18 SDF4 TAS1R3 SCNN1D MIR200B LOC105378948 RNF223
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss MMP23B ATAD3C SAMD11 LINC00115 TMEM240 KLHL17 LOC284600 ATAD3B LOC100287934 CCNL2 LOC107985729 LOC100288069 TTLL10-AS1 MMP23A MIR429 MIB2 SLC35E2 AGRN CPTP UBE2J2 B3GALT6 TMEM88B FAM87B C1orf159 LOC102724312 LINC01342 PLEKHN1 MRPL20 TNFRSF18 LOC100130417 TAS1R3 MIR200B DVL1 AURKAIP1 CDK11A VWA1 FAM132A SSU72 CPSF3L LOC148413 LOC107985728 ATAD3A TTLL10 PUSL1 ISG15 LINC01128 MIR6726 FNDC10 HES4 MIR6727 ACAP3 MIR6808 MXRA8 ANKRD65 LOC100288175 CDK11B SLC35E2B MIR200A NOC2L TNFRSF4 NADK PERM1 FAM41C SDF4 SCNN1D LOC105378948 RNF223
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss MMP23B TMEM240 LOC100134822 CFAP74 LOC100287934 CCNL2 TTLL10-AS1 MIR429 LOC107984841 MIB2 LOC102725121 LOC105378947 PLEKHN1 MRPL20 TNFRSF18 TAS1R3 DVL1 AURKAIP1 VWA1 SSU72 LOC105378592 MIR6723 CPSF3L MIR1302-2 ATAD3A TTLL10 MIR6726 MIR6859-1 MIR6727 LOC105378589 MIR6808 MXRA8 ANKRD65 CALML6 FAM138A LOC100506504 CDK11B SLC35E2B MIR200A NOC2L PRKCZ PERM1 FAM41C SDF4 LOC105378948 RNF223 WASH7P ATAD3C SAMD11 GNB1 LINC00115 KLHL17 LOC284600 ATAD3B LOC729737 LOC100132287 LOC107985729 LOC100288069 OR4F16 MMP23A SLC35E2 AGRN MTND2P28 CPTP UBE2J2 LOC105378591 B3GALT6 TMEM88B FAM87B C1orf159 LOC102724312 LINC01342 MORN1 TMEM52 LOC100130417 MIR200B CDK11A FAAP20 OR4F29 LOC100133331 FAM132A MTND1P23 MIR6859-2 GABRD LOC148413 LOC107985728 PUSL1 ISG15 LINC01128 LOC101928626 SKI DDX11L1 FNDC10 HES4 OR4F5 LOC105378949 ACAP3 LOC100288175 TNFRSF4 NADK LOC100129534 SCNN1D
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss FAM132A UBE2J2 B3GALT6 SDF4 SCNN1D ACAP3 MIR6726
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss FAM132A SAMD11 LINC00115 KLHL17 LOC107985728 LOC284600 TTLL10 ISG15 LINC01128 TTLL10-AS1 MIR429 AGRN HES4 UBE2J2 B3GALT6 FAM87B LOC100288175 C1orf159 LINC01342 MIR200A PLEKHN1 NOC2L TNFRSF4 TNFRSF18 LOC100130417 PERM1 FAM41C SDF4 MIR200B LOC105378948 RNF223
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion VWA1 FAM132A SSU72 MMP23B ATAD3C TMEM240 CPSF3L LOC148413 ATAD3A TTLL10 PUSL1 ATAD3B CCNL2 LOC107985729 TTLL10-AS1 MIR429 MIR6726 MIB2 FNDC10 CPTP UBE2J2 B3GALT6 TMEM88B MIR6727 ACAP3 MIR6808 MXRA8 ANKRD65 C1orf159 CDK11B LOC102724312 LINC01342 MIR200A TNFRSF4 MRPL20 TNFRSF18 SDF4 TAS1R3 SCNN1D MIR200B DVL1 AURKAIP1
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss CPTP UBE2J2 MIR6727 ACAP3 MIR6808 MXRA8 CPSF3L PUSL1 TAS1R3 SCNN1D DVL1 AURKAIP1 MIR6726
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain FAM132A SAMD11 KLHL17 CPSF3L LOC148413 LOC107985728 LOC284600 TTLL10 PUSL1 CCNL2 ISG15 TTLL10-AS1 MIR429 MIR6726 AGRN HES4 CPTP UBE2J2 B3GALT6 TMEM88B MIR6727 ACAP3 MIR6808 MXRA8 ANKRD65 LOC100288175 C1orf159 LINC01342 MIR200A PLEKHN1 NOC2L TNFRSF4 MRPL20 TNFRSF18 LOC100130417 PERM1 SDF4 TAS1R3 SCNN1D MIR200B LOC105378948 DVL1 AURKAIP1 RNF223
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain FAM132A UBE2J2 B3GALT6 C1orf159 LINC01342 MIR200A TNFRSF4 TNFRSF18 TTLL10 SDF4 SCNN1D MIR200B LOC105378948 TTLL10-AS1 MIR429 RNF223
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss FAM132A UBE2J2 B3GALT6 ACAP3 C1orf159 LINC01342 MIR200A TNFRSF4 TNFRSF18 TTLL10 SDF4 SCNN1D MIR200B TTLL10-AS1 MIR429 RNF223 MIR6726
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain FAM132A UBE2J2 B3GALT6 MIR6727 ACAP3 MIR200A CPSF3L TNFRSF4 TNFRSF18 TTLL10 PUSL1 SDF4 SCNN1D MIR200B TTLL10-AS1 MIR429 MIR6726

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-65nqn:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline