Assay Details
Target Gene Details
Entrez Gene ID: | 284498 |
Gene Name: | chromosome 1 open reading frame 167 |
Gene Aliases: |
- |
Location: |
Chr.1:11762188-11789585 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf167 | NM_001010881.1 | NP_001010881.1 | ||
| XM_011541269.2 | XP_011539571.2 | |||
| XM_011541271.2 | XP_011539573.2 | |||
| XM_011541272.2 | XP_011539574.1 | |||
| XM_011541274.2 | XP_011539576.2 | |||
| XM_011541275.2 | XP_011539577.2 | |||
| XM_011541276.2 | XP_011539578.1 | |||
| XM_011541277.2 | XP_011539579.1 | |||
| XM_011541278.2 | XP_011539580.1 | |||
| XM_017001062.1 | XP_016856551.1 | |||
| XM_017001063.1 | XP_016856552.1 | |||
| XM_017001064.1 | XP_016856553.1 | |||
| AL833920.1 | CAD38776.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832758 | Chr.1:11637026 - 11777087 on Build GRCh38 | Loss |
|
| nsv508880 | Chr.1:11721383 - 11771263 on Build GRCh38 | Insertion |
|

Genomic Map