This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03333427_cn
See other SAMD11 CNV Assays ›
Gene Symbol
SAMD11
Assay Reference Genome
Location

Chr.1:940959 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03333427_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

148398

Gene Name:

sterile alpha motif domain containing 11

Gene Aliases:

MRS

Location:

Chr.1:925741-944581 on Build GRCh38

Assay Gene Location:

Within Intron 7
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SAMD11 NM_152486.2 NP_689699.2
AK054643.1 BAB70781.1
BC024295.2 AAH24295.1
BC033213.1 AAH33213.1
CN293867.1
JX093066.1
JX093067.1
JX093068.1
JX093069.1
JX093070.1
JX093071.1
JX093072.1
JX093073.1
JX093074.1
JX093075.1
JX093076.1
JX093077.1
JX093078.1
JX093079.1
JX093080.1
JX093081.1
JX093082.1
JX093083.1
JX093084.1
JX093085.1
JX093086.1
JX093087.1
JX093088.1
JX093089.1
JX093090.1
JX093091.1
JX093092.1
JX093093.1
JX093094.1
JX093095.1
JX093096.1
JX093097.1
JX093098.1
JX093099.1
JX093100.1
JX093101.1
JX093102.1
JX093103.1
JX093104.1
JX093105.1
JX093106.1
JX093107.1
JX093108.1
JX093109.1
JX093110.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv15n54 Chr.1:934063 - 943937 on Build GRCh38 Loss SAMD11
esv2762302 Chr.1:914637 - 1106320 on Build GRCh38 Gain LOC284600 LOC100288175 NOC2L ISG15 LOC105378948 C1orf159 PLEKHN1 KLHL17 LOC107985728 PERM1 HES4 AGRN SAMD11 LOC100130417 RNF223
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss SLC35E2 LOC101928626 LOC105378949 NADK LOC105378948 ATAD3B MIR6808 TNFRSF18 FAM138A CALML6 ATAD3C AURKAIP1 CDK11A SDF4 LOC100288175 FAAP20 MTND2P28 PLEKHN1 MRPL20 CFAP74 LOC100506504 LOC100134822 CPSF3L MIR6727 LOC107984841 TMEM240 SKI TTLL10 LOC148413 FAM41C DDX11L1 B3GALT6 TMEM88B MXRA8 LOC102724312 MIR6859-1 OR4F29 LOC100132287 PERM1 MTND1P23 MIR6723 LOC102725121 LOC284600 OR4F16 MIB2 LOC105378592 SSU72 SLC35E2B ANKRD65 LOC100287934 ATAD3A TTLL10-AS1 RNF223 ACAP3 LOC100133331 TAS1R3 TMEM52 NOC2L FNDC10 LOC105378947 MIR6859-2 PRKCZ MIR429 SAMD11 FAM132A FAM87B MIR1302-2 LOC729737 CPTP CDK11B GABRD TNFRSF4 C1orf159 WASH7P MIR200A MIR6726 KLHL17 LOC107985728 DVL1 MMP23A SCNN1D LOC107985729 LOC100129534 LINC01342 LOC100288069 MIR200B OR4F5 AGRN PUSL1 LINC00115 MMP23B ISG15 LOC105378589 VWA1 CCNL2 GNB1 HES4 LOC100130417 UBE2J2 LOC105378591 LINC01128 MORN1
nsv950451 Chr.1:899421 - 1094520 on Build GRCh38 Deletion LOC284600 LOC100288175 NOC2L ISG15 LOC105378948 C1orf159 PLEKHN1 KLHL17 LOC107985728 PERM1 HES4 AGRN SAMD11 LOC100130417 RNF223
nsv509035 Chr.1:891406 - 985724 on Build GRCh38 Insertion LOC284600 KLHL17 LOC107985728 PERM1 SAMD11 LOC100130417 NOC2L PLEKHN1
dgv8n54 Chr.1:925754 - 943937 on Build GRCh38 Loss SAMD11
nsv428334 Chr.1:874371 - 1220569 on Build GRCh38 Gain FAM41C LINC01342 NOC2L MIR200B LOC105378948 MIR429 PERM1 AGRN SAMD11 TNFRSF18 LOC284600 SDF4 LOC100288175 ISG15 TNFRSF4 C1orf159 PLEKHN1 MIR200A KLHL17 LOC107985728 HES4 LOC100130417 TTLL10-AS1 RNF223 TTLL10
dgv5n100 Chr.1:585989 - 1114424 on Build GRCh38 Gain FAM41C LOC100133331 LOC101928626 LOC100288069 NOC2L LOC105378948 LOC105378947 PERM1 AGRN SAMD11 MTND1P23 MIR6723 FAM87B LOC284600 OR4F16 LINC00115 LOC100288175 MTND2P28 ISG15 C1orf159 PLEKHN1 KLHL17 LOC100287934 LOC107985728 HES4 LOC100130417 LOC107984841 RNF223 LINC01128
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss SLC35E2 TAS1R3 NOC2L NADK LOC105378948 FNDC10 ATAD3B MIR6808 MIR429 SAMD11 TNFRSF18 FAM132A ATAD3C FAM87B AURKAIP1 CDK11A SDF4 CPTP LOC100288175 CDK11B TNFRSF4 C1orf159 PLEKHN1 MIR200A MIR6726 MRPL20 KLHL17 LOC107985728 CPSF3L MIR6727 DVL1 MMP23A TMEM240 SCNN1D TTLL10 LOC148413 LOC107985729 FAM41C B3GALT6 TMEM88B LINC01342 LOC100288069 MXRA8 MIR200B LOC102724312 PERM1 AGRN PUSL1 LOC284600 LINC00115 MIB2 MMP23B SSU72 ISG15 SLC35E2B VWA1 ANKRD65 CCNL2 LOC100287934 ATAD3A HES4 LOC100130417 UBE2J2 TTLL10-AS1 RNF223 LINC01128 ACAP3
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss FAM41C B3GALT6 LINC01342 NOC2L MIR200B LOC105378948 MIR429 PERM1 AGRN SAMD11 TNFRSF18 FAM132A FAM87B LOC284600 LINC00115 SDF4 LOC100288175 ISG15 TNFRSF4 C1orf159 PLEKHN1 MIR200A KLHL17 LOC107985728 HES4 LOC100130417 UBE2J2 TTLL10-AS1 RNF223 TTLL10 LINC01128
nsv1013524 Chr.1:939510 - 1161955 on Build GRCh38 Gain LOC100288175 LINC01342 NOC2L ISG15 LOC105378948 C1orf159 PLEKHN1 KLHL17 PERM1 HES4 AGRN SAMD11 RNF223
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain B3GALT6 TAS1R3 TMEM88B LINC01342 NOC2L MXRA8 MIR200B LOC105378948 MIR6808 MIR429 PERM1 AGRN PUSL1 SAMD11 TNFRSF18 FAM132A LOC284600 AURKAIP1 SDF4 CPTP LOC100288175 ISG15 TNFRSF4 C1orf159 PLEKHN1 MIR200A MIR6726 ANKRD65 MRPL20 KLHL17 CCNL2 LOC107985728 CPSF3L HES4 MIR6727 DVL1 LOC100130417 UBE2J2 TTLL10-AS1 SCNN1D RNF223 TTLL10 ACAP3 LOC148413
nsv544895 Chr.1:917392 - 1054900 on Build GRCh38 Loss LOC284600 KLHL17 LOC107985728 PERM1 HES4 AGRN SAMD11 LOC100130417 NOC2L ISG15 PLEKHN1
nsv1160644 Chr.1:939652 - 1220663 on Build GRCh38 Deletion SDF4 LOC100288175 LINC01342 NOC2L ISG15 MIR200B LOC105378948 TNFRSF4 C1orf159 PLEKHN1 MIR200A KLHL17 MIR429 PERM1 HES4 AGRN SAMD11 TTLL10-AS1 TNFRSF18 RNF223 TTLL10
nsv832980 Chr.1:848279 - 1007037 on Build GRCh38 Gain FAM41C LOC284600 KLHL17 LOC107985728 PERM1 HES4 SAMD11 LOC100130417 NOC2L LINC01128 PLEKHN1
nsv544898 Chr.1:924665 - 943937 on Build GRCh38 Gain LOC107985728 SAMD11

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0