This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03335104_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1268647 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03335104_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_017000238.1 XP_016855727.1
AF296658.1 AAK52609.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM794599.1
BM926567.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss SCNN1D UBE2J2 MIR6726 ACAP3
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication TTLL10-AS1 SCNN1D RNF223 LOC100288175 MIR200B PUSL1 TNFRSF4 AGRN B3GALT6 MIR6727 C1orf159 MIR6726 SDF4 CPSF3L MIR429 LOC105378948 UBE2J2 TNFRSF18 ACAP3 TAS1R3 TTLL10 LINC01342 FAM132A MIR200A CPTP
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss MMP23A PLEKHN1 MIB2 LOC107985728 CCNL2 LOC102724312 SCNN1D ISG15 ATAD3C MIR200B TNFRSF4 NOC2L AGRN TMEM240 MRPL20 DVL1 MIR6727 FAM41C MIR6726 LOC107985729 CPSF3L LOC284600 ANKRD65 FAM87B KLHL17 FNDC10 TMEM88B ATAD3B AURKAIP1 UBE2J2 TNFRSF18 TAS1R3 TTLL10 MXRA8 LINC01342 FAM132A LOC100287934 MIR200A CPTP TTLL10-AS1 NADK PERM1 SLC35E2B SSU72 RNF223 LOC100288175 PUSL1 LINC00115 B3GALT6 CDK11B C1orf159 SLC35E2 SDF4 ATAD3A CDK11A LOC148413 MMP23B LOC100130417 LOC100288069 MIR429 LINC01128 LOC105378948 HES4 MIR6808 ACAP3 SAMD11 VWA1
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss SCNN1D UBE2J2 MIR6726 SDF4 ACAP3 FAM132A B3GALT6
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss PLEKHN1 MIR1302-2 LOC107985728 PRKCZ MIR6859-2 SCNN1D WASH7P FAAP20 TNFRSF4 OR4F29 TMEM240 DVL1 MIR6726 CPSF3L LOC284600 FAM87B LOC105378947 KLHL17 TMEM88B ATAD3B AURKAIP1 UBE2J2 MIR6859-1 TTLL10 MXRA8 LINC01342 LOC100287934 LOC105378949 CFAP74 NADK SLC35E2B FAM138A LOC100132287 SSU72 LOC105378589 RNF223 PUSL1 SKI LINC00115 B3GALT6 LOC107984841 C1orf159 LOC100129534 SLC35E2 SDF4 CDK11A LOC148413 MMP23B LOC100130417 LOC105378591 LINC01128 MORN1 LOC100134822 MMP23A LOC729737 MIB2 LOC100133331 GNB1 CCNL2 LOC102724312 OR4F5 ISG15 ATAD3C MIR200B NOC2L MIR6723 AGRN OR4F16 MRPL20 MIR6727 FAM41C LOC107985729 ANKRD65 LOC101928626 GABRD FNDC10 LOC100506504 TNFRSF18 TAS1R3 DDX11L1 FAM132A MIR200A CPTP TTLL10-AS1 PERM1 CALML6 LOC100288175 CDK11B MTND2P28 ATAD3A LOC105378592 LOC100288069 LOC102725121 MIR429 LOC105378948 HES4 MIR6808 TMEM52 ACAP3 MTND1P23 SAMD11 VWA1
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss TTLL10-AS1 PLEKHN1 PERM1 LOC107985728 ISG15 RNF223 LOC100288175 MIR200B TNFRSF4 NOC2L LINC00115 AGRN B3GALT6 C1orf159 FAM41C SDF4 LOC284600 FAM87B LOC100130417 MIR429 LINC01128 LOC105378948 KLHL17 HES4 UBE2J2 TNFRSF18 TTLL10 LINC01342 FAM132A MIR200A SAMD11
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion TTLL10-AS1 MIB2 CCNL2 SSU72 LOC102724312 SCNN1D ATAD3C MIR200B PUSL1 TNFRSF4 TMEM240 B3GALT6 MRPL20 CDK11B DVL1 MIR6727 C1orf159 MIR6726 SDF4 ATAD3A LOC107985729 CPSF3L LOC148413 ANKRD65 MMP23B MIR429 FNDC10 TMEM88B ATAD3B AURKAIP1 UBE2J2 MIR6808 TNFRSF18 ACAP3 TAS1R3 TTLL10 MXRA8 LINC01342 FAM132A MIR200A VWA1 CPTP
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss DVL1 MIR6727 MIR6726 CPSF3L SCNN1D AURKAIP1 UBE2J2 PUSL1 MIR6808 ACAP3 TAS1R3 MXRA8 CPTP
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain TTLL10-AS1 PLEKHN1 PERM1 LOC107985728 CCNL2 SCNN1D ISG15 RNF223 LOC100288175 MIR200B PUSL1 TNFRSF4 NOC2L AGRN B3GALT6 MRPL20 DVL1 MIR6727 C1orf159 MIR6726 SDF4 CPSF3L LOC284600 LOC148413 ANKRD65 LOC100130417 MIR429 LOC105378948 KLHL17 TMEM88B AURKAIP1 HES4 UBE2J2 MIR6808 TNFRSF18 ACAP3 TAS1R3 TTLL10 MXRA8 LINC01342 FAM132A MIR200A SAMD11 CPTP
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain TTLL10-AS1 C1orf159 SDF4 MIR429 LOC105378948 SCNN1D RNF223 UBE2J2 MIR200B TNFRSF18 TNFRSF4 TTLL10 LINC01342 FAM132A MIR200A B3GALT6
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss TTLL10-AS1 C1orf159 MIR6726 SDF4 MIR429 SCNN1D RNF223 UBE2J2 MIR200B TNFRSF18 ACAP3 TNFRSF4 TTLL10 LINC01342 FAM132A MIR200A B3GALT6
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain TTLL10-AS1 MIR6727 MIR6726 SDF4 CPSF3L MIR429 SCNN1D UBE2J2 MIR200B PUSL1 TNFRSF18 ACAP3 TNFRSF4 TTLL10 FAM132A MIR200A B3GALT6

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-65nqn:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline