This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03336756_cn
See other MIB2 CNV Assays ›
Gene Symbol
MIB2
Assay Reference Genome
Location

Chr.1:1622546 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03336756_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

142678

Gene Name:

mindbomb E3 ubiquitin protein ligase 2

Gene Aliases:

ZZANK1, ZZZ5

Location:

Chr.1:1615188-1630610 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MIB2 NM_001170686.1 NP_001164157.1
NM_001170687.1 NP_001164158.1
NM_001170688.1 NP_001164159.1
NM_001170689.1 NP_001164160.1
NM_080875.2 NP_543151.2
NR_033183.1
XM_006710372.1 XP_006710435.1
XM_011540731.2 XP_011539033.1
XM_011540736.2 XP_011539038.1
XM_011540737.2 XP_011539039.1
XM_011540741.2 XP_011539043.1
XM_011540742.2 XP_011539044.1
XM_017000349.1 XP_016855838.1
XM_017000350.1 XP_016855839.1
XM_017000351.1 XP_016855840.1
XM_017000352.1 XP_016855841.1
XM_017000353.1 XP_016855842.1
XM_017000354.1 XP_016855843.1
XM_017000355.1 XP_016855844.1
XM_017000356.1 XP_016855845.1
XM_017000357.1 XP_016855846.1
XM_017000358.1 XP_016855847.1
XM_017000359.1 XP_016855848.1
XM_017000360.1 XP_016855849.1
XM_017000361.1 XP_016855850.1
XM_017000362.1 XP_016855851.1
XM_017000363.1 XP_016855852.1
XM_017000364.1 XP_016855853.1
XM_017000365.1 XP_016855854.1
XM_017000366.1 XP_016855855.1
AB074480.1 BAB92950.1
AB076691.1 BAC00992.1
AB076692.1 BAC00993.1
AB076693.1 BAC00994.1
AB097000.1 BAC77353.1
AK098785.1 BAC05413.1
AK122751.1
AK122863.1
AK128167.1
AK128553.1
AK293642.1
AK302757.1
AK309362.1
BC016490.1 AAH16490.1
BC037542.1 AAH37542.1
DA391220.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion MMP23A SLC35E2B SLC35E2 NADK FNDC10 GNB1 MIB2 CDK11B MMP23B CDK11A
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss MMP23A SLC35E2B SLC35E2 FNDC10 MIB2 CDK11B MMP23B SSU72 CDK11A
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss LOC105378948 TTLL10 CCNL2 MIR200B MIR6726 LOC107985728 LOC284600 DVL1 LOC148413 LOC100287934 LOC107985729 AURKAIP1 AGRN HES4 LOC100288069 KLHL17 SLC35E2B FNDC10 ATAD3A NOC2L C1orf159 FAM41C NADK CDK11B MXRA8 FAM87B SCNN1D LOC100288175 LOC102724312 MMP23A LINC01342 MIB2 ANKRD65 MMP23B VWA1 ATAD3C LINC01128 ACAP3 PERM1 LINC00115 MRPL20 SLC35E2 TTLL10-AS1 TAS1R3 TMEM240 PUSL1 FAM132A CPTP TMEM88B LOC100130417 B3GALT6 MIR200A PLEKHN1 CPSF3L TNFRSF4 ATAD3B SDF4 SSU72 RNF223 CDK11A SAMD11 MIR6808 UBE2J2 MIR6727 ISG15 MIR429 TNFRSF18
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss TTLL10 LOC105378949 CCNL2 LOC729737 MIR200B MIR1302-2 LOC107985728 LOC284600 CFAP74 AURKAIP1 SKI LOC100133331 LOC100129534 LOC105378947 ATAD3A NOC2L LOC105378592 NADK DDX11L1 GNB1 LOC107984841 MTND2P28 LOC100288175 LOC102724312 LOC102725121 OR4F16 LINC01342 MIB2 ANKRD65 VWA1 ACAP3 PERM1 LINC00115 MRPL20 SLC35E2 OR4F5 TAS1R3 PUSL1 OR4F29 TMEM88B LOC100130417 MIR200A PLEKHN1 CPSF3L FAAP20 WASH7P SAMD11 UBE2J2 MIR6727 MIR429 LOC100134822 TMEM52 LOC101928626 LOC105378948 LOC100506504 MIR6726 LOC105378591 DVL1 PRKCZ LOC148413 LOC100287934 LOC107985729 MTND1P23 AGRN LOC105378589 HES4 MORN1 CALML6 LOC100288069 KLHL17 SLC35E2B FNDC10 MIR6859-1 C1orf159 FAM41C CDK11B GABRD MXRA8 FAM87B SCNN1D MMP23A MIR6859-2 MMP23B ATAD3C LINC01128 FAM138A LOC100132287 TTLL10-AS1 TMEM240 MIR6723 FAM132A CPTP B3GALT6 TNFRSF4 ATAD3B SDF4 SSU72 RNF223 CDK11A MIR6808 ISG15 TNFRSF18
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication MMP23A SLC35E2B SLC35E2 NADK FNDC10 GNB1 MIB2 CDK11B MMP23B SSU72 CDK11A
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion LOC102724312 TTLL10 LINC01342 MIB2 CCNL2 ANKRD65 MIR200B MIR6726 MMP23B VWA1 ATAD3C ACAP3 DVL1 MRPL20 LOC148413 LOC107985729 AURKAIP1 TTLL10-AS1 TAS1R3 TMEM240 PUSL1 FAM132A CPTP TMEM88B B3GALT6 FNDC10 ATAD3A MIR200A CPSF3L TNFRSF4 ATAD3B SDF4 SSU72 C1orf159 MIR6808 CDK11B UBE2J2 MIR6727 MIR429 TNFRSF18 MXRA8 SCNN1D
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss MMP23A SLC35E2B SLC35E2 NADK FNDC10 MIB2 CDK11B MMP23B SSU72 CDK11A
nsv545017 Chr.1:1615322 - 1663759 on Build GRCh38 Loss SLC35E2B MIB2 CDK11B MMP23B
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss LOC102724312 MMP23A LOC105378949 MIB2 CCNL2 ANKRD65 MMP23B VWA1 ATAD3C LOC105378591 PRKCZ MRPL20 LOC148413 SLC35E2 CFAP74 LOC107985729 TMEM240 LOC105378589 CALML6 TMEM88B SLC35E2B FNDC10 ATAD3A ATAD3B SSU72 CDK11A NADK GNB1 CDK11B GABRD TMEM52

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
Notch signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0