This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03337997_cn
See other CHD1L CNV Assays ›
Gene Symbol
CHD1L
Assay Reference Genome
Location

Chr.1:147196728 on build GRCh38
Cytoband
1q21.1
Assay ID Hs03337997_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9557

Gene Name:

chromodomain helicase DNA binding protein 1 like

Gene Aliases:

ALC1, CHDL

Location:

Chr.1:147172804-147295766 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CHD1L XM_017002858.1 XP_016858347.1

Target Gene Details

Entrez Gene ID:

2330

Gene Name:

flavin containing monooxygenase 5

Gene Aliases:

-

Location:

Chr.1:147183963-147225798 on Build GRCh38

Assay Gene Location:

Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FMO5 NM_001144829.2 NP_001138301.1
NM_001144830.2 NP_001138302.1
NM_001461.3 NP_001452.2
XM_005272946.4 XP_005273003.1
XM_005272947.4 XP_005273004.1
XM_005272948.4 XP_005273005.1
XM_006711245.3 XP_006711308.1
XM_011509350.2 XP_011507652.1
XM_011509351.2 XP_011507653.1
XM_017000801.1 XP_016856290.1
XM_017000802.1 XP_016856291.1
AK222728.1 BAD96448.1
AK225739.1
AK314647.1
BC035687.1 AAH35687.1
L37080.1 AAA67849.1
Z47553.1 CAA87633.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv327n100 Chr.1:146028214 - 148860637 on Build GRCh38 Gain CH17-408M7.1 LINC01138 LOC101927468 NBPF12 MIR5087 FMO5 RNVU1-8 LOC100132057 GPR89B NUDT4P2 NUDT4P1 CHD1L PRKAB2 LOC101928979 NBPF14 LINC00624 LOC100996517 NOTCH2NL RNVU1-6 RNVU1-7 RNU1-13P LOC284561 LOC105371226 NBPF13P LOC105371242 PDZK1P1 PPIAL4G PDE4DIP NBPF11 RNVU1-1 PDIA3P1 BCL9 LOC728989 HYDIN2 MIR6077 GJA8 NBPF10 RNVU1-3 LOC100996763 ACP6 GJA5 LOC100505824
nsv1005055 Chr.1:145642988 - 148991068 on Build GRCh38 Loss CH17-408M7.1 LIX1L LOC101927468 NBPF12 RNF115 RNVU1-8 ANKRD34A GPR89B NUDT4P2 NUDT4P1 TXNIP CHD1L PRKAB2 NOTCH2NL RNVU1-7 NBPF11 PDIA3P1 LOC728989 MIR6077 GJA8 NBPF10 RBM8A LOC100996763 MIR6736 NUDT17 LOC100505824 HFE2 LINC01138 MIR5087 POLR3GL FMO5 LOC100132057 PDZK1 LOC101928979 NBPF14 LINC00624 LOC100996517 ITGA10 RNVU1-6 RNU1-13P LOC284561 LOC105371226 NBPF13P LOC105371242 PDZK1P1 PPIAL4G PEX11B PDE4DIP RNVU1-1 BCL9 HYDIN2 CD160 POLR3C RNVU1-3 GNRHR2 ACP6 ANKRD35 PIAS3 GJA5 GPR89A
dgv67e199 Chr.1:147188956 - 147198583 on Build GRCh38 Deletion CHD1L FMO5
nsv1121713 Chr.1:147010816 - 147207577 on Build GRCh38 Deletion LOC728989 NBPF13P CHD1L PRKAB2 FMO5 RNVU1-8 PDIA3P1
esv2761299 Chr.1:146068341 - 148926801 on Build GRCh38 Gain+Loss CH17-408M7.1 LINC01138 LOC101927468 NBPF12 MIR5087 FMO5 RNVU1-8 LOC100132057 GPR89B NUDT4P2 NUDT4P1 CHD1L PRKAB2 NBPF14 LINC00624 LOC100996517 NOTCH2NL RNVU1-7 RNU1-13P LOC284561 LOC105371226 NBPF13P LOC105371242 PDZK1P1 PPIAL4G PDE4DIP NBPF11 RNVU1-1 PDIA3P1 BCL9 LOC728989 HYDIN2 MIR6077 GJA8 NBPF10 RNVU1-3 LOC100996763 ACP6 GJA5 LOC100505824
nsv509457 Chr.1:145948024 - 149021821 on Build GRCh38 Insertion HFE2 CH17-408M7.1 LIX1L LINC01138 LOC101927468 NBPF12 MIR5087 POLR3GL FMO5 RNVU1-8 ANKRD34A LOC100132057 GPR89B NUDT4P2 NUDT4P1 TXNIP CHD1L PRKAB2 LOC101928979 NBPF14 LINC00624 LOC100996517 NOTCH2NL RNVU1-6 RNVU1-7 RNU1-13P LOC284561 LOC105371226 NBPF13P LOC105371242 PDZK1P1 PPIAL4G PDE4DIP NBPF11 RNVU1-1 PDIA3P1 BCL9 LOC728989 HYDIN2 MIR6077 GJA8 NBPF10 RNVU1-3 LOC100996763 ACP6 GJA5 LOC100505824
nsv1132304 Chr.1:146186094 - 148977674 on Build GRCh38 Duplication CH17-408M7.1 LINC01138 LOC101927468 NBPF12 MIR5087 FMO5 RNVU1-8 LOC100132057 GPR89B NUDT4P2 NUDT4P1 CHD1L PRKAB2 NBPF14 LINC00624 LOC100996517 NOTCH2NL RNVU1-7 RNU1-13P LOC284561 LOC105371226 NBPF13P LOC105371242 PDZK1P1 PPIAL4G PDE4DIP NBPF11 RNVU1-1 PDIA3P1 BCL9 LOC728989 HYDIN2 MIR6077 GJA8 RNVU1-3 LOC100996763 ACP6 GJA5 LOC100505824
nsv1003688 Chr.1:146036817 - 148934526 on Build GRCh38 Gain CH17-408M7.1 LINC01138 LOC101927468 NBPF12 MIR5087 FMO5 RNVU1-8 LOC100132057 GPR89B NUDT4P2 NUDT4P1 CHD1L PRKAB2 LOC101928979 NBPF14 LINC00624 LOC100996517 NOTCH2NL RNVU1-6 RNVU1-7 RNU1-13P LOC284561 LOC105371226 NBPF13P LOC105371242 PDZK1P1 PPIAL4G PDE4DIP NBPF11 RNVU1-1 PDIA3P1 BCL9 LOC728989 HYDIN2 MIR6077 GJA8 NBPF10 RNVU1-3 LOC100996763 ACP6 GJA5 LOC100505824
dgv66e199 Chr.1:146055970 - 149056514 on Build GRCh38 Deletion CH17-408M7.1 LINC01138 LOC101927468 NBPF12 MIR5087 FMO5 RNVU1-8 LOC100132057 GPR89B NUDT4P2 NUDT4P1 CHD1L PRKAB2 LOC101928979 NBPF9 NBPF14 LINC00624 LOC100996517 NOTCH2NL RNVU1-7 RNU1-13P LOC284561 LOC105371226 NBPF13P LOC105371242 PDZK1P1 PPIAL4G PDE4DIP NBPF11 RNVU1-1 PDIA3P1 BCL9 LOC728989 HYDIN2 MIR6077 GJA8 NBPF10 RNVU1-3 LOC100996763 ACP6 GJA5 LOC100505824
esv3587435 Chr.1:147189056 - 147198713 on Build GRCh38 Loss CHD1L FMO5
nsv437047 Chr.1:147171976 - 147369886 on Build GRCh38 Loss CHD1L PRKAB2 FMO5 PDIA3P1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
chromatin remodeling
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
global genome nucleotide-excision repair
biological_process
drug metabolic process
oxidation-reduction process
nucleotide binding
nucleic acid binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
ATPase activity
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-55ff9ddd88-mtlvn:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0