This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03338997_cn
See other CPSF3L CNV Assays ›
Gene Symbol
CPSF3L
Assay Reference Genome
Location

Chr.1:1317629 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03338997_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

54973

Gene Name:

cleavage and polyadenylation specific factor 3-like

Gene Aliases:

CPSF73L, INT11, INTS11, RC-68, RC68

Location:

Chr.1:1311585-1324687 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CPSF3L NM_001256456.1 NP_001243385.1
NM_001256460.1 NP_001243389.1
NM_001256462.1 NP_001243391.1
NM_001256463.1 NP_001243392.1
NM_017871.5 NP_060341.2
XM_011541647.1 XP_011539949.1
XM_011541648.1 XP_011539950.1
XM_011541650.1 XP_011539952.1
XM_017001557.1 XP_016857046.1
XM_017001558.1 XP_016857047.1
AK021939.1 BAB13943.1
AK023356.1 BAB14541.1
AK056652.1
AK058018.1
AK092190.1
AK291387.1
AK297350.1
AL136813.1 CAB66747.1
BC000675.2 AAH00675.2
BC007978.2 AAH07978.1
BC008041.2 AAH08041.1
BC020199.1 AAH20199.1
CR533557.1 CAG38588.1
DC295791.1
DC299237.1
DC299993.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss KLHL17 MIR6859-1 CALML6 TNFRSF18 PRKCZ CDK11B MRPL20 LOC105378592 CFAP74 LOC100287934 MIR6808 FAM87B LOC100129534 MMP23B ISG15 MIR200B PERM1 MTND1P23 SLC35E2 LOC284600 TMEM240 NADK TMEM52 VWA1 C1orf159 LOC107985728 WASH7P B3GALT6 MMP23A MIR6726 LOC100130417 FAM138A GABRD MIB2 MIR1302-2 FAM41C LOC100506504 LOC100288069 ACAP3 ATAD3A LOC105378947 FNDC10 LOC100288175 MIR6859-2 LOC102724312 LINC01342 LOC105378589 SCNN1D FAM132A SKI FAAP20 MTND2P28 DVL1 LOC105378591 MIR200A TNFRSF4 LOC107985729 LOC148413 ATAD3B TTLL10-AS1 SDF4 SAMD11 CPSF3L ATAD3C LOC100133331 LOC105378948 LOC107984841 DDX11L1 OR4F16 RNF223 CPTP TMEM88B LOC729737 SLC35E2B LINC00115 TTLL10 MIR429 PUSL1 AURKAIP1 SSU72 MORN1 UBE2J2 AGRN PLEKHN1 LOC101928626 LOC100134822 OR4F29 LOC102725121 MIR6723 MXRA8 ANKRD65 CDK11A LOC100132287 TAS1R3 OR4F5 LOC105378949 CCNL2 HES4 LINC01128 GNB1 MIR6727 NOC2L
nsv544971 Chr.1:1294063 - 1351426 on Build GRCh38 Loss MIR6808 CPTP ACAP3 CPSF3L PUSL1 TAS1R3 MIR6727 MIR6726 DVL1
nsv544969 Chr.1:1290683 - 1374597 on Build GRCh38 Loss MIR6808 CPTP SCNN1D ACAP3 CPSF3L PUSL1 AURKAIP1 TAS1R3 MIR6727 MIR6726 DVL1 MXRA8
nsv997291 Chr.1:1306835 - 1376336 on Build GRCh38 Gain MIR6808 CPTP ACAP3 CPSF3L PUSL1 AURKAIP1 TAS1R3 MIR6727 DVL1 MXRA8
nsv470680 Chr.1:1070426 - 1378635 on Build GRCh38 Loss UBE2J2 SDF4 CPSF3L TNFRSF18 ACAP3 RNF223 MXRA8 MIR6808 CPTP LINC01342 SCNN1D FAM132A TAS1R3 MIR200B DVL1 TTLL10 MIR200A TNFRSF4 MIR429 PUSL1 AURKAIP1 C1orf159 TTLL10-AS1 MIR6727 B3GALT6 MIR6726
dgv6n100 Chr.1:1301750 - 1362354 on Build GRCh38 Gain+Loss MIR6808 CPTP ACAP3 CPSF3L PUSL1 TAS1R3 MIR6727 DVL1 MXRA8
esv3584992 Chr.1:1314419 - 1330342 on Build GRCh38 Gain CPTP CPSF3L
nsv1160788 Chr.1:1282706 - 1376336 on Build GRCh38 Deletion MIR6808 CPTP SCNN1D ACAP3 CPSF3L PUSL1 AURKAIP1 TAS1R3 MIR6727 MIR6726 DVL1 MXRA8
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication UBE2J2 AGRN SDF4 CPSF3L TNFRSF18 ACAP3 LOC105378948 LOC100288175 RNF223 CPTP LINC01342 SCNN1D FAM132A TAS1R3 MIR200B TTLL10 MIR200A TNFRSF4 MIR429 PUSL1 C1orf159 TTLL10-AS1 MIR6727 B3GALT6 MIR6726
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss KLHL17 SDF4 SAMD11 CPSF3L ATAD3C TNFRSF18 CDK11B MRPL20 LOC105378948 RNF223 LOC100287934 MIR6808 CPTP FAM87B TMEM88B MMP23B ISG15 SLC35E2B MIR200B PERM1 SLC35E2 LINC00115 TTLL10 LOC284600 TMEM240 MIR429 PUSL1 NADK VWA1 AURKAIP1 C1orf159 LOC107985728 SSU72 B3GALT6 MMP23A MIR6726 LOC100130417 UBE2J2 AGRN PLEKHN1 MIB2 FAM41C LOC100288069 ACAP3 ATAD3A FNDC10 LOC100288175 MXRA8 ANKRD65 LOC102724312 CDK11A LINC01342 SCNN1D FAM132A TAS1R3 CCNL2 DVL1 MIR200A HES4 TNFRSF4 LOC107985729 LOC148413 LINC01128 ATAD3B TTLL10-AS1 MIR6727 NOC2L
esv3890636 Chr.1:1304555 - 1353203 on Build GRCh38 Loss MIR6808 CPTP ACAP3 CPSF3L PUSL1 TAS1R3 MIR6727 DVL1 MXRA8
nsv1000169 Chr.1:1281678 - 1362354 on Build GRCh38 Gain MIR6808 CPTP SCNN1D ACAP3 CPSF3L PUSL1 TAS1R3 MIR6727 MIR6726 DVL1 MXRA8
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion UBE2J2 SDF4 CPSF3L MIB2 ATAD3C TNFRSF18 CDK11B MRPL20 ACAP3 ATAD3A FNDC10 MXRA8 MIR6808 ANKRD65 CPTP LOC102724312 TMEM88B LINC01342 MMP23B SCNN1D FAM132A TAS1R3 MIR200B CCNL2 DVL1 TTLL10 MIR200A TNFRSF4 LOC107985729 LOC148413 TMEM240 MIR429 PUSL1 ATAD3B VWA1 AURKAIP1 C1orf159 SSU72 TTLL10-AS1 MIR6727 B3GALT6 MIR6726
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss MIR6808 UBE2J2 CPTP SCNN1D CPSF3L TAS1R3 DVL1 ACAP3 PUSL1 AURKAIP1 MIR6727 MIR6726 MXRA8
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain UBE2J2 AGRN KLHL17 SDF4 PLEKHN1 SAMD11 CPSF3L TNFRSF18 MRPL20 ACAP3 LOC105378948 LOC100288175 RNF223 MXRA8 MIR6808 ANKRD65 CPTP TMEM88B LINC01342 ISG15 SCNN1D FAM132A TAS1R3 MIR200B PERM1 CCNL2 DVL1 TTLL10 MIR200A HES4 LOC284600 TNFRSF4 LOC148413 MIR429 PUSL1 AURKAIP1 C1orf159 LOC107985728 TTLL10-AS1 MIR6727 B3GALT6 MIR6726 LOC100130417 NOC2L
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain UBE2J2 SDF4 SCNN1D FAM132A CPSF3L MIR200B TNFRSF18 TTLL10 MIR200A TNFRSF4 MIR429 ACAP3 PUSL1 TTLL10-AS1 MIR6727 B3GALT6 MIR6726

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

snRNA processing
snRNA transcription from RNA polymerase II promoter
protein binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0