This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03342291_cn
See other PLCH2 CNV Assays ›
Gene Symbol
PLCH2
Assay Reference Genome
Location

Chr.1:2435725 on build GRCh38
Cytoband
1p36.32
Assay ID Hs03342291_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9651

Gene Name:

phospholipase C eta 2

Gene Aliases:

PLC-L4, PLC-eta2, PLCL4, PLCeta2

Location:

Chr.1:2412481-2505530 on Build GRCh38

Assay Gene Location:

Within Intron 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
PLCH2 XM_011542455.2 XP_011540757.1
XM_011542456.2 XP_011540758.1
XM_017002870.1 XP_016858359.1
XM_017002871.1 XP_016858360.1
XM_017002872.1 XP_016858361.1
XM_017002873.1 XP_016858362.1
XM_017002874.1 XP_016858363.1
AK122591.2

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv545078 Chr.1:2434877 - 2499618 on Build GRCh38 Loss PLCH2
nsv951557 Chr.1:2266062 - 2564661 on Build GRCh38 Deletion MORN1 TNFRSF14 LOC115110 PLCH2 HES5 PANK4 LOC100129534 SKI RER1 PEX10
nsv545072 Chr.1:2401592 - 2475448 on Build GRCh38 Loss PLCH2 RER1 PEX10
nsv545075 Chr.1:2428542 - 2458765 on Build GRCh38 Loss PLCH2
nsv470684 Chr.1:2394570 - 2930923 on Build GRCh38 Loss MMEL1 TNFRSF14 LOC115110 PLCH2 HES5 FAM213B PANK4 RER1 TTC34 LOC100996583 PEX10

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
lipid catabolic process
intracellular signal transduction
inositol phosphate metabolic process
phosphatidylinositol metabolic process
molecular_function
phosphatidylinositol phospholipase C activity
signal transducer activity
calcium ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline