This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03342680_cn
See other SLC35E2B CNV Assays ›
Gene Symbol
SLC35E2B
Assay Reference Genome
Location

Chr.1:1670853 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03342680_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

728661

Gene Name:

solute carrier family 35 member E2B

Gene Aliases:

SLC35E2

Location:

Chr.1:1661478-1692804 on Build GRCh38

Assay Gene Location:

Within Intron 7
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SLC35E2B NM_001110781.2 NP_001104251.1
NM_001290264.1 NP_001277193.1
XM_011542072.1 XP_011540374.1
AB007916.3
AK125619.1
AK307886.1
BC110653.1 AAI10654.1
CR936618.1 CAI56761.1
DA794179.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv37n54 Chr.1:1663759 - 1696548 on Build GRCh38 Loss SLC35E2B
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion SLC35E2 CDK11B MMP23B FNDC10 NADK MIB2 MMP23A SLC35E2B GNB1 CDK11A
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss SLC35E2 SSU72 CDK11B MMP23B FNDC10 MIB2 MMP23A SLC35E2B CDK11A
esv3575582 Chr.1:1624086 - 1721336 on Build GRCh38 Gain CDK11B MMP23B MIB2 MMP23A SLC35E2B CDK11A
esv3585010 Chr.1:1670280 - 1751138 on Build GRCh38 Loss SLC35E2 MMP23A SLC35E2B CDK11A
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC102725121 ATAD3A OR4F29 CCNL2 CPSF3L LOC100288175 CDK11B MIR200A LOC101928626 LOC105378947 LOC102724312 CALML6 WASH7P MMP23B MORN1 MIB2 LOC100288069 CDK11A CPTP HES4 MTND2P28 NOC2L FAM138A TMEM88B LOC100129534 LOC729737 LOC148413 MIR6723 NADK LOC107985729 MIR1302-2 MXRA8 MTND1P23 LOC105378949 LOC100287934 LOC100134822 TNFRSF4 SAMD11 TTLL10 RNF223 ANKRD65 ACAP3 ATAD3C LOC100506504 ISG15 LINC01128 PRKCZ PERM1 TTLL10-AS1 LOC100130417 SLC35E2 MIR6808 FNDC10 B3GALT6 TMEM240 LOC105378591 AURKAIP1 LINC00115 MIR200B MIR429 SKI GABRD LOC100132287 SLC35E2B LOC284600 MRPL20 LINC01342 LOC100133331 C1orf159 MIR6727 ATAD3B FAM41C AGRN GNB1 OR4F16 LOC107985728 PUSL1 PLEKHN1 LOC105378592 DVL1 TMEM52 VWA1 LOC105378589 CFAP74 MIR6726 LOC107984841 SDF4 KLHL17 TAS1R3 MMP23A TNFRSF18 FAM87B OR4F5 LOC105378948 SSU72 DDX11L1 FAM132A UBE2J2 MIR6859-2 SCNN1D FAAP20 MIR6859-1
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication SLC35E2 SSU72 CDK11B MMP23B FNDC10 NADK MIB2 MMP23A SLC35E2B GNB1 CDK11A
dgv2e199 Chr.1:1653096 - 1721890 on Build GRCh38 Deletion CDK11B MMP23A SLC35E2B CDK11A
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss SLC35E2 SSU72 CDK11B MMP23B FNDC10 NADK MIB2 MMP23A SLC35E2B CDK11A
esv3585008 Chr.1:1663929 - 1681002 on Build GRCh38 Loss SLC35E2B
esv3585007 Chr.1:1647187 - 1714507 on Build GRCh38 Loss CDK11B MMP23A SLC35E2B CDK11A
nsv950516 Chr.1:1663141 - 1715961 on Build GRCh38 Deletion MMP23A SLC35E2B CDK11A
esv27786 Chr.1:1632502 - 1752267 on Build GRCh38 Gain SLC35E2 CDK11B MMP23B NADK MMP23A SLC35E2B CDK11A
nsv545022 Chr.1:1665581 - 1683909 on Build GRCh38 Loss SLC35E2B
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss ATAD3A TTLL10-AS1 CCNL2 LOC100130417 CPSF3L SLC35E2 MIR6808 LOC100288175 CDK11B MIR200A FNDC10 B3GALT6 LOC102724312 TMEM240 AURKAIP1 LINC00115 MIR200B MMP23B MIR429 MIB2 LOC100288069 SLC35E2B CDK11A LOC284600 MRPL20 LINC01342 C1orf159 MIR6727 ATAD3B FAM41C CPTP HES4 AGRN NOC2L LOC107985728 TMEM88B PUSL1 PLEKHN1 LOC148413 DVL1 NADK VWA1 LOC107985729 MXRA8 LOC100287934 MIR6726 SDF4 TNFRSF4 SAMD11 TTLL10 RNF223 ANKRD65 KLHL17 TAS1R3 MMP23A TNFRSF18 FAM87B LOC105378948 ACAP3 ATAD3C SSU72 FAM132A UBE2J2 ISG15 SCNN1D LINC01128 PERM1
nsv545018 Chr.1:1638499 - 1688102 on Build GRCh38 Loss CDK11B SLC35E2B
dgv3n67 Chr.1:1653920 - 1735718 on Build GRCh38 Gain SLC35E2 CDK11B MMP23A SLC35E2B CDK11A
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss TMEM88B ATAD3A LOC148413 NADK CCNL2 TMEM52 VWA1 LOC107985729 SLC35E2 LOC105378589 CDK11B LOC105378949 FNDC10 CFAP74 LOC102724312 TMEM240 LOC105378591 CALML6 MMP23B GABRD ANKRD65 MIB2 MMP23A SLC35E2B CDK11A ATAD3C SSU72 MRPL20 ATAD3B GNB1 PRKCZ

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

secondary carrier transporter

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline