This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03343200_cn
See other MIB2 CNV Assays ›
Gene Symbol
MIB2
Assay Reference Genome
Location

Chr.1:1615232 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03343200_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

142678

Gene Name:

mindbomb E3 ubiquitin protein ligase 2

Gene Aliases:

ZZANK1, ZZZ5

Location:

Chr.1:1615188-1630610 on Build GRCh38

Assay Gene Location:

Overlaps - Exon 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MIB2 XM_011540731.2 XP_011539033.1
XM_011540736.2 XP_011539038.1
XM_011540737.2 XP_011539039.1
XM_011540741.2 XP_011539043.1
XM_011540742.2 XP_011539044.1
XM_017000349.1 XP_016855838.1
XM_017000350.1 XP_016855839.1
XM_017000351.1 XP_016855840.1
XM_017000352.1 XP_016855841.1
XM_017000353.1 XP_016855842.1
XM_017000354.1 XP_016855843.1
XM_017000355.1 XP_016855844.1
XM_017000356.1 XP_016855845.1
XM_017000357.1 XP_016855846.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion SLC35E2 NADK FNDC10 MMP23B MMP23A CDK11A SLC35E2B CDK11B MIB2 GNB1
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss SLC35E2 FNDC10 MMP23B SSU72 MMP23A CDK11A SLC35E2B CDK11B MIB2
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss DVL1 SLC35E2 PUSL1 ACAP3 SAMD11 CCNL2 CDK11A PLEKHN1 CPSF3L VWA1 LINC00115 MMP23B MIR6727 MIR6808 LOC148413 CPTP TAS1R3 FNDC10 FAM41C AURKAIP1 CDK11B MIR200A C1orf159 MIR6726 LINC01128 SDF4 PERM1 TMEM88B MRPL20 LINC01342 RNF223 ATAD3B TNFRSF18 HES4 TMEM240 SLC35E2B MIR429 B3GALT6 LOC100130417 SSU72 LOC100287934 LOC107985728 LOC284600 LOC107985729 MMP23A NOC2L TTLL10-AS1 AGRN ISG15 KLHL17 TNFRSF4 LOC100288175 SCNN1D LOC100288069 FAM132A MIB2 MIR200B FAM87B NADK MXRA8 UBE2J2 ATAD3C LOC102724312 LOC105378948 TTLL10 ANKRD65 ATAD3A
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss SLC35E2 PUSL1 LOC105378589 CFAP74 CDK11A PLEKHN1 VWA1 LINC00115 MIR6727 FAAP20 LOC101928626 CPTP MTND1P23 LOC107984841 PRKCZ AURKAIP1 MIR6859-2 CDK11B MIR6726 LINC01128 LOC729737 TMEM88B MRPL20 LINC01342 CALML6 RNF223 ATAD3B MIR6859-1 HES4 TMEM240 OR4F29 LOC100287934 LOC107985728 LOC107985729 MMP23A TTLL10-AS1 ISG15 KLHL17 DDX11L1 SCNN1D LOC100288069 LOC100506504 LOC105378591 FAM87B NADK UBE2J2 LOC105378947 ATAD3C LOC105378948 ANKRD65 LOC100129534 ATAD3A DVL1 ACAP3 SAMD11 MIR1302-2 CCNL2 LOC105378592 LOC100132287 GNB1 CPSF3L MMP23B MIR6808 OR4F5 FAM138A MORN1 LOC148413 LOC100133331 TAS1R3 FNDC10 FAM41C LOC100134822 LOC102725121 MIR200A C1orf159 SKI SDF4 PERM1 GABRD MIR6723 OR4F16 TNFRSF18 SLC35E2B MIR429 B3GALT6 LOC105378949 LOC100130417 SSU72 LOC284600 NOC2L AGRN TMEM52 TNFRSF4 LOC100288175 MTND2P28 FAM132A MIB2 MIR200B WASH7P MXRA8 LOC102724312 TTLL10
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication SLC35E2 NADK FNDC10 MMP23B SSU72 MMP23A CDK11A SLC35E2B CDK11B MIB2 GNB1
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion DVL1 TNFRSF18 PUSL1 TMEM240 ACAP3 CCNL2 MIR429 B3GALT6 CPSF3L VWA1 MMP23B MIR6727 MIR6808 SSU72 LOC107985729 LOC148413 TTLL10-AS1 CPTP TAS1R3 FNDC10 TNFRSF4 SCNN1D AURKAIP1 FAM132A CDK11B MIR200A MIB2 MIR200B C1orf159 MIR6726 SDF4 MXRA8 UBE2J2 TMEM88B ATAD3C LOC102724312 MRPL20 LINC01342 TTLL10 ATAD3B ANKRD65 ATAD3A
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss SLC35E2 NADK FNDC10 MMP23B SSU72 MMP23A CDK11A SLC35E2B CDK11B MIB2
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss SLC35E2 LOC105378589 CFAP74 TMEM240 CCNL2 CDK11A SLC35E2B GNB1 VWA1 LOC105378949 MMP23B SSU72 LOC107985729 MMP23A LOC148413 FNDC10 TMEM52 PRKCZ CDK11B MIB2 LOC105378591 NADK TMEM88B GABRD ATAD3C LOC102724312 MRPL20 CALML6 ATAD3B ANKRD65 ATAD3A

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
Notch signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0