This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03348105_cn
See other NOC2L CNV Assays ›
Gene Symbol
NOC2L
Assay Reference Genome
Location

Chr.1:954155 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03348105_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

26155

Gene Name:

NOC2 like nucleolar associated transcriptional repressor

Gene Aliases:

NET15, NET7, NIR, PPP1R112

Location:

Chr.1:944203-959299 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NOC2L NM_015658.3 NP_056473.2
AK024284.1
AK225239.1
AK315080.1
AL050019.1 CAB43240.2
BC003555.1 AAH03555.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2762302 Chr.1:914637 - 1106320 on Build GRCh38 Gain LOC100288175 RNF223 AGRN LOC100130417 SAMD11 LOC105378948 LOC107985728 NOC2L HES4 KLHL17 PLEKHN1 PERM1 LOC284600 C1orf159 ISG15
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC100133331 GNB1 MIR1302-2 MIR6808 MIR200A NOC2L ATAD3B PUSL1 SKI C1orf159 LOC105378589 LINC01342 LINC01128 CALML6 MIR429 TNFRSF18 MIR6726 TMEM52 VWA1 RNF223 TTLL10-AS1 LOC100132287 SAMD11 LOC107985728 PERM1 TAS1R3 LOC100129534 MIB2 MORN1 MTND2P28 UBE2J2 FAAP20 MIR6723 OR4F5 CDK11B OR4F16 TMEM88B CPSF3L LOC107984841 WASH7P LOC105378947 KLHL17 TMEM240 FNDC10 LOC729737 MIR200B DDX11L1 MRPL20 LOC102724312 LOC100288069 MXRA8 LOC100506504 PRKCZ LOC102725121 SCNN1D ANKRD65 SLC35E2B SDF4 CFAP74 LOC107985729 LOC105378948 PLEKHN1 CDK11A LOC284600 LOC105378592 MMP23B SSU72 TTLL10 NADK LINC00115 LOC100287934 CCNL2 FAM138A SLC35E2 MIR6859-1 B3GALT6 LOC105378591 FAM41C AURKAIP1 ATAD3C DVL1 MMP23A FAM87B ACAP3 TNFRSF4 LOC100130417 LOC105378949 GABRD ATAD3A MIR6859-2 FAM132A ISG15 MTND1P23 LOC100288175 LOC101928626 AGRN MIR6727 HES4 OR4F29 CPTP LOC148413 LOC100134822
nsv950451 Chr.1:899421 - 1094520 on Build GRCh38 Deletion LOC100288175 RNF223 AGRN LOC100130417 SAMD11 LOC105378948 LOC107985728 NOC2L HES4 KLHL17 PLEKHN1 PERM1 LOC284600 C1orf159 ISG15
nsv509035 Chr.1:891406 - 985724 on Build GRCh38 Insertion PLEKHN1 PERM1 LOC284600 LOC100130417 SAMD11 LOC107985728 NOC2L KLHL17
nsv428334 Chr.1:874371 - 1220569 on Build GRCh38 Gain FAM41C SDF4 TNFRSF4 LOC100130417 MIR200A LOC105378948 NOC2L KLHL17 PLEKHN1 MIR200B LOC284600 C1orf159 LINC01342 ISG15 MIR429 TTLL10 TNFRSF18 LOC100288175 RNF223 TTLL10-AS1 AGRN SAMD11 LOC107985728 HES4 PERM1
dgv5n100 Chr.1:585989 - 1114424 on Build GRCh38 Gain FAM41C LOC100133331 FAM87B MIR6723 LOC100130417 OR4F16 LOC105378948 LOC107984841 NOC2L LOC105378947 KLHL17 PLEKHN1 LOC284600 C1orf159 ISG15 LINC01128 MTND1P23 LOC100288069 LOC100288175 RNF223 LOC101928626 AGRN LINC00115 LOC100287934 SAMD11 LOC107985728 HES4 PERM1 MTND2P28
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss ANKRD65 SLC35E2B SDF4 MIR6808 MIR200A LOC107985729 LOC105378948 NOC2L ATAD3B PLEKHN1 CDK11A LOC284600 PUSL1 C1orf159 LINC01342 LINC01128 MMP23B MIR429 SSU72 TTLL10 TNFRSF18 MIR6726 NADK VWA1 RNF223 TTLL10-AS1 LINC00115 LOC100287934 SAMD11 CCNL2 LOC107985728 SLC35E2 PERM1 TAS1R3 MIB2 B3GALT6 UBE2J2 FAM41C AURKAIP1 ATAD3C DVL1 MMP23A FAM87B ACAP3 TNFRSF4 LOC100130417 CDK11B TMEM88B CPSF3L ATAD3A KLHL17 TMEM240 FNDC10 MIR200B MRPL20 FAM132A ISG15 LOC102724312 LOC100288069 LOC100288175 AGRN MIR6727 HES4 MXRA8 CPTP LOC148413 SCNN1D
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss FAM41C SDF4 FAM87B TNFRSF4 LOC100130417 MIR200A LOC105378948 NOC2L KLHL17 PLEKHN1 MIR200B LOC284600 C1orf159 LINC01342 FAM132A ISG15 LINC01128 MIR429 TTLL10 TNFRSF18 LOC100288175 RNF223 TTLL10-AS1 AGRN LINC00115 SAMD11 LOC107985728 HES4 PERM1 B3GALT6 UBE2J2
nsv1013524 Chr.1:939510 - 1161955 on Build GRCh38 Gain LOC100288175 RNF223 AGRN SAMD11 LOC105378948 NOC2L HES4 KLHL17 PLEKHN1 PERM1 C1orf159 LINC01342 ISG15
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain AURKAIP1 DVL1 ANKRD65 SDF4 ACAP3 TNFRSF4 LOC100130417 MIR6808 MIR200A TMEM88B CPSF3L LOC105378948 NOC2L KLHL17 PLEKHN1 MIR200B LOC284600 PUSL1 C1orf159 MRPL20 LINC01342 FAM132A ISG15 MIR429 TTLL10 TNFRSF18 MIR6726 LOC100288175 RNF223 TTLL10-AS1 AGRN MIR6727 SAMD11 CCNL2 LOC107985728 HES4 PERM1 MXRA8 TAS1R3 CPTP B3GALT6 LOC148413 SCNN1D UBE2J2
nsv544895 Chr.1:917392 - 1054900 on Build GRCh38 Loss PLEKHN1 PERM1 LOC284600 AGRN ISG15 LOC100130417 SAMD11 LOC107985728 NOC2L HES4 KLHL17
nsv1160644 Chr.1:939652 - 1220663 on Build GRCh38 Deletion TNFRSF18 LOC100288175 RNF223 TTLL10-AS1 SDF4 AGRN TNFRSF4 MIR200A SAMD11 LOC105378948 NOC2L HES4 KLHL17 PLEKHN1 PERM1 MIR200B C1orf159 LINC01342 ISG15 MIR429 TTLL10
nsv832980 Chr.1:848279 - 1007037 on Build GRCh38 Gain FAM41C PLEKHN1 PERM1 LOC284600 LINC01128 LOC100130417 SAMD11 LOC107985728 NOC2L HES4 KLHL17

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of B cell apoptotic process
transcription, DNA-templated
apoptotic process
chromatin assembly
nucleolus to nucleoplasm transport
cellular response to UV
negative regulation of histone acetylation
ribosomal large subunit biogenesis
regulation of signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway
chromatin binding
transcription corepressor activity
protein binding
nucleosome binding
histone binding
poly(A) RNA binding
repressing transcription factor binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-2x6nv:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline