This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03348126_cn
See other NOC2L CNV Assays ›
Gene Symbol
NOC2L
Assay Reference Genome
Location

Chr.1:953540 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03348126_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

26155

Gene Name:

NOC2 like nucleolar associated transcriptional repressor

Gene Aliases:

NET15, NET7, NIR, PPP1R112

Location:

Chr.1:944203-959299 on Build GRCh38

Assay Gene Location:

Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NOC2L NM_015658.3 NP_056473.2
AK022756.1 BAB14230.1
AK024284.1
AK225239.1
AK315080.1
AL050019.1 CAB43240.2
BC003555.1 AAH03555.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2762302 Chr.1:914637 - 1106320 on Build GRCh38 Gain LOC105378948 PLEKHN1 PERM1 NOC2L RNF223 LOC100288175 LOC284600 KLHL17 LOC107985728 C1orf159 ISG15 HES4 LOC100130417 SAMD11 AGRN
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss ACAP3 MIR429 MIR200A CALML6 KLHL17 LOC105378592 FAAP20 TNFRSF4 LINC01342 FAM87B LOC100132287 CPTP TTLL10-AS1 TNFRSF18 CCNL2 LOC100506504 OR4F16 CDK11B TMEM88B LOC100288069 LOC105378591 MIR6723 MIR6808 ANKRD65 NADK PUSL1 LOC100130417 LOC100287934 LOC100134822 SDF4 LOC100133331 GABRD FAM132A DVL1 SCNN1D LOC105378949 LINC01128 PRKCZ LOC100288175 ISG15 SLC35E2 MMP23A LOC729737 MIR6859-1 LOC105378947 LOC148413 AURKAIP1 TTLL10 OR4F5 DDX11L1 CDK11A TMEM52 HES4 MIR6727 LOC102725121 AGRN SKI LOC107984841 LOC105378948 PLEKHN1 MIR200B RNF223 UBE2J2 LOC284600 VWA1 SAMD11 FAM138A MMP23B MTND1P23 WASH7P B3GALT6 MTND2P28 PERM1 TMEM240 ATAD3B NOC2L MORN1 FAM41C LOC107985728 C1orf159 LOC107985729 ATAD3A MIB2 CFAP74 OR4F29 MRPL20 MIR6726 CPSF3L LOC102724312 GNB1 FNDC10 SSU72 LINC00115 LOC100129534 MXRA8 MIR6859-2 ATAD3C SLC35E2B MIR1302-2 LOC101928626 LOC105378589 TAS1R3
nsv950451 Chr.1:899421 - 1094520 on Build GRCh38 Deletion LOC105378948 PLEKHN1 PERM1 NOC2L RNF223 LOC100288175 LOC284600 KLHL17 LOC107985728 C1orf159 ISG15 HES4 LOC100130417 SAMD11 AGRN
nsv509035 Chr.1:891406 - 985724 on Build GRCh38 Insertion PLEKHN1 LOC100130417 PERM1 SAMD11 NOC2L LOC284600 KLHL17 LOC107985728
nsv428334 Chr.1:874371 - 1220569 on Build GRCh38 Gain MIR429 SDF4 PERM1 NOC2L MIR200A TTLL10 FAM41C KLHL17 LOC107985728 C1orf159 TNFRSF4 LINC01342 HES4 AGRN TTLL10-AS1 TNFRSF18 LOC105378948 PLEKHN1 MIR200B RNF223 LOC100288175 LOC284600 ISG15 LOC100130417 SAMD11
dgv5n100 Chr.1:585989 - 1114424 on Build GRCh38 Gain LOC105378947 MTND2P28 PERM1 LOC100133331 NOC2L FAM41C KLHL17 LOC107985728 C1orf159 HES4 FAM87B AGRN LOC107984841 LOC105378948 LINC00115 PLEKHN1 LINC01128 OR4F16 RNF223 LOC100288175 LOC284600 LOC100288069 MIR6723 ISG15 LOC100130417 SAMD11 LOC100287934 LOC101928626 MTND1P23
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss LOC148413 ACAP3 MIR429 AURKAIP1 MIR200A TTLL10 KLHL17 CDK11A TNFRSF4 LINC01342 HES4 FAM87B MIR6727 AGRN CPTP TTLL10-AS1 TNFRSF18 CCNL2 LOC105378948 PLEKHN1 MIR200B RNF223 UBE2J2 CDK11B LOC284600 TMEM88B VWA1 LOC100288069 MIR6808 ANKRD65 NADK PUSL1 LOC100130417 SAMD11 LOC100287934 MMP23B B3GALT6 SDF4 PERM1 TMEM240 ATAD3B NOC2L FAM41C LOC107985728 C1orf159 LOC107985729 ATAD3A MIB2 FAM132A MRPL20 MIR6726 DVL1 CPSF3L LOC102724312 SCNN1D FNDC10 SSU72 LINC00115 LINC01128 LOC100288175 MXRA8 ISG15 SLC35E2 ATAD3C SLC35E2B TAS1R3 MMP23A
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss B3GALT6 MIR429 SDF4 PERM1 NOC2L MIR200A TTLL10 FAM41C KLHL17 LOC107985728 C1orf159 TNFRSF4 LINC01342 HES4 FAM132A FAM87B AGRN TTLL10-AS1 TNFRSF18 LOC105378948 LINC00115 PLEKHN1 LINC01128 MIR200B RNF223 UBE2J2 LOC100288175 LOC284600 ISG15 LOC100130417 SAMD11
nsv1013524 Chr.1:939510 - 1161955 on Build GRCh38 Gain LOC105378948 PLEKHN1 PERM1 NOC2L RNF223 LOC100288175 KLHL17 C1orf159 ISG15 LINC01342 HES4 SAMD11 AGRN
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain LOC148413 ACAP3 B3GALT6 MIR429 AURKAIP1 SDF4 PERM1 NOC2L MIR200A TTLL10 KLHL17 LOC107985728 C1orf159 TNFRSF4 LINC01342 HES4 FAM132A MRPL20 MIR6726 DVL1 CPSF3L MIR6727 SCNN1D AGRN CPTP TTLL10-AS1 TNFRSF18 CCNL2 LOC105378948 PLEKHN1 MIR200B RNF223 UBE2J2 LOC100288175 LOC284600 TMEM88B MXRA8 MIR6808 ANKRD65 ISG15 PUSL1 LOC100130417 SAMD11 TAS1R3
nsv544895 Chr.1:917392 - 1054900 on Build GRCh38 Loss ISG15 HES4 PLEKHN1 LOC100130417 PERM1 SAMD11 NOC2L LOC284600 KLHL17 LOC107985728 AGRN
nsv1160644 Chr.1:939652 - 1220663 on Build GRCh38 Deletion TTLL10-AS1 MIR429 TNFRSF18 LOC105378948 SDF4 PLEKHN1 PERM1 MIR200B NOC2L RNF223 MIR200A LOC100288175 TTLL10 KLHL17 C1orf159 TNFRSF4 ISG15 LINC01342 HES4 SAMD11 AGRN
nsv832980 Chr.1:848279 - 1007037 on Build GRCh38 Gain HES4 PLEKHN1 LINC01128 LOC100130417 PERM1 SAMD11 NOC2L LOC284600 FAM41C KLHL17 LOC107985728

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of B cell apoptotic process
transcription, DNA-templated
apoptotic process
chromatin assembly
nucleolus to nucleoplasm transport
cellular response to UV
negative regulation of histone acetylation
ribosomal large subunit biogenesis
regulation of signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway
chromatin binding
transcription corepressor activity
protein binding
nucleosome binding
histone binding
poly(A) RNA binding
repressing transcription factor binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline