This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03349098_cn
See other CHD1L CNV Assays ›
Gene Symbol
CHD1L
Assay Reference Genome
Location

Chr.1:147197189 on build GRCh38
Cytoband
1q21.1
Assay ID Hs03349098_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9557

Gene Name:

chromodomain helicase DNA binding protein 1 like

Gene Aliases:

ALC1, CHDL

Location:

Chr.1:147172804-147295766 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CHD1L XM_017002858.1 XP_016858347.1

Target Gene Details

Entrez Gene ID:

2330

Gene Name:

flavin containing monooxygenase 5

Gene Aliases:

-

Location:

Chr.1:147183963-147225798 on Build GRCh38

Assay Gene Location:

Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FMO5 NM_001144829.2 NP_001138301.1
NM_001144830.2 NP_001138302.1
NM_001461.3 NP_001452.2
XM_005272946.4 XP_005273003.1
XM_005272947.4 XP_005273004.1
XM_005272948.4 XP_005273005.1
XM_006711245.3 XP_006711308.1
XM_011509350.2 XP_011507652.1
XM_011509351.2 XP_011507653.1
XM_017000801.1 XP_016856290.1
XM_017000802.1 XP_016856291.1
AK222728.1 BAD96448.1
AK225739.1
AK314647.1
BC035687.1 AAH35687.1
L37080.1 AAA67849.1
Z47553.1 CAA87633.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv327n100 Chr.1:146028214 - 148860637 on Build GRCh38 Gain CH17-408M7.1 NOTCH2NL NBPF12 GJA5 GJA8 RNVU1-8 NUDT4P2 NUDT4P1 LOC105371242 MIR6077 CHD1L PPIAL4G PRKAB2 GPR89B RNVU1-3 PDE4DIP RNVU1-1 LOC105371226 FMO5 NBPF13P LINC01138 RNVU1-7 HYDIN2 RNVU1-6 LOC101928979 PDZK1P1 ACP6 RNU1-13P LOC100505824 LOC100996517 PDIA3P1 LINC00624 NBPF14 LOC100132057 NBPF11 NBPF10 LOC100996763 BCL9 LOC101927468 LOC728989 LOC284561 MIR5087
nsv1005055 Chr.1:145642988 - 148991068 on Build GRCh38 Loss NBPF12 RNVU1-8 NUDT4P2 NUDT4P1 CHD1L RNVU1-3 PDE4DIP NBPF13P RBM8A LINC01138 CD160 RNVU1-6 HFE2 PDZK1P1 ACP6 NUDT17 PDZK1 ANKRD34A LOC100132057 NBPF11 PIAS3 LOC100996763 BCL9 LOC101927468 LOC284561 MIR5087 CH17-408M7.1 NOTCH2NL GJA5 GJA8 LOC105371242 MIR6077 PPIAL4G PRKAB2 GPR89B RNF115 RNVU1-1 ITGA10 LOC105371226 FMO5 POLR3GL RNVU1-7 HYDIN2 GNRHR2 LOC101928979 GPR89A PEX11B POLR3C RNU1-13P LOC100505824 MIR6736 LOC100996517 PDIA3P1 LINC00624 NBPF14 TXNIP NBPF10 LIX1L LOC728989 ANKRD35
dgv67e199 Chr.1:147188956 - 147198583 on Build GRCh38 Deletion FMO5 CHD1L
nsv1121713 Chr.1:147010816 - 147207577 on Build GRCh38 Deletion PDIA3P1 FMO5 NBPF13P RNVU1-8 LOC728989 CHD1L PRKAB2
esv2761299 Chr.1:146068341 - 148926801 on Build GRCh38 Gain+Loss CH17-408M7.1 NOTCH2NL NBPF12 GJA5 GJA8 RNVU1-8 NUDT4P2 NUDT4P1 LOC105371242 MIR6077 CHD1L PPIAL4G PRKAB2 GPR89B RNVU1-3 PDE4DIP RNVU1-1 LOC105371226 FMO5 NBPF13P LINC01138 RNVU1-7 HYDIN2 PDZK1P1 ACP6 RNU1-13P LOC100505824 LOC100996517 PDIA3P1 LINC00624 NBPF14 LOC100132057 NBPF11 NBPF10 LOC100996763 BCL9 LOC101927468 LOC728989 LOC284561 MIR5087
nsv509457 Chr.1:145948024 - 149021821 on Build GRCh38 Insertion CH17-408M7.1 NOTCH2NL NBPF12 GJA5 GJA8 RNVU1-8 NUDT4P2 NUDT4P1 LOC105371242 MIR6077 CHD1L PPIAL4G PRKAB2 GPR89B RNVU1-3 PDE4DIP RNVU1-1 LOC105371226 FMO5 NBPF13P POLR3GL LINC01138 RNVU1-7 HYDIN2 RNVU1-6 HFE2 LOC101928979 PDZK1P1 ACP6 RNU1-13P LOC100505824 LOC100996517 PDIA3P1 LINC00624 NBPF14 ANKRD34A LOC100132057 NBPF11 TXNIP NBPF10 LIX1L LOC100996763 BCL9 LOC101927468 LOC728989 LOC284561 MIR5087
nsv1132304 Chr.1:146186094 - 148977674 on Build GRCh38 Duplication CH17-408M7.1 NOTCH2NL NBPF12 GJA5 GJA8 RNVU1-8 NUDT4P2 NUDT4P1 LOC105371242 MIR6077 CHD1L PPIAL4G PRKAB2 GPR89B RNVU1-3 PDE4DIP RNVU1-1 LOC105371226 FMO5 NBPF13P LINC01138 RNVU1-7 HYDIN2 PDZK1P1 ACP6 RNU1-13P LOC100505824 LOC100996517 PDIA3P1 LINC00624 NBPF14 LOC100132057 NBPF11 LOC100996763 BCL9 LOC101927468 LOC728989 LOC284561 MIR5087
nsv1003688 Chr.1:146036817 - 148934526 on Build GRCh38 Gain CH17-408M7.1 NOTCH2NL NBPF12 GJA5 GJA8 RNVU1-8 NUDT4P2 NUDT4P1 LOC105371242 MIR6077 CHD1L PPIAL4G PRKAB2 GPR89B RNVU1-3 PDE4DIP RNVU1-1 LOC105371226 FMO5 NBPF13P LINC01138 RNVU1-7 HYDIN2 RNVU1-6 LOC101928979 PDZK1P1 ACP6 RNU1-13P LOC100505824 LOC100996517 PDIA3P1 LINC00624 NBPF14 LOC100132057 NBPF11 NBPF10 LOC100996763 BCL9 LOC101927468 LOC728989 LOC284561 MIR5087
dgv66e199 Chr.1:146055970 - 149056514 on Build GRCh38 Deletion CH17-408M7.1 NOTCH2NL NBPF12 GJA5 GJA8 RNVU1-8 NUDT4P2 NUDT4P1 LOC105371242 MIR6077 CHD1L PPIAL4G PRKAB2 GPR89B RNVU1-3 PDE4DIP RNVU1-1 LOC105371226 FMO5 NBPF13P LINC01138 RNVU1-7 HYDIN2 NBPF9 LOC101928979 PDZK1P1 ACP6 RNU1-13P LOC100505824 LOC100996517 PDIA3P1 LINC00624 NBPF14 LOC100132057 NBPF11 NBPF10 LOC100996763 BCL9 LOC101927468 LOC728989 LOC284561 MIR5087
esv3587435 Chr.1:147189056 - 147198713 on Build GRCh38 Loss FMO5 CHD1L
nsv437047 Chr.1:147171976 - 147369886 on Build GRCh38 Loss PDIA3P1 FMO5 CHD1L PRKAB2

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
chromatin remodeling
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
global genome nucleotide-excision repair
biological_process
drug metabolic process
oxidation-reduction process
nucleotide binding
nucleic acid binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
ATPase activity
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline