This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03349682_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1264122 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03349682_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Intron 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss UBE2J2 MIR6726 SCNN1D ACAP3
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication MIR200B LOC105378948 C1orf159 AGRN FAM132A PUSL1 CPSF3L TTLL10-AS1 TTLL10 TNFRSF4 CPTP TNFRSF18 MIR6726 B3GALT6 MIR429 MIR200A MIR6727 TAS1R3 UBE2J2 LOC100288175 SCNN1D ACAP3 RNF223 SDF4 LINC01342
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss SLC35E2 KLHL17 ATAD3C AGRN LOC100287934 TMEM240 MRPL20 LINC00115 MIB2 PUSL1 ATAD3A CDK11A CPSF3L TTLL10-AS1 TTLL10 LOC284600 VWA1 DVL1 LOC100288069 TNFRSF4 CPTP SLC35E2B PERM1 TNFRSF18 ATAD3B NOC2L MIR6808 CCNL2 MIR429 SAMD11 MIR6727 TAS1R3 UBE2J2 LOC100288175 SCNN1D LOC100130417 LINC01342 CDK11B FNDC10 MIR200B LOC105378948 ANKRD65 LOC107985728 C1orf159 SSU72 MMP23B FAM132A MMP23A LOC107985729 LOC148413 LOC102724312 NADK AURKAIP1 FAM41C MIR6726 B3GALT6 LINC01128 FAM87B PLEKHN1 ISG15 MIR200A MXRA8 HES4 TMEM88B ACAP3 RNF223 SDF4
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss SLC35E2 KLHL17 ATAD3C TMEM240 MRPL20 TMEM52 MIB2 OR4F29 LOC100506504 ATAD3A CDK11A CPSF3L FAM138A DVL1 LOC100288069 LOC107984841 CPTP SLC35E2B PERM1 TNFRSF18 MIR429 SAMD11 CFAP74 TAS1R3 UBE2J2 LOC729737 LOC100288175 LOC100130417 PRKCZ MORN1 LINC01342 CDK11B LOC105378948 ANKRD65 FAAP20 SKI MMP23B LOC100129534 MIR6723 MTND1P23 LOC102724312 NADK FAM41C MIR6726 OR4F5 B3GALT6 LINC01128 FAM87B PLEKHN1 LOC105378947 MIR200A ACAP3 RNF223 GABRD MIR6859-2 AGRN GNB1 LOC100287934 LINC00115 PUSL1 TTLL10-AS1 TTLL10 LOC284600 MTND2P28 VWA1 TNFRSF4 CALML6 ATAD3B NOC2L MIR6808 CCNL2 MIR6727 LOC100134822 SCNN1D LOC105378949 FNDC10 MIR200B LOC100133331 LOC102725121 LOC105378592 WASH7P LOC105378589 LOC107985728 C1orf159 MIR6859-1 SSU72 OR4F16 FAM132A MMP23A LOC100132287 LOC107985729 LOC148413 LOC105378591 AURKAIP1 DDX11L1 LOC101928626 ISG15 MXRA8 HES4 TMEM88B MIR1302-2 SDF4
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss FAM132A UBE2J2 MIR6726 SCNN1D B3GALT6 ACAP3 SDF4
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss MIR200B KLHL17 LOC105378948 LOC107985728 C1orf159 AGRN LINC00115 FAM132A TTLL10-AS1 TTLL10 LOC284600 TNFRSF4 PERM1 TNFRSF18 FAM41C NOC2L B3GALT6 MIR429 LINC01128 FAM87B PLEKHN1 ISG15 SAMD11 MIR200A UBE2J2 LOC100288175 HES4 LOC100130417 RNF223 SDF4 LINC01342
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion MIR200B ANKRD65 C1orf159 ATAD3C SSU72 MMP23B TMEM240 MRPL20 FAM132A MIB2 PUSL1 ATAD3A LOC107985729 CPSF3L TTLL10-AS1 TTLL10 VWA1 DVL1 LOC148413 LOC102724312 TNFRSF4 CPTP TNFRSF18 AURKAIP1 ATAD3B MIR6808 CCNL2 MIR6726 B3GALT6 MIR429 MIR200A MXRA8 MIR6727 TAS1R3 UBE2J2 SCNN1D TMEM88B ACAP3 SDF4 LINC01342 CDK11B FNDC10
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss CPTP AURKAIP1 MIR6808 MIR6726 MXRA8 MIR6727 TAS1R3 UBE2J2 PUSL1 SCNN1D CPSF3L ACAP3 DVL1
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain MIR200B KLHL17 LOC105378948 ANKRD65 LOC107985728 C1orf159 AGRN MRPL20 FAM132A PUSL1 CPSF3L TTLL10-AS1 TTLL10 LOC284600 DVL1 LOC148413 TNFRSF4 CPTP PERM1 TNFRSF18 AURKAIP1 NOC2L MIR6808 CCNL2 MIR6726 B3GALT6 MIR429 PLEKHN1 ISG15 SAMD11 MIR200A MXRA8 MIR6727 TAS1R3 UBE2J2 LOC100288175 SCNN1D HES4 TMEM88B LOC100130417 ACAP3 RNF223 SDF4 LINC01342
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain MIR200B TNFRSF4 TNFRSF18 LOC105378948 B3GALT6 C1orf159 MIR429 MIR200A FAM132A UBE2J2 SCNN1D TTLL10-AS1 RNF223 SDF4 LINC01342 TTLL10
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss MIR200B TNFRSF4 TNFRSF18 MIR6726 B3GALT6 C1orf159 MIR429 MIR200A FAM132A UBE2J2 SCNN1D ACAP3 TTLL10-AS1 RNF223 SDF4 LINC01342 TTLL10
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain MIR200B TNFRSF4 TNFRSF18 MIR6726 B3GALT6 MIR429 MIR200A MIR6727 FAM132A UBE2J2 PUSL1 SCNN1D CPSF3L ACAP3 TTLL10-AS1 SDF4 TTLL10

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-65nqn:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline