This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03350665_cn
See other CHD1L CNV Assays ›
Gene Symbol
CHD1L
Assay Reference Genome
Location

Chr.1:147195614 on build GRCh38
Cytoband
1q21.1
Assay ID Hs03350665_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9557

Gene Name:

chromodomain helicase DNA binding protein 1 like

Gene Aliases:

ALC1, CHDL

Location:

Chr.1:147172804-147295766 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CHD1L XM_017002858.1 XP_016858347.1

Target Gene Details

Entrez Gene ID:

2330

Gene Name:

flavin containing monooxygenase 5

Gene Aliases:

-

Location:

Chr.1:147183963-147225798 on Build GRCh38

Assay Gene Location:

Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FMO5 NM_001144829.2 NP_001138301.1
NM_001144830.2 NP_001138302.1
NM_001461.3 NP_001452.2
XM_005272946.4 XP_005273003.1
XM_005272947.4 XP_005273004.1
XM_005272948.4 XP_005273005.1
XM_006711245.3 XP_006711308.1
XM_011509350.2 XP_011507652.1
XM_011509351.2 XP_011507653.1
XM_017000801.1 XP_016856290.1
XM_017000802.1 XP_016856291.1
AK222728.1 BAD96448.1
AK225739.1
AK314647.1
BC035687.1 AAH35687.1
L37080.1 AAA67849.1
Z47553.1 CAA87633.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv327n100 Chr.1:146028214 - 148860637 on Build GRCh38 Gain NBPF10 RNVU1-3 PDE4DIP MIR5087 LOC100996763 LINC01138 HYDIN2 PDZK1P1 LOC105371242 LOC100505824 NOTCH2NL LOC101927468 LOC100132057 RNVU1-8 NUDT4P2 LOC101928979 LOC284561 NBPF11 RNVU1-1 NBPF14 LOC728989 RNVU1-7 GJA8 NBPF12 RNU1-13P NUDT4P1 ACP6 FMO5 LOC105371226 GPR89B LINC00624 RNVU1-6 NBPF13P PPIAL4G LOC100996517 GJA5 MIR6077 CH17-408M7.1 CHD1L PRKAB2 PDIA3P1 BCL9
nsv1005055 Chr.1:145642988 - 148991068 on Build GRCh38 Loss TXNIP PDE4DIP LOC100996763 LINC01138 MIR6736 NUDT4P2 HFE2 LOC101928979 LOC284561 POLR3GL GNRHR2 RNVU1-1 LOC728989 ANKRD34A NBPF12 RNU1-13P NUDT4P1 ACP6 LOC105371226 LIX1L GPR89B LINC00624 RNVU1-6 NBPF13P MIR6077 CHD1L PDIA3P1 BCL9 NBPF10 ITGA10 RNVU1-3 MIR5087 ANKRD35 HYDIN2 PDZK1P1 PEX11B LOC105371242 LOC100505824 NOTCH2NL LOC101927468 LOC100132057 RNVU1-8 RNF115 GPR89A NBPF11 CD160 NUDT17 NBPF14 RNVU1-7 GJA8 FMO5 POLR3C RBM8A PPIAL4G LOC100996517 GJA5 CH17-408M7.1 PRKAB2 PIAS3 PDZK1
nsv1077657 Chr.1:147010816 - 147207577 on Build GRCh38 Deletion NBPF13P LOC728989 RNVU1-8 CHD1L PRKAB2 FMO5 PDIA3P1
esv2761299 Chr.1:146068341 - 148926801 on Build GRCh38 Gain+Loss NBPF10 RNVU1-3 PDE4DIP MIR5087 LOC100996763 LINC01138 HYDIN2 PDZK1P1 LOC105371242 LOC100505824 NOTCH2NL LOC101927468 LOC100132057 RNVU1-8 NUDT4P2 LOC284561 NBPF11 RNVU1-1 NBPF14 LOC728989 RNVU1-7 GJA8 NBPF12 RNU1-13P NUDT4P1 ACP6 FMO5 LOC105371226 GPR89B LINC00624 NBPF13P PPIAL4G LOC100996517 GJA5 MIR6077 CH17-408M7.1 CHD1L PRKAB2 PDIA3P1 BCL9
dgv67e199 Chr.1:147188956 - 147198583 on Build GRCh38 Deletion CHD1L FMO5
nsv509457 Chr.1:145948024 - 149021821 on Build GRCh38 Insertion TXNIP NBPF10 RNVU1-3 PDE4DIP MIR5087 LOC100996763 LINC01138 HYDIN2 PDZK1P1 LOC105371242 LOC100505824 NOTCH2NL LOC101927468 LOC100132057 RNVU1-8 NUDT4P2 HFE2 LOC101928979 LOC284561 NBPF11 POLR3GL RNVU1-1 NBPF14 LOC728989 RNVU1-7 GJA8 ANKRD34A NBPF12 RNU1-13P NUDT4P1 ACP6 FMO5 LOC105371226 LIX1L GPR89B LINC00624 RNVU1-6 NBPF13P PPIAL4G LOC100996517 GJA5 MIR6077 CH17-408M7.1 CHD1L PRKAB2 PDIA3P1 BCL9
nsv1132304 Chr.1:146186094 - 148977674 on Build GRCh38 Duplication RNVU1-3 PDE4DIP MIR5087 LOC100996763 LINC01138 HYDIN2 PDZK1P1 LOC105371242 LOC100505824 NOTCH2NL LOC101927468 LOC100132057 RNVU1-8 NUDT4P2 LOC284561 NBPF11 RNVU1-1 NBPF14 LOC728989 RNVU1-7 GJA8 NBPF12 RNU1-13P NUDT4P1 ACP6 FMO5 LOC105371226 GPR89B LINC00624 NBPF13P PPIAL4G LOC100996517 GJA5 MIR6077 CH17-408M7.1 CHD1L PRKAB2 PDIA3P1 BCL9
nsv1003688 Chr.1:146036817 - 148934526 on Build GRCh38 Gain NBPF10 RNVU1-3 PDE4DIP MIR5087 LOC100996763 LINC01138 HYDIN2 PDZK1P1 LOC105371242 LOC100505824 NOTCH2NL LOC101927468 LOC100132057 RNVU1-8 NUDT4P2 LOC101928979 LOC284561 NBPF11 RNVU1-1 NBPF14 LOC728989 RNVU1-7 GJA8 NBPF12 RNU1-13P NUDT4P1 ACP6 FMO5 LOC105371226 GPR89B LINC00624 RNVU1-6 NBPF13P PPIAL4G LOC100996517 GJA5 MIR6077 CH17-408M7.1 CHD1L PRKAB2 PDIA3P1 BCL9
dgv66e199 Chr.1:146055970 - 149056514 on Build GRCh38 Deletion NBPF10 RNVU1-3 PDE4DIP MIR5087 LOC100996763 LINC01138 HYDIN2 PDZK1P1 LOC105371242 LOC100505824 NOTCH2NL LOC101927468 LOC100132057 RNVU1-8 NUDT4P2 LOC101928979 LOC284561 NBPF9 NBPF11 RNVU1-1 NBPF14 LOC728989 RNVU1-7 GJA8 NBPF12 RNU1-13P NUDT4P1 ACP6 FMO5 LOC105371226 GPR89B LINC00624 NBPF13P PPIAL4G LOC100996517 GJA5 MIR6077 CH17-408M7.1 CHD1L PRKAB2 PDIA3P1 BCL9
esv3587435 Chr.1:147189056 - 147198713 on Build GRCh38 Loss CHD1L FMO5
nsv437047 Chr.1:147171976 - 147369886 on Build GRCh38 Loss CHD1L PRKAB2 FMO5 PDIA3P1

Back To Top

More Information


Additional Information:

For this assay, SNP(s) [rs72708573] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
chromatin remodeling
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
global genome nucleotide-excision repair
biological_process
drug metabolic process
oxidation-reduction process
nucleotide binding
nucleic acid binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
ATPase activity
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0