This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03351009_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1257944 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03351009_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1
BQ881730.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss UBE2J2 MIR6726 SCNN1D ACAP3
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication MIR6726 TNFRSF18 RNF223 LOC100288175 FAM132A CPSF3L AGRN UBE2J2 MIR429 LINC01342 TTLL10-AS1 TNFRSF4 PUSL1 TTLL10 MIR200A MIR6727 ACAP3 LOC105378948 CPTP SCNN1D C1orf159 MIR200B SDF4 TAS1R3 B3GALT6
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss DVL1 TNFRSF18 SLC35E2 VWA1 KLHL17 AURKAIP1 FNDC10 CPSF3L AGRN MMP23B LINC01342 TTLL10-AS1 LINC00115 PUSL1 CDK11B NOC2L SAMD11 CDK11A TTLL10 MIR200A FAM87B ATAD3C LOC105378948 LOC148413 TMEM240 SLC35E2B SDF4 ATAD3A MIR6726 LOC107985729 NADK PLEKHN1 LINC01128 RNF223 LOC100288175 FAM132A FAM41C UBE2J2 MIR429 LOC107985728 LOC100287934 LOC284600 CCNL2 MMP23A TNFRSF4 TMEM88B MIR6727 MIR6808 ACAP3 LOC100130417 LOC100288069 ISG15 SSU72 MRPL20 ATAD3B CPTP SCNN1D ANKRD65 C1orf159 HES4 LOC102724312 MIR200B MIB2 TAS1R3 PERM1 B3GALT6 MXRA8
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss SLC35E2 GNB1 LINC00115 DDX11L1 PUSL1 FAM138A CDK11B SAMD11 FAM87B PRKCZ ATAD3C LOC105378948 FAAP20 MIR1302-2 LOC101928626 LOC148413 GABRD SLC35E2B SDF4 LOC100132287 OR4F29 ATAD3A MIR6859-1 MIR6726 LINC01128 RNF223 FAM132A OR4F5 UBE2J2 MIR429 LOC100506504 LOC107985728 LOC100133331 LOC729737 LOC100287934 MMP23A TMEM88B MIR6727 CFAP74 SSU72 MRPL20 CPTP ANKRD65 HES4 LOC102724312 B3GALT6 MXRA8 OR4F16 DVL1 TNFRSF18 TMEM52 CALML6 LOC105378591 VWA1 KLHL17 SKI AURKAIP1 FNDC10 CPSF3L AGRN MMP23B MTND2P28 LINC01342 TTLL10-AS1 MIR6859-2 LOC105378589 NOC2L CDK11A TTLL10 MIR200A LOC100134822 WASH7P LOC105378949 TMEM240 MIR6723 LOC107985729 NADK PLEKHN1 LOC107984841 MORN1 LOC100288175 LOC105378947 LOC100129534 LOC102725121 FAM41C LOC105378592 LOC284600 CCNL2 TNFRSF4 MIR6808 ACAP3 LOC100130417 LOC100288069 MTND1P23 ISG15 ATAD3B SCNN1D C1orf159 MIR200B MIB2 TAS1R3 PERM1
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss UBE2J2 MIR6726 SCNN1D SDF4 ACAP3 FAM132A B3GALT6
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss TNFRSF18 PLEKHN1 LINC01128 RNF223 KLHL17 LOC100288175 FAM132A FAM41C AGRN UBE2J2 MIR429 LOC107985728 LINC01342 TTLL10-AS1 LOC284600 LINC00115 TNFRSF4 NOC2L SAMD11 TTLL10 MIR200A FAM87B LOC100130417 LOC105378948 ISG15 C1orf159 HES4 MIR200B SDF4 PERM1 B3GALT6
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion ATAD3A MIR6726 DVL1 LOC107985729 TNFRSF18 VWA1 AURKAIP1 FAM132A FNDC10 CPSF3L UBE2J2 MMP23B MIR429 LINC01342 TTLL10-AS1 CCNL2 TNFRSF4 PUSL1 CDK11B TTLL10 MIR200A TMEM88B MIR6727 MIR6808 ACAP3 ATAD3C SSU72 LOC148413 MRPL20 ATAD3B CPTP SCNN1D TMEM240 ANKRD65 C1orf159 LOC102724312 MIR200B SDF4 MIB2 TAS1R3 B3GALT6 MXRA8
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain MIR6726 DVL1 TNFRSF18 PLEKHN1 RNF223 KLHL17 LOC100288175 AURKAIP1 FAM132A CPSF3L AGRN UBE2J2 MIR429 LOC107985728 LINC01342 TTLL10-AS1 LOC284600 CCNL2 TNFRSF4 PUSL1 NOC2L SAMD11 TTLL10 MIR200A TMEM88B MIR6727 MIR6808 ACAP3 LOC100130417 LOC105378948 ISG15 LOC148413 MRPL20 CPTP SCNN1D ANKRD65 C1orf159 HES4 MIR200B SDF4 TAS1R3 PERM1 B3GALT6 MXRA8
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain TNFRSF18 TTLL10 MIR200A RNF223 LOC105378948 FAM132A UBE2J2 MIR429 SCNN1D LINC01342 C1orf159 MIR200B TTLL10-AS1 SDF4 B3GALT6 TNFRSF4
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss MIR6726 TNFRSF18 TTLL10 MIR200A RNF223 ACAP3 FAM132A UBE2J2 MIR429 SCNN1D LINC01342 C1orf159 MIR200B TTLL10-AS1 SDF4 B3GALT6 TNFRSF4
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain PUSL1 MIR6726 TNFRSF18 TTLL10 MIR200A MIR6727 ACAP3 FAM132A CPSF3L UBE2J2 MIR429 SCNN1D MIR200B TTLL10-AS1 SDF4 B3GALT6 TNFRSF4

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-97vht:80/100.66.77.199:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline