This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03352195_cn
See other GNB1 CNV Assays ›
Gene Symbol
GNB1
Assay Reference Genome
Location

Chr.1:1799474 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03352195_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2782

Gene Name:

G protein subunit beta 1

Gene Aliases:

MRD42

Location:

Chr.1:1785285-1891117 on Build GRCh38

Assay Gene Location:

Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GNB1 NM_001282538.1 NP_001269467.1
NM_001282539.1 NP_001269468.1
NM_002074.4 NP_002065.1
XM_017001059.1 XP_016856548.1
XM_017001060.1 XP_016856549.1
XM_017001061.1 XP_016856550.1
AF070603.1 AAC28655.1
AF501882.1 AAM15918.1
AK122949.1
AK312614.1
BC004186.2 AAH04186.1
BC005888.2 AAH05888.1
BC008991.1 AAH08991.1
BC114618.1 AAI14619.1
BT007305.1 AAP35969.1
CR456784.1 CAG33065.1
M36430.1 AAA63265.1
X04526.1 CAA28207.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion SLC35E2B FNDC10 CDK11A SLC35E2 MMP23A GNB1 CDK11B MIB2 NADK MMP23B
nsv834091 Chr.1:1701359 - 1880638 on Build GRCh38 Loss CDK11A SLC35E2 MMP23A GNB1 NADK
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication SSU72 SLC35E2B FNDC10 CDK11A SLC35E2 MMP23A GNB1 CDK11B MIB2 NADK MMP23B
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss PRKCZ MTND1P23 FAM41C LOC101928626 CCNL2 TMEM240 VWA1 LOC105378948 LOC102725121 TNFRSF18 B3GALT6 SAMD11 LOC105378591 CPTP MORN1 KLHL17 MIR200A LOC100133331 MIR6723 LOC284600 SLC35E2 LOC729737 LOC107985729 PUSL1 LOC100134822 TTLL10 LOC105378592 ATAD3C WASH7P LINC01128 DVL1 MIR200B LOC100506504 FAM87B OR4F16 GABRD LINC01342 LOC105378949 LOC100129534 TTLL10-AS1 PLEKHN1 TAS1R3 PERM1 SCNN1D ANKRD65 ATAD3B HES4 SLC35E2B MMP23A CALML6 MIB2 TMEM52 AURKAIP1 SSU72 ATAD3A UBE2J2 MXRA8 LOC102724312 MTND2P28 LOC105378947 LINC00115 DDX11L1 LOC105378589 TMEM88B OR4F5 SKI GNB1 ACAP3 MIR6859-1 OR4F29 MMP23B CFAP74 CPSF3L FNDC10 C1orf159 RNF223 CDK11B SDF4 LOC107984841 MIR6727 LOC100288069 LOC148413 CDK11A LOC100132287 MIR6808 LOC100130417 FAM132A LOC100288175 LOC100287934 ISG15 FAM138A FAAP20 MIR1302-2 AGRN NADK LOC107985728 NOC2L TNFRSF4 MIR6859-2 MRPL20 MIR429 MIR6726
nsv1160926 Chr.1:1772699 - 1905488 on Build GRCh38 Duplication LOC105378949 GNB1 NADK
dgv10n100 Chr.1:1779975 - 1884542 on Build GRCh38 Gain GNB1 NADK
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss CFAP74 PRKCZ FNDC10 SLC35E2 CDK11B LOC107985729 ANKRD65 ATAD3B LOC148413 CCNL2 SLC35E2B CDK11A TMEM240 MMP23A VWA1 CALML6 MIB2 ATAD3C TMEM52 SSU72 ATAD3A LOC102724312 NADK LOC105378591 LOC105378589 GABRD LOC105378949 TMEM88B GNB1 MRPL20 MMP23B

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

protein folding
signal transduction
adenylate cyclase-activating dopamine receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
G-protein coupled acetylcholine receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
Ras protein signal transduction
cell proliferation
cardiac muscle cell apoptotic process
rhodopsin mediated signaling pathway
platelet activation
sensory perception of taste
retina development in camera-type eye
cellular response to glucagon stimulus
cellular response to prostaglandin E stimulus
cellular response to hypoxia
cellular response to catecholamine stimulus
GTPase activity
signal transducer activity
protein binding
spectrin binding
type 1 angiotensin receptor binding
protein complex binding
GTPase binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline