This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03354605_cn
See other TNFRSF4 CNV Assays ›
Gene Symbol
TNFRSF4
Assay Reference Genome
Location

Chr.1:1214430 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03354605_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

7293

Gene Name:

TNF receptor superfamily member 4

Gene Aliases:

ACT35, CD134, IMD16, OX40, TXGP1L

Location:

Chr.1:1211326-1216812 on Build GRCh38

Assay Gene Location:

Within Exon 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
TNFRSF4 XM_011542075.2 1 206 XP_011540377.1
XM_017002231.1 XP_016857720.1
XM_017002232.1 XP_016857721.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication TNFRSF4 LOC105378948 SDF4 TTLL10-AS1 UBE2J2 SCNN1D PUSL1 TNFRSF18 LINC01342 RNF223 B3GALT6 TAS1R3 ACAP3 MIR6726 TTLL10 MIR429 MIR200B FAM132A LOC100288175 MIR6727 CPSF3L AGRN CPTP MIR200A C1orf159
nsv428334 Chr.1:874371 - 1220569 on Build GRCh38 Gain HES4 TNFRSF4 LOC105378948 SDF4 FAM41C PERM1 TTLL10-AS1 LOC284600 NOC2L TNFRSF18 LINC01342 LOC100130417 RNF223 SAMD11 PLEKHN1 TTLL10 MIR429 MIR200B ISG15 LOC100288175 LOC107985728 KLHL17 AGRN MIR200A C1orf159
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss HES4 CDK11B LOC105378948 SDF4 PERM1 MMP23B TMEM240 MIB2 MMP23A UBE2J2 VWA1 LOC284600 NOC2L TMEM88B LOC100130417 RNF223 SSU72 LOC107985729 B3GALT6 LOC102724312 NADK ATAD3C LINC00115 FAM87B TTLL10 MIR200B ISG15 CPTP MIR200A SLC35E2 C1orf159 TNFRSF4 FAM41C LOC148413 ATAD3A LOC100288069 FNDC10 TTLL10-AS1 AURKAIP1 SCNN1D PUSL1 TNFRSF18 LINC01342 SLC35E2B ATAD3B SAMD11 TAS1R3 ACAP3 LOC100287934 DVL1 MIR6726 PLEKHN1 MRPL20 MIR6808 ANKRD65 MXRA8 MIR429 FAM132A LOC100288175 LOC107985728 MIR6727 KLHL17 CPSF3L AGRN CCNL2 LINC01128 CDK11A
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss CDK11B LOC105378948 SDF4 CALML6 TMEM240 MMP23A UBE2J2 VWA1 NOC2L TMEM88B LOC100130417 TMEM52 SSU72 LOC107985729 B3GALT6 FAAP20 OR4F16 MIR1302-2 NADK ATAD3C MTND2P28 LINC00115 LOC107984841 LOC100132287 MIR200B ISG15 LOC100134822 SKI CPTP MIR200A SLC35E2 LOC100506504 TNFRSF4 LOC105378591 ATAD3A LOC100288069 WASH7P PUSL1 LOC105378947 MORN1 SLC35E2B ATAD3B SAMD11 ACAP3 OR4F29 LOC105378949 DVL1 PLEKHN1 MIR6808 MIR429 FAM132A LOC100288175 KLHL17 CPSF3L LOC105378589 AGRN LINC01128 HES4 PERM1 MMP23B LOC101928626 DDX11L1 MIB2 LOC284600 FAM138A MIR6723 RNF223 LOC102724312 LOC100129534 FAM87B TTLL10 LOC102725121 C1orf159 FAM41C PRKCZ LOC100133331 LOC148413 FNDC10 MIR6859-2 TTLL10-AS1 AURKAIP1 GABRD SCNN1D TNFRSF18 LINC01342 MTND1P23 TAS1R3 LOC100287934 LOC105378592 LOC729737 CFAP74 MIR6726 MRPL20 ANKRD65 MXRA8 LOC107985728 MIR6727 GNB1 CCNL2 MIR6859-1 CDK11A OR4F5
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion TNFRSF4 CDK11B SDF4 LOC148413 ATAD3A FNDC10 TTLL10-AS1 AURKAIP1 MMP23B TMEM240 MIB2 UBE2J2 VWA1 SCNN1D PUSL1 TNFRSF18 TMEM88B LINC01342 SSU72 ATAD3B LOC107985729 B3GALT6 TAS1R3 ACAP3 LOC102724312 DVL1 ATAD3C MIR6726 MRPL20 MIR6808 ANKRD65 MXRA8 TTLL10 MIR429 MIR200B FAM132A MIR6727 CPSF3L CPTP CCNL2 MIR200A C1orf159
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss HES4 TNFRSF4 LOC105378948 SDF4 FAM41C PERM1 TTLL10-AS1 UBE2J2 LOC284600 NOC2L TNFRSF18 LINC01342 LOC100130417 RNF223 SAMD11 B3GALT6 PLEKHN1 LINC00115 FAM87B TTLL10 MIR429 MIR200B ISG15 FAM132A LOC100288175 LOC107985728 KLHL17 AGRN LINC01128 MIR200A C1orf159
nsv1160644 Chr.1:939652 - 1220663 on Build GRCh38 Deletion HES4 TNFRSF4 LOC105378948 SDF4 PERM1 TTLL10-AS1 PLEKHN1 TTLL10 MIR429 MIR200B ISG15 NOC2L TNFRSF18 LOC100288175 LINC01342 KLHL17 RNF223 AGRN MIR200A SAMD11 C1orf159
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain HES4 TNFRSF4 LOC105378948 SDF4 LOC148413 PERM1 TTLL10-AS1 AURKAIP1 UBE2J2 SCNN1D PUSL1 LOC284600 NOC2L TNFRSF18 TMEM88B LINC01342 LOC100130417 RNF223 SAMD11 B3GALT6 TAS1R3 ACAP3 DVL1 MIR6726 PLEKHN1 MRPL20 MIR6808 ANKRD65 MXRA8 TTLL10 MIR429 MIR200B ISG15 FAM132A LOC100288175 LOC107985728 MIR6727 KLHL17 CPSF3L AGRN CPTP CCNL2 MIR200A C1orf159
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain TNFRSF4 LOC105378948 SDF4 TTLL10-AS1 TTLL10 MIR429 UBE2J2 SCNN1D MIR200B FAM132A TNFRSF18 LINC01342 RNF223 MIR200A B3GALT6 C1orf159
nsv470680 Chr.1:1070426 - 1378635 on Build GRCh38 Loss TNFRSF4 SDF4 TTLL10-AS1 AURKAIP1 UBE2J2 SCNN1D PUSL1 TNFRSF18 LINC01342 RNF223 B3GALT6 TAS1R3 ACAP3 DVL1 MIR6726 MIR6808 MXRA8 TTLL10 MIR429 MIR200B FAM132A MIR6727 CPSF3L CPTP MIR200A C1orf159
nsv544956 Chr.1:1171093 - 1241217 on Build GRCh38 Loss TTLL10 TNFRSF4 SDF4 TNFRSF18 TTLL10-AS1 B3GALT6
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain TNFRSF4 SDF4 MIR6726 TTLL10-AS1 TTLL10 MIR429 UBE2J2 SCNN1D PUSL1 MIR200B FAM132A TNFRSF18 MIR6727 CPSF3L MIR200A B3GALT6 ACAP3

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

inflammatory response
immune response
multicellular organism development
positive regulation of B cell proliferation
response to lipopolysaccharide
tumor necrosis factor-mediated signaling pathway
T cell proliferation
regulation of cell proliferation
regulation of apoptotic process
positive regulation of MAPK cascade
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of transcription, DNA-templated
viral entry into host cell
positive regulation of immunoglobulin secretion
apoptotic signaling pathway
virus receptor activity
tumor necrosis factor-activated receptor activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0