This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03355729_cn
See other NOC2L CNV Assays ›
Gene Symbol
NOC2L
Assay Reference Genome
Location

Chr.1:944932 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03355729_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

26155

Gene Name:

NOC2 like nucleolar associated transcriptional repressor

Gene Aliases:

NET15, NET7, NIR, PPP1R112

Location:

Chr.1:944203-959299 on Build GRCh38

Assay Gene Location:

Within Intron 18
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NOC2L NM_015658.3 NP_056473.2
AK022756.1 BAB14230.1
AK024284.1
AK092491.1 BAC03905.1
AK225239.1
AK315080.1
AL050019.1 CAB43240.2
BC003555.1 AAH03555.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2762302 Chr.1:914637 - 1106320 on Build GRCh38 Gain ISG15 AGRN HES4 NOC2L SAMD11 LOC105378948 KLHL17 LOC100130417 LOC284600 LOC100288175 C1orf159 LOC107985728 PLEKHN1 PERM1 RNF223
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss AURKAIP1 MRPL20 SLC35E2 OR4F16 LOC102725121 SCNN1D SKI ATAD3A ATAD3B CFAP74 DDX11L1 CDK11B FAM41C AGRN MIR6808 LOC107984841 TNFRSF18 FAM87B LOC729737 TTLL10 CCNL2 LOC148413 MIR6726 GNB1 ANKRD65 PERM1 MMP23B LOC100506504 TMEM52 PUSL1 LOC102724312 GABRD MIR429 LOC107985728 LINC01128 MXRA8 SLC35E2B SDF4 MIR6859-1 LINC00115 MORN1 HES4 LOC100133331 LOC105378948 LOC100130417 MTND2P28 LOC284600 LOC100288175 MIR6723 MTND1P23 CPSF3L ISG15 NOC2L SAMD11 TMEM240 NADK FAAP20 LOC105378949 LOC100134822 FAM132A ACAP3 LOC100132287 UBE2J2 LOC101928626 LOC100287934 MIR6859-2 CPTP LOC100288069 MMP23A FNDC10 OR4F5 MIR1302-2 LOC105378592 WASH7P LOC100129534 OR4F29 VWA1 TMEM88B CALML6 PRKCZ TNFRSF4 CDK11A KLHL17 LOC105378589 FAM138A MIR6727 DVL1 B3GALT6 TTLL10-AS1 LOC105378591 LOC105378947 LINC01342 LOC107985729 MIR200B SSU72 TAS1R3 C1orf159 MIR200A ATAD3C PLEKHN1 RNF223 MIB2
nsv950451 Chr.1:899421 - 1094520 on Build GRCh38 Deletion ISG15 AGRN HES4 NOC2L SAMD11 LOC105378948 KLHL17 LOC100130417 LOC284600 LOC100288175 C1orf159 LOC107985728 PLEKHN1 PERM1 RNF223
nsv509035 Chr.1:891406 - 985724 on Build GRCh38 Insertion LOC100130417 LOC284600 LOC107985728 NOC2L SAMD11 PLEKHN1 PERM1 KLHL17
dgv9n54 Chr.1:925754 - 952840 on Build GRCh38 Loss NOC2L SAMD11
nsv428334 Chr.1:874371 - 1220569 on Build GRCh38 Gain ISG15 TNFRSF4 NOC2L SAMD11 KLHL17 MIR429 LOC107985728 SDF4 TTLL10-AS1 FAM41C AGRN LINC01342 TNFRSF18 HES4 LOC105378948 TTLL10 MIR200B LOC100130417 LOC284600 LOC100288175 C1orf159 MIR200A PLEKHN1 PERM1 RNF223
dgv5n100 Chr.1:585989 - 1114424 on Build GRCh38 Gain ISG15 NOC2L SAMD11 KLHL17 OR4F16 LOC107985728 LINC01128 LOC101928626 LINC00115 LOC100287934 FAM41C LOC105378947 AGRN LOC107984841 HES4 FAM87B LOC100133331 LOC100288069 LOC105378948 LOC100130417 MTND2P28 LOC284600 LOC100288175 C1orf159 MIR6723 MTND1P23 PLEKHN1 PERM1 RNF223
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss ISG15 AURKAIP1 NOC2L SAMD11 TMEM240 MRPL20 NADK SLC35E2 SCNN1D FAM132A ACAP3 ATAD3A ATAD3B UBE2J2 LOC100287934 CDK11B FAM41C CPTP AGRN MIR6808 TNFRSF18 FAM87B LOC100288069 MMP23A FNDC10 TTLL10 CCNL2 LOC148413 MIR6726 ANKRD65 PERM1 MMP23B VWA1 TMEM88B TNFRSF4 CDK11A KLHL17 PUSL1 LOC102724312 MIR6727 MIR429 DVL1 LOC107985728 LINC01128 MXRA8 B3GALT6 SLC35E2B SDF4 TTLL10-AS1 LINC00115 LINC01342 HES4 LOC105378948 LOC107985729 MIR200B SSU72 LOC100130417 LOC284600 TAS1R3 LOC100288175 C1orf159 MIR200A ATAD3C PLEKHN1 RNF223 CPSF3L MIB2
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss ISG15 TNFRSF4 NOC2L SAMD11 KLHL17 FAM132A MIR429 LOC107985728 LINC01128 B3GALT6 UBE2J2 SDF4 TTLL10-AS1 LINC00115 FAM41C AGRN LINC01342 TNFRSF18 HES4 FAM87B LOC105378948 TTLL10 MIR200B LOC100130417 LOC284600 LOC100288175 C1orf159 MIR200A PLEKHN1 PERM1 RNF223
nsv1013524 Chr.1:939510 - 1161955 on Build GRCh38 Gain ISG15 AGRN LINC01342 HES4 NOC2L SAMD11 LOC105378948 KLHL17 LOC100288175 C1orf159 PLEKHN1 PERM1 RNF223
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain TMEM88B ISG15 AURKAIP1 TNFRSF4 NOC2L SAMD11 MRPL20 KLHL17 PUSL1 MIR6727 SCNN1D FAM132A ACAP3 MIR429 DVL1 LOC107985728 MXRA8 B3GALT6 UBE2J2 SDF4 TTLL10-AS1 CPTP AGRN LINC01342 MIR6808 TNFRSF18 HES4 LOC105378948 TTLL10 CCNL2 MIR200B LOC148413 LOC100130417 LOC284600 TAS1R3 LOC100288175 C1orf159 MIR200A MIR6726 ANKRD65 PLEKHN1 PERM1 RNF223 CPSF3L
nsv544895 Chr.1:917392 - 1054900 on Build GRCh38 Loss ISG15 LOC100130417 AGRN LOC284600 HES4 LOC107985728 NOC2L SAMD11 PLEKHN1 PERM1 KLHL17
nsv1160644 Chr.1:939652 - 1220663 on Build GRCh38 Deletion ISG15 AGRN LINC01342 TNFRSF18 HES4 TNFRSF4 NOC2L SAMD11 LOC105378948 TTLL10 KLHL17 MIR200B MIR429 LOC100288175 C1orf159 MIR200A SDF4 PLEKHN1 PERM1 TTLL10-AS1 RNF223
nsv832980 Chr.1:848279 - 1007037 on Build GRCh38 Gain FAM41C LOC100130417 LOC284600 HES4 LOC107985728 NOC2L SAMD11 LINC01128 PLEKHN1 PERM1 KLHL17

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of B cell apoptotic process
transcription, DNA-templated
apoptotic process
chromatin assembly
nucleolus to nucleoplasm transport
cellular response to UV
negative regulation of histone acetylation
ribosomal large subunit biogenesis
regulation of signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway
chromatin binding
transcription corepressor activity
protein binding
nucleosome binding
histone binding
poly(A) RNA binding
repressing transcription factor binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-65nqn:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline