This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03356306_cn
See other CD160 CNV Assays ›
Gene Symbol
CD160
Assay Reference Genome
Location

Chr.1:145720921 on build GRCh38
Cytoband
1q21.1
Assay ID Hs03356306_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

11126

Gene Name:

CD160 molecule

Gene Aliases:

BY55, NK1, NK28

Location:

Chr.1:145719312-145739288 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CD160 NM_007053.3 NP_008984.1
NR_103845.1
XM_005272929.3 XP_005272986.1
XM_011509104.2 XP_011507406.1
AF060981.1 AAC72302.1
AK128370.1
BC014465.1 AAH14465.1
CD638675.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1015025 Chr.1:145642988 - 145851919 on Build GRCh38 Gain PIAS3 RNF115 GPR89A PDZK1 NUDT17 CD160 POLR3C MIR6736
esv3587422 Chr.1:145649153 - 145751059 on Build GRCh38 Gain RNF115 GPR89A PDZK1 CD160
esv3572342 Chr.1:145601946 - 145809476 on Build GRCh38 Gain RNF115 GPR89A PDZK1 CD160 NBPF25P
nsv469752 Chr.1:145639438 - 145790042 on Build GRCh38 Loss RNF115 GPR89A PDZK1 CD160
nsv831392 Chr.1:145632374 - 145813165 on Build GRCh38 Gain RNF115 GPR89A PDZK1 CD160
esv3894089 Chr.1:145672100 - 145824903 on Build GRCh38 Loss RNF115 PDZK1 CD160 POLR3C
dgv531n54 Chr.1:145689573 - 145824903 on Build GRCh38 Loss RNF115 PDZK1 CD160 POLR3C
dgv532n54 Chr.1:145624561 - 145813526 on Build GRCh38 Gain RNF115 GPR89A PDZK1 CD160
dgv95e212 Chr.1:145631218 - 145812561 on Build GRCh38 Gain RNF115 GPR89A PDZK1 CD160
nsv470735 Chr.1:145687602 - 145809105 on Build GRCh38 Gain RNF115 PDZK1 CD160

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

cellular defense response
cell surface receptor signaling pathway
cell proliferation
regulation of immune response
defense response to Gram-negative bacterium
receptor activity
receptor binding
protein binding
MHC class I receptor activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline