This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03357545_cn
See other GNB1 CNV Assays ›
Gene Symbol
GNB1
Assay Reference Genome
Location

Chr.1:1796281 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03357545_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2782

Gene Name:

G protein subunit beta 1

Gene Aliases:

MRD42

Location:

Chr.1:1785285-1891117 on Build GRCh38

Assay Gene Location:

Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GNB1 NM_001282538.1 NP_001269467.1
NM_001282539.1 NP_001269468.1
NM_002074.4 NP_002065.1
XM_017001059.1 XP_016856548.1
XM_017001060.1 XP_016856549.1
XM_017001061.1 XP_016856550.1
AF070603.1 AAC28655.1
AF501882.1 AAM15918.1
AK122949.1
AK312614.1
BC004186.2 AAH04186.1
BC005888.2 AAH05888.1
BC008991.1 AAH08991.1
BC114618.1 AAI14619.1
BT007305.1 AAP35969.1
CR456784.1 CAG33065.1
M36430.1 AAA63265.1
X04526.1 CAA28207.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion SLC35E2B FNDC10 CDK11B GNB1 NADK MMP23B CDK11A MIB2 SLC35E2 MMP23A
nsv834091 Chr.1:1701359 - 1880638 on Build GRCh38 Loss GNB1 NADK CDK11A SLC35E2 MMP23A
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication SLC35E2B FNDC10 CDK11B GNB1 NADK MMP23B CDK11A MIB2 SLC35E2 MMP23A SSU72
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss ISG15 CALML6 LOC105378947 MIR6723 LINC00115 LINC01342 SLC35E2B LOC100288175 LOC100134822 ATAD3B HES4 GABRD LOC284600 FAM87B SDF4 LOC102724312 OR4F5 PRKCZ PERM1 TTLL10 CDK11A MIR6859-2 MIR6808 LINC01128 MIR1302-2 FAM41C LOC100129534 LOC100506504 PLEKHN1 LOC729737 MIR6727 LOC101928626 MIR6726 FNDC10 ANKRD65 CCNL2 TMEM88B MIR200A MXRA8 CPSF3L TNFRSF18 B3GALT6 ATAD3C SAMD11 TTLL10-AS1 TAS1R3 LOC102725121 MORN1 SCNN1D C1orf159 SSU72 DVL1 NOC2L DDX11L1 GNB1 PUSL1 TMEM240 MMP23A LOC105378948 CDK11B LOC100133331 MIR429 AURKAIP1 LOC107985728 ACAP3 OR4F29 RNF223 MIR6859-1 LOC100287934 LOC148413 MIB2 VWA1 KLHL17 OR4F16 LOC105378949 LOC105378592 AGRN WASH7P TNFRSF4 LOC100132287 LOC105378589 FAM138A LOC107985729 MRPL20 NADK MMP23B TMEM52 SLC35E2 SKI LOC105378591 MIR200B LOC107984841 MTND1P23 CPTP MTND2P28 CFAP74 FAM132A FAAP20 LOC100288069 LOC100130417 ATAD3A UBE2J2
nsv1160926 Chr.1:1772699 - 1905488 on Build GRCh38 Duplication GNB1 NADK LOC105378949
dgv10n100 Chr.1:1779975 - 1884542 on Build GRCh38 Gain GNB1 NADK
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss CALML6 FNDC10 ANKRD65 GNB1 CCNL2 TMEM240 TMEM88B LOC105378589 MMP23A SLC35E2B LOC107985729 ATAD3B MRPL20 CDK11B NADK MMP23B TMEM52 GABRD SLC35E2 LOC105378591 LOC102724312 ATAD3C PRKCZ LOC148413 CDK11A MIB2 VWA1 CFAP74 LOC105378949 ATAD3A SSU72

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

protein folding
signal transduction
adenylate cyclase-activating dopamine receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
G-protein coupled acetylcholine receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
Ras protein signal transduction
cell proliferation
cardiac muscle cell apoptotic process
rhodopsin mediated signaling pathway
platelet activation
sensory perception of taste
retina development in camera-type eye
cellular response to glucagon stimulus
cellular response to prostaglandin E stimulus
cellular response to hypoxia
cellular response to catecholamine stimulus
GTPase activity
signal transducer activity
protein binding
spectrin binding
type 1 angiotensin receptor binding
protein complex binding
GTPase binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline