This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03358215_cn
See other MIB2 CNV Assays ›
Gene Symbol
MIB2
Assay Reference Genome
Location

Chr.1:1620657 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03358215_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

142678

Gene Name:

mindbomb E3 ubiquitin protein ligase 2

Gene Aliases:

ZZANK1, ZZZ5

Location:

Chr.1:1615188-1630610 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MIB2 NM_001170686.1 NP_001164157.1
NM_001170687.1 NP_001164158.1
NM_001170688.1 NP_001164159.1
NM_001170689.1 NP_001164160.1
NM_080875.2 NP_543151.2
NR_033183.1
XM_006710372.1 XP_006710435.1
XM_011540731.2 XP_011539033.1
XM_011540736.2 XP_011539038.1
XM_011540737.2 XP_011539039.1
XM_011540741.2 XP_011539043.1
XM_011540742.2 XP_011539044.1
XM_017000349.1 XP_016855838.1
XM_017000350.1 XP_016855839.1
XM_017000351.1 XP_016855840.1
XM_017000352.1 XP_016855841.1
XM_017000353.1 XP_016855842.1
XM_017000354.1 XP_016855843.1
XM_017000355.1 XP_016855844.1
XM_017000356.1 XP_016855845.1
XM_017000357.1 XP_016855846.1
XM_017000358.1 XP_016855847.1
XM_017000359.1 XP_016855848.1
XM_017000360.1 XP_016855849.1
XM_017000361.1 XP_016855850.1
XM_017000362.1 XP_016855851.1
XM_017000363.1 XP_016855852.1
XM_017000364.1 XP_016855853.1
XM_017000365.1 XP_016855854.1
XM_017000366.1 XP_016855855.1
AB074480.1 BAB92950.1
AB076691.1 BAC00992.1
AB076692.1 BAC00993.1
AB076693.1 BAC00994.1
AB097000.1 BAC77353.1
AK098785.1 BAC05413.1
AK122751.1
AK122863.1
AK128167.1
AK128553.1
AK293642.1
AK302757.1
AK309362.1
BC016490.1 AAH16490.1
BC037542.1 AAH37542.1
DA391220.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion SLC35E2B CDK11B NADK MIB2 CDK11A MMP23B FNDC10 GNB1 MMP23A SLC35E2
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss SSU72 SLC35E2B CDK11B MIB2 CDK11A MMP23B FNDC10 MMP23A SLC35E2
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss FAM41C PLEKHN1 LOC100288175 VWA1 SCNN1D TNFRSF18 LOC100287934 MIR6727 NADK LINC01342 LINC00115 FAM87B ANKRD65 LOC107985728 LOC102724312 LOC100130417 KLHL17 MMP23A SLC35E2 MRPL20 CPTP LOC105378948 LOC148413 LOC107985729 TTLL10 HES4 FAM132A RNF223 MIR6726 AURKAIP1 LOC100288069 LOC284600 MIR200A PERM1 LINC01128 SLC35E2B MIR429 CCNL2 AGRN FNDC10 MIR6808 ATAD3B CPSF3L TNFRSF4 PUSL1 CDK11A MMP23B NOC2L SSU72 CDK11B SAMD11 C1orf159 TAS1R3 TMEM88B ATAD3A MXRA8 DVL1 SDF4 TTLL10-AS1 UBE2J2 ATAD3C B3GALT6 ACAP3 MIB2 TMEM240 MIR200B ISG15
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss FAM41C LOC100288175 SCNN1D MIR6727 CFAP74 LINC01342 MIR1302-2 LINC00115 FAM87B LOC100506504 MIR6723 KLHL17 MMP23A SLC35E2 CPTP LOC105378948 LOC105378589 LOC107985729 LOC100133331 TTLL10 RNF223 MIR6726 LOC101928626 LOC284600 LOC729737 MIR200A LINC01128 MIR429 CCNL2 MIR6808 ATAD3B FAM138A MTND2P28 CPSF3L TNFRSF4 PUSL1 CDK11A MMP23B GNB1 OR4F29 SSU72 CDK11B SAMD11 LOC107984841 C1orf159 TMEM88B LOC102725121 ATAD3A MXRA8 SKI ATAD3C PRKCZ ACAP3 MIB2 MORN1 TMEM240 MIR200B FAAP20 ISG15 PLEKHN1 VWA1 TNFRSF18 WASH7P LOC100287934 TMEM52 OR4F5 NADK ANKRD65 LOC107985728 LOC102724312 LOC100130417 LOC105378592 MRPL20 LOC148413 MIR6859-1 HES4 FAM132A AURKAIP1 LOC100129534 CALML6 LOC100288069 DDX11L1 LOC100132287 MIR6859-2 PERM1 SLC35E2B OR4F16 AGRN FNDC10 LOC105378947 GABRD NOC2L TAS1R3 LOC105378591 DVL1 LOC105378949 SDF4 TTLL10-AS1 UBE2J2 MTND1P23 LOC100134822 B3GALT6
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication SSU72 SLC35E2B CDK11B NADK MIB2 CDK11A MMP23B FNDC10 GNB1 MMP23A SLC35E2
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion MIR429 VWA1 SCNN1D CCNL2 TNFRSF18 FNDC10 MIR6727 MIR6808 ATAD3B CPSF3L LINC01342 TNFRSF4 ANKRD65 PUSL1 LOC102724312 MMP23B SSU72 CDK11B MRPL20 CPTP C1orf159 TAS1R3 TMEM88B LOC148413 LOC107985729 TTLL10 ATAD3A MXRA8 FAM132A DVL1 MIR6726 SDF4 TTLL10-AS1 UBE2J2 AURKAIP1 ATAD3C B3GALT6 ACAP3 MIB2 TMEM240 MIR200A MIR200B
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss SSU72 SLC35E2B CDK11B NADK MIB2 CDK11A MMP23B FNDC10 MMP23A SLC35E2
nsv545017 Chr.1:1615322 - 1663759 on Build GRCh38 Loss SLC35E2B CDK11B MIB2 MMP23B
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss SLC35E2B VWA1 CCNL2 FNDC10 TMEM52 CFAP74 ATAD3B NADK ANKRD65 CDK11A LOC102724312 MMP23B GABRD GNB1 MMP23A SLC35E2 SSU72 CDK11B MRPL20 TMEM88B LOC105378591 LOC148413 LOC105378589 LOC107985729 ATAD3A LOC105378949 CALML6 ATAD3C PRKCZ MIB2 TMEM240

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
Notch signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0