This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03359455_cn
See other BMP8A CNV Assays ›
Gene Symbol
BMP8A
Assay Reference Genome
Location

Chr.1:39497509 on build GRCh38
Cytoband
1p34.3
Assay ID Hs03359455_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

353500

Gene Name:

bone morphogenetic protein 8a

Gene Aliases:

-

Location:

Chr.1:39491626-39529869 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
BMP8A NM_181809.3 NP_861525.2
XM_006710616.3 XP_006710679.1
XM_011541381.2 XP_011539683.1
XM_011541382.2 XP_011539684.1
XM_017001198.1 XP_016856687.1
AY303954.1 AAP74559.1
BC131617.1

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic non-DGV Variation

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

ossification
diet induced thermogenesis
positive regulation of pathway-restricted SMAD protein phosphorylation
BMP signaling pathway
growth
regulation of apoptotic process
regulation of MAPK cascade
negative regulation of insulin secretion
cell development
cartilage development
SMAD protein signal transduction
regulation of energy homeostasis
cytokine activity
transforming growth factor beta receptor binding
growth factor activity
BMP receptor binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0