This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03360608_cn
See other GNB1 CNV Assays ›
Gene Symbol
GNB1
Assay Reference Genome
Location

Chr.1:1788179 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03360608_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2782

Gene Name:

G protein subunit beta 1

Gene Aliases:

MRD42

Location:

Chr.1:1785285-1891117 on Build GRCh38

Assay Gene Location:

Within Intron 11
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GNB1 NM_001282538.1 NP_001269467.1
NM_001282539.1 NP_001269468.1
NM_002074.4 NP_002065.1
XM_017001059.1 XP_016856548.1
XM_017001060.1 XP_016856549.1
XM_017001061.1 XP_016856550.1
AF070597.1 AAC28652.1
AF070603.1 AAC28655.1
AF501882.1 AAM15918.1
AK096614.1 3 2246
AK122949.1
AK312614.1
BC004186.2 AAH04186.1
BC005888.2 AAH05888.1
BC008991.1 AAH08991.1
BC114618.1 AAI14619.1
BT007305.1 AAP35969.1
CR456784.1 CAG33065.1
M36430.1 AAA63265.1
X04526.1 CAA28207.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion GNB1 CDK11A NADK MMP23A SLC35E2B MMP23B CDK11B MIB2 SLC35E2 FNDC10
nsv834091 Chr.1:1701359 - 1880638 on Build GRCh38 Loss GNB1 CDK11A NADK MMP23A SLC35E2
nsv951380 Chr.1:1785162 - 1791261 on Build GRCh38 Deletion GNB1
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication GNB1 CDK11A NADK MMP23A SLC35E2B MMP23B CDK11B MIB2 SLC35E2 FNDC10 SSU72
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss TTLL10-AS1 SAMD11 NOC2L SKI LINC01128 MTND1P23 LINC01342 MIR200A FAM138A LOC100288069 CFAP74 LOC100287934 PUSL1 PRKCZ TMEM240 SCNN1D MORN1 OR4F29 FAM41C AURKAIP1 LOC105378591 DVL1 OR4F16 ACAP3 TMEM88B TAS1R3 LOC100506504 FAM132A LINC00115 FAAP20 LOC105378948 MXRA8 SDF4 MIR429 MIR6859-1 MMP23B C1orf159 LOC102725121 UBE2J2 MTND2P28 SLC35E2B LOC107984841 SLC35E2 LOC105378592 LOC100134822 PLEKHN1 LOC100288175 AGRN CPSF3L NADK VWA1 RNF223 FNDC10 MIR6808 LOC100132287 MIR6727 LOC107985729 CCNL2 MIR6723 CDK11B MIR6726 LOC105378947 ATAD3C LOC148413 OR4F5 LOC102724312 TNFRSF18 ANKRD65 LOC284600 LOC729737 CDK11A LOC105378949 LOC105378589 MIB2 CPTP SSU72 CALML6 ISG15 TNFRSF4 LOC107985728 ATAD3B HES4 MIR200B B3GALT6 LOC100129534 GABRD MIR6859-2 FAM87B WASH7P LOC101928626 KLHL17 PERM1 GNB1 TTLL10 MIR1302-2 MMP23A LOC100130417 DDX11L1 TMEM52 ATAD3A LOC100133331 MRPL20
nsv1160926 Chr.1:1772699 - 1905488 on Build GRCh38 Duplication GNB1 NADK LOC105378949
dgv10n100 Chr.1:1779975 - 1884542 on Build GRCh38 Gain GNB1 NADK
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss LOC107985729 TMEM88B CCNL2 CDK11B ATAD3C LOC148413 GABRD MMP23B LOC102724312 CFAP74 PRKCZ ANKRD65 TMEM240 CDK11A LOC105378949 SLC35E2B LOC105378589 MIB2 SLC35E2 SSU72 LOC105378591 CALML6 GNB1 NADK MMP23A VWA1 FNDC10 ATAD3B TMEM52 ATAD3A MRPL20

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

protein folding
signal transduction
adenylate cyclase-activating dopamine receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
G-protein coupled acetylcholine receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
Ras protein signal transduction
cell proliferation
cardiac muscle cell apoptotic process
rhodopsin mediated signaling pathway
platelet activation
sensory perception of taste
retina development in camera-type eye
cellular response to glucagon stimulus
cellular response to prostaglandin E stimulus
cellular response to hypoxia
cellular response to catecholamine stimulus
GTPase activity
signal transducer activity
protein binding
spectrin binding
type 1 angiotensin receptor binding
protein complex binding
GTPase binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline