This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03361444_cn
See other CHD1L CNV Assays ›
Gene Symbol
CHD1L
Assay Reference Genome
Location

Chr.1:147198487 on build GRCh38
Cytoband
1q21.1
Assay ID Hs03361444_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9557

Gene Name:

chromodomain helicase DNA binding protein 1 like

Gene Aliases:

ALC1, CHDL

Location:

Chr.1:147172804-147295766 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CHD1L XM_017002858.1 XP_016858347.1

Target Gene Details

Entrez Gene ID:

2330

Gene Name:

flavin containing monooxygenase 5

Gene Aliases:

-

Location:

Chr.1:147183963-147225798 on Build GRCh38

Assay Gene Location:

Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FMO5 NM_001144829.2 NP_001138301.1
NM_001144830.2 NP_001138302.1
NM_001461.3 NP_001452.2
XM_005272946.4 XP_005273003.1
XM_005272947.4 XP_005273004.1
XM_005272948.4 XP_005273005.1
XM_006711245.3 XP_006711308.1
XM_011509350.2 XP_011507652.1
XM_011509351.2 XP_011507653.1
XM_017000801.1 XP_016856290.1
XM_017000802.1 XP_016856291.1
AK222728.1 BAD96448.1
AK225739.1
AK314647.1
BC035687.1 AAH35687.1
L37080.1 AAA67849.1
Z47553.1 CAA87633.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv327n100 Chr.1:146028214 - 148860637 on Build GRCh38 Gain LINC01138 PDIA3P1 NBPF13P LOC284561 FMO5 BCL9 LOC101927468 GJA8 LOC101928979 MIR5087 LOC100132057 NBPF14 PRKAB2 CH17-408M7.1 LOC100996517 RNVU1-7 MIR6077 RNVU1-1 ACP6 PDZK1P1 CHD1L NBPF12 LOC100505824 LINC00624 NBPF10 HYDIN2 RNU1-13P GJA5 GPR89B LOC105371226 RNVU1-8 PDE4DIP RNVU1-6 LOC728989 NOTCH2NL RNVU1-3 LOC100996763 NUDT4P1 NBPF11 NUDT4P2 LOC105371242 PPIAL4G
nsv1005055 Chr.1:145642988 - 148991068 on Build GRCh38 Loss LINC01138 PDZK1 FMO5 BCL9 LOC101927468 GPR89A ITGA10 GJA8 HFE2 CH17-408M7.1 LOC100996517 RNVU1-1 ACP6 PDZK1P1 CHD1L NBPF12 LINC00624 HYDIN2 PEX11B GJA5 GPR89B PDE4DIP RNVU1-6 NOTCH2NL RNVU1-3 NBPF11 PPIAL4G MIR6736 NUDT17 PDIA3P1 NBPF13P LOC284561 POLR3C CD160 ANKRD35 RBM8A LOC101928979 MIR5087 LOC100132057 NBPF14 PRKAB2 TXNIP LIX1L RNVU1-7 MIR6077 PIAS3 POLR3GL LOC100505824 NBPF10 RNU1-13P RNF115 LOC105371226 RNVU1-8 LOC728989 LOC100996763 NUDT4P1 NUDT4P2 LOC105371242 ANKRD34A GNRHR2
dgv67e199 Chr.1:147188956 - 147198583 on Build GRCh38 Deletion FMO5 CHD1L
nsv1121713 Chr.1:147010816 - 147207577 on Build GRCh38 Deletion PDIA3P1 NBPF13P PRKAB2 FMO5 LOC728989 CHD1L RNVU1-8
esv2761299 Chr.1:146068341 - 148926801 on Build GRCh38 Gain+Loss LINC01138 PDIA3P1 NBPF13P LOC284561 FMO5 BCL9 LOC101927468 GJA8 MIR5087 LOC100132057 NBPF14 PRKAB2 CH17-408M7.1 LOC100996517 RNVU1-7 MIR6077 RNVU1-1 ACP6 PDZK1P1 CHD1L NBPF12 LOC100505824 LINC00624 NBPF10 HYDIN2 RNU1-13P GJA5 GPR89B LOC105371226 RNVU1-8 PDE4DIP LOC728989 NOTCH2NL RNVU1-3 LOC100996763 NUDT4P1 NBPF11 NUDT4P2 LOC105371242 PPIAL4G
nsv509457 Chr.1:145948024 - 149021821 on Build GRCh38 Insertion LINC01138 PDIA3P1 NBPF13P LOC284561 FMO5 BCL9 LOC101927468 GJA8 LOC101928979 MIR5087 LOC100132057 HFE2 NBPF14 PRKAB2 CH17-408M7.1 LOC100996517 TXNIP LIX1L RNVU1-7 MIR6077 RNVU1-1 ACP6 PDZK1P1 POLR3GL CHD1L NBPF12 LOC100505824 LINC00624 NBPF10 HYDIN2 RNU1-13P GJA5 GPR89B LOC105371226 RNVU1-8 PDE4DIP RNVU1-6 LOC728989 NOTCH2NL RNVU1-3 LOC100996763 NUDT4P1 NBPF11 NUDT4P2 LOC105371242 ANKRD34A PPIAL4G
nsv1132304 Chr.1:146186094 - 148977674 on Build GRCh38 Duplication LINC01138 PDIA3P1 NBPF13P LOC284561 FMO5 BCL9 LOC101927468 GJA8 MIR5087 LOC100132057 NBPF14 PRKAB2 CH17-408M7.1 LOC100996517 RNVU1-7 MIR6077 RNVU1-1 ACP6 PDZK1P1 CHD1L NBPF12 LOC100505824 LINC00624 HYDIN2 RNU1-13P GJA5 GPR89B LOC105371226 RNVU1-8 PDE4DIP LOC728989 NOTCH2NL RNVU1-3 LOC100996763 NUDT4P1 NBPF11 NUDT4P2 LOC105371242 PPIAL4G
nsv1003688 Chr.1:146036817 - 148934526 on Build GRCh38 Gain LINC01138 PDIA3P1 NBPF13P LOC284561 FMO5 BCL9 LOC101927468 GJA8 LOC101928979 MIR5087 LOC100132057 NBPF14 PRKAB2 CH17-408M7.1 LOC100996517 RNVU1-7 MIR6077 RNVU1-1 ACP6 PDZK1P1 CHD1L NBPF12 LOC100505824 LINC00624 NBPF10 HYDIN2 RNU1-13P GJA5 GPR89B LOC105371226 RNVU1-8 PDE4DIP RNVU1-6 LOC728989 NOTCH2NL RNVU1-3 LOC100996763 NUDT4P1 NBPF11 NUDT4P2 LOC105371242 PPIAL4G
dgv66e199 Chr.1:146055970 - 149056514 on Build GRCh38 Deletion LINC01138 PDIA3P1 NBPF13P LOC284561 FMO5 BCL9 LOC101927468 GJA8 LOC101928979 MIR5087 LOC100132057 NBPF9 NBPF14 PRKAB2 CH17-408M7.1 LOC100996517 RNVU1-7 MIR6077 RNVU1-1 ACP6 PDZK1P1 CHD1L NBPF12 LOC100505824 LINC00624 NBPF10 HYDIN2 RNU1-13P GJA5 GPR89B LOC105371226 RNVU1-8 PDE4DIP LOC728989 NOTCH2NL RNVU1-3 LOC100996763 NUDT4P1 NBPF11 NUDT4P2 LOC105371242 PPIAL4G
esv3587435 Chr.1:147189056 - 147198713 on Build GRCh38 Loss FMO5 CHD1L
nsv437047 Chr.1:147171976 - 147369886 on Build GRCh38 Loss PDIA3P1 PRKAB2 FMO5 CHD1L

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
chromatin remodeling
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
global genome nucleotide-excision repair
biological_process
drug metabolic process
oxidation-reduction process
nucleotide binding
nucleic acid binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
ATPase activity
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0