This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03361635_cn
See other NOC2L CNV Assays ›
Gene Symbol
NOC2L
Assay Reference Genome
Location

Chr.1:955299 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03361635_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

26155

Gene Name:

NOC2 like nucleolar associated transcriptional repressor

Gene Aliases:

NET15, NET7, NIR, PPP1R112

Location:

Chr.1:944203-959299 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NOC2L NM_015658.3 NP_056473.2
AK024284.1
AK225239.1
AK315080.1
AL050019.1 CAB43240.2
BC003555.1 AAH03555.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2762302 Chr.1:914637 - 1106320 on Build GRCh38 Gain LOC100288175 C1orf159 KLHL17 ISG15 NOC2L HES4 LOC107985728 PLEKHN1 LOC284600 PERM1 SAMD11 AGRN LOC105378948 RNF223 LOC100130417
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC105378949 NADK LOC102724312 MIR6859-1 SLC35E2 SKI LOC105378947 LOC100506504 LOC105378591 MXRA8 FAM87B LOC100288175 LOC148413 MIR6808 NOC2L CPTP OR4F16 LOC105378592 LOC100133331 MIR1302-2 LOC107985729 LOC100287934 LOC284600 WASH7P PRKCZ OR4F5 ACAP3 TTLL10 HES4 LOC107985728 MORN1 CDK11B TMEM240 TMEM88B FAM41C FAM138A MMP23B LINC00115 LOC107984841 SDF4 RNF223 ANKRD65 LOC100129534 TNFRSF4 MIR6726 TNFRSF18 SSU72 LOC105378589 FAM132A ATAD3A B3GALT6 GNB1 FAAP20 LOC100134822 AGRN TMEM52 ATAD3B LINC01342 C1orf159 KLHL17 ISG15 PLEKHN1 LOC101928626 TTLL10-AS1 CPSF3L MIR429 CDK11A MMP23A ATAD3C MIR6723 CFAP74 MTND2P28 OR4F29 MIR6859-2 VWA1 LOC105378948 MIR200B CCNL2 AURKAIP1 DDX11L1 LOC729737 TAS1R3 PUSL1 LOC100288069 MIR6727 PERM1 MTND1P23 SCNN1D SAMD11 LOC102725121 UBE2J2 MIB2 MRPL20 LOC100130417 GABRD LOC100132287 SLC35E2B MIR200A CALML6 LINC01128 FNDC10 DVL1
nsv950451 Chr.1:899421 - 1094520 on Build GRCh38 Deletion LOC100288175 C1orf159 KLHL17 ISG15 NOC2L HES4 LOC107985728 PLEKHN1 LOC284600 PERM1 SAMD11 AGRN LOC105378948 RNF223 LOC100130417
nsv509035 Chr.1:891406 - 985724 on Build GRCh38 Insertion KLHL17 PERM1 NOC2L SAMD11 LOC107985728 PLEKHN1 LOC284600 LOC100130417
nsv428334 Chr.1:874371 - 1220569 on Build GRCh38 Gain HES4 LOC107985728 FAM41C PERM1 SAMD11 AGRN SDF4 RNF223 LOC100130417 LINC01342 LOC100288175 C1orf159 KLHL17 ISG15 NOC2L PLEKHN1 TTLL10-AS1 MIR429 LOC284600 TNFRSF4 MIR200A TNFRSF18 LOC105378948 MIR200B TTLL10
dgv5n100 Chr.1:585989 - 1114424 on Build GRCh38 Gain HES4 LOC107985728 FAM41C LOC100288069 PERM1 MTND1P23 SAMD11 LINC00115 AGRN LOC105378947 LOC107984841 RNF223 LOC100130417 FAM87B LOC100288175 C1orf159 KLHL17 ISG15 NOC2L OR4F16 PLEKHN1 LOC100133331 LOC101928626 LOC100287934 LOC284600 MIR6723 MTND2P28 LINC01128 LOC105378948
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss ATAD3A NADK B3GALT6 LOC102724312 SLC35E2 AGRN MXRA8 ATAD3B LINC01342 FAM87B LOC100288175 C1orf159 KLHL17 LOC148413 MIR6808 ISG15 NOC2L CPTP PLEKHN1 TTLL10-AS1 LOC107985729 CPSF3L MIR429 CDK11A LOC100287934 MMP23A LOC284600 ATAD3C VWA1 LOC105378948 MIR200B CCNL2 ACAP3 AURKAIP1 TTLL10 HES4 LOC107985728 CDK11B TMEM240 TAS1R3 TMEM88B FAM41C PUSL1 LOC100288069 MIR6727 MMP23B PERM1 SCNN1D SAMD11 LINC00115 UBE2J2 MIB2 SDF4 MRPL20 RNF223 LOC100130417 ANKRD65 SLC35E2B TNFRSF4 MIR200A MIR6726 TNFRSF18 LINC01128 SSU72 FNDC10 DVL1 FAM132A
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss HES4 LOC107985728 B3GALT6 FAM41C PERM1 SAMD11 LINC00115 AGRN UBE2J2 SDF4 RNF223 LOC100130417 LINC01342 FAM87B LOC100288175 C1orf159 KLHL17 ISG15 NOC2L PLEKHN1 TTLL10-AS1 MIR429 LOC284600 TNFRSF4 MIR200A TNFRSF18 LINC01128 LOC105378948 MIR200B TTLL10 FAM132A
nsv1013524 Chr.1:939510 - 1161955 on Build GRCh38 Gain LINC01342 LOC100288175 C1orf159 KLHL17 ISG15 NOC2L HES4 PLEKHN1 PERM1 SAMD11 AGRN LOC105378948 RNF223
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain HES4 LOC107985728 B3GALT6 TAS1R3 TMEM88B PUSL1 MIR6727 PERM1 SCNN1D SAMD11 AGRN UBE2J2 MXRA8 SDF4 MRPL20 RNF223 LOC100130417 LINC01342 LOC100288175 C1orf159 KLHL17 LOC148413 MIR6808 ISG15 NOC2L CPTP ANKRD65 PLEKHN1 TTLL10-AS1 CPSF3L MIR429 LOC284600 TNFRSF4 MIR200A MIR6726 TNFRSF18 LOC105378948 MIR200B CCNL2 ACAP3 DVL1 AURKAIP1 TTLL10 FAM132A
nsv544895 Chr.1:917392 - 1054900 on Build GRCh38 Loss KLHL17 PERM1 ISG15 NOC2L HES4 SAMD11 AGRN LOC107985728 PLEKHN1 LOC284600 LOC100130417
nsv1160644 Chr.1:939652 - 1220663 on Build GRCh38 Deletion LINC01342 LOC100288175 C1orf159 KLHL17 ISG15 NOC2L HES4 PLEKHN1 TTLL10-AS1 MIR429 TNFRSF4 PERM1 SAMD11 AGRN MIR200A TNFRSF18 LOC105378948 MIR200B SDF4 TTLL10 RNF223
nsv832980 Chr.1:848279 - 1007037 on Build GRCh38 Gain KLHL17 PERM1 NOC2L HES4 SAMD11 LOC107985728 PLEKHN1 LINC01128 FAM41C LOC284600 LOC100130417

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of B cell apoptotic process
transcription, DNA-templated
apoptotic process
chromatin assembly
nucleolus to nucleoplasm transport
cellular response to UV
negative regulation of histone acetylation
ribosomal large subunit biogenesis
regulation of signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway
chromatin binding
transcription corepressor activity
protein binding
nucleosome binding
histone binding
poly(A) RNA binding
repressing transcription factor binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline