This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03361941_cn
See other GPR153 CNV Assays ›
Gene Symbol
GPR153
Assay Reference Genome
Location

Chr.1:6248542 on build GRCh38
Cytoband
1p36.31
Assay ID Hs03361941_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

387509

Gene Name:

G protein-coupled receptor 153

Gene Aliases:

PGR1

Location:

Chr.1:6247346-6260975 on Build GRCh38

Assay Gene Location:

Within Exon 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GPR153 NM_207370.2 6 2886 NP_997253.2
XM_011541434.2 6 2990 XP_011539736.1
XM_017001250.1 5 2972 XP_016856739.1
CR627473.1 1 1522 CAH10674.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv8n67 Chr.1:6220621 - 6264303 on Build GRCh38 Gain ACOT7 ICMT RNF207 GPR153 HES3 LINC00337
nsv526371 Chr.1:6247195 - 6307159 on Build GRCh38 Loss ACOT7 GPR153
nsv831425 Chr.1:6129291 - 6264418 on Build GRCh38 Loss ACOT7 ICMT RPL22 RNF207 GPR153 LOC102724450 CHD5 HES3 LINC00337
nsv545236 Chr.1:6161915 - 6262876 on Build GRCh38 Loss ICMT RPL22 RNF207 GPR153 LOC102724450 CHD5 HES3 LINC00337
nsv545237 Chr.1:6240990 - 6250176 on Build GRCh38 Loss GPR153 HES3
nsv545238 Chr.1:6248510 - 6257923 on Build GRCh38 Loss GPR153
nsv508825 Chr.1:6221275 - 6286549 on Build GRCh38 Insertion ACOT7 ICMT RNF207 GPR153 HES3 LINC00337

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
G-protein coupled receptor activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline