This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03362245_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1258617 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03362245_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1
BQ881730.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss ACAP3 MIR6726 UBE2J2 SCNN1D
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication SDF4 MIR200A MIR6727 SCNN1D MIR200B TAS1R3 LOC100288175 LOC105378948 CPSF3L TTLL10-AS1 PUSL1 RNF223 MIR429 C1orf159 TNFRSF18 B3GALT6 CPTP FAM132A ACAP3 MIR6726 UBE2J2 TTLL10 TNFRSF4 AGRN LINC01342
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss FAM87B MMP23B MMP23A LOC148413 SDF4 ATAD3A TAS1R3 PERM1 TMEM88B HES4 LOC100130417 LOC284600 SAMD11 LOC100288175 LOC105378948 CDK11B CCNL2 CPSF3L LOC100287934 LINC01128 TTLL10-AS1 TMEM240 RNF223 C1orf159 SSU72 MXRA8 CPTP FAM132A KLHL17 NADK MIR6726 ANKRD65 TTLL10 AGRN DVL1 LINC00115 FAM41C AURKAIP1 MIB2 PLEKHN1 CDK11A NOC2L MIR200A MIR6727 MIR6808 VWA1 SCNN1D MIR200B LOC107985728 PUSL1 MIR429 TNFRSF18 B3GALT6 ATAD3B ACAP3 UBE2J2 ATAD3C TNFRSF4 SLC35E2 LOC107985729 ISG15 MRPL20 SLC35E2B LOC100288069 LOC102724312 LINC01342 FNDC10
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss CFAP74 LOC148413 SDF4 PERM1 TMEM88B HES4 LOC100130417 LOC100288175 CCNL2 CPSF3L LOC100287934 TTLL10-AS1 TMEM240 RNF223 MXRA8 KLHL17 ANKRD65 TTLL10 DVL1 LINC00115 OR4F29 FAM41C LOC105378591 AURKAIP1 OR4F5 FAM138A CDK11A PRKCZ MIR200A LOC100129534 VWA1 LOC101928626 LOC107984841 CALML6 LOC105378589 MIR6723 ATAD3B MIR6859-1 UBE2J2 ATAD3C TNFRSF4 MTND1P23 DDX11L1 GABRD LOC105378949 OR4F16 LOC100288069 LOC102724312 MORN1 LINC01342 FNDC10 FAM87B MMP23B MMP23A TMEM52 LOC105378947 ATAD3A TAS1R3 FAAP20 GNB1 LOC284600 SAMD11 LOC105378948 CDK11B MIR6859-2 LINC01128 C1orf159 LOC100134822 SSU72 CPTP FAM132A NADK MIR6726 AGRN LOC102725121 LOC105378592 MIB2 PLEKHN1 LOC100132287 NOC2L MIR6727 MIR6808 SCNN1D MIR200B LOC107985728 MTND2P28 SKI PUSL1 MIR429 MIR1302-2 TNFRSF18 LOC100506504 B3GALT6 LOC729737 WASH7P ACAP3 SLC35E2 LOC107985729 ISG15 MRPL20 SLC35E2B LOC100133331
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss B3GALT6 SDF4 FAM132A ACAP3 MIR6726 UBE2J2 SCNN1D
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss FAM87B NOC2L SDF4 MIR200A MIR200B LOC107985728 PERM1 HES4 LOC100130417 LOC284600 SAMD11 LOC100288175 LOC105378948 LINC01128 TTLL10-AS1 RNF223 MIR429 C1orf159 TNFRSF18 B3GALT6 FAM132A KLHL17 UBE2J2 TTLL10 TNFRSF4 AGRN ISG15 LINC00115 FAM41C PLEKHN1 LINC01342
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion MMP23B LOC148413 SDF4 MIR200A MIR6727 MIR6808 VWA1 SCNN1D ATAD3A MIR200B TAS1R3 TMEM88B CDK11B CCNL2 CPSF3L TTLL10-AS1 PUSL1 TMEM240 MIR429 C1orf159 TNFRSF18 SSU72 B3GALT6 MXRA8 CPTP FAM132A ATAD3B ACAP3 MIR6726 UBE2J2 ATAD3C ANKRD65 TTLL10 TNFRSF4 LOC107985729 MRPL20 DVL1 AURKAIP1 MIB2 LOC102724312 LINC01342 FNDC10
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain NOC2L LOC148413 SDF4 MIR200A MIR6727 MIR6808 SCNN1D MIR200B LOC107985728 TAS1R3 PERM1 TMEM88B HES4 LOC100130417 LOC284600 SAMD11 LOC100288175 LOC105378948 CCNL2 CPSF3L TTLL10-AS1 PUSL1 RNF223 MIR429 C1orf159 TNFRSF18 B3GALT6 MXRA8 CPTP FAM132A KLHL17 ACAP3 MIR6726 UBE2J2 ANKRD65 TTLL10 TNFRSF4 AGRN ISG15 MRPL20 DVL1 AURKAIP1 PLEKHN1 LINC01342
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain B3GALT6 SDF4 FAM132A MIR200A UBE2J2 SCNN1D MIR200B TTLL10 TNFRSF4 LOC105378948 TTLL10-AS1 RNF223 MIR429 C1orf159 TNFRSF18 LINC01342
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss B3GALT6 SDF4 FAM132A MIR200A ACAP3 MIR6726 UBE2J2 SCNN1D MIR200B TTLL10 TNFRSF4 TTLL10-AS1 RNF223 MIR429 C1orf159 TNFRSF18 LINC01342
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain B3GALT6 SDF4 FAM132A MIR200A MIR6727 ACAP3 MIR6726 UBE2J2 SCNN1D MIR200B TTLL10 TNFRSF4 CPSF3L TTLL10-AS1 PUSL1 MIR429 TNFRSF18

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline