This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03362311_cn
See other NOC2L CNV Assays ›
Gene Symbol
NOC2L
Assay Reference Genome
Location

Chr.1:950888 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03362311_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

26155

Gene Name:

NOC2 like nucleolar associated transcriptional repressor

Gene Aliases:

NET15, NET7, NIR, PPP1R112

Location:

Chr.1:944203-959299 on Build GRCh38

Assay Gene Location:

Within Intron 12
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NOC2L NM_015658.3 NP_056473.2
AK022756.1 BAB14230.1
AK024284.1
AK225239.1
AK315080.1
AL050019.1 CAB43240.2
BC003555.1 AAH03555.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2762302 Chr.1:914637 - 1106320 on Build GRCh38 Gain NOC2L C1orf159 HES4 PERM1 LOC100288175 LOC284600 ISG15 LOC100130417 KLHL17 SAMD11 LOC107985728 LOC105378948 PLEKHN1 AGRN RNF223
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss GNB1 C1orf159 TAS1R3 CFAP74 ANKRD65 LOC105378589 ATAD3C AURKAIP1 PRKCZ MIR6808 LOC107985729 TTLL10 CCNL2 ATAD3A MIR6723 DVL1 FAM87B LOC148413 TNFRSF4 VWA1 MIR6859-2 MMP23A LOC105378948 PLEKHN1 TMEM52 LOC105378592 LOC105378947 OR4F16 SSU72 LINC00115 LOC105378949 CALML6 LOC100133331 AGRN OR4F5 ATAD3B OR4F29 CDK11A LOC101928626 ISG15 TMEM240 MIR1302-2 LOC100506504 MRPL20 FNDC10 RNF223 HES4 FAM132A CPTP MTND2P28 LOC100288175 MIR6727 MORN1 DDX11L1 LOC729737 MIR6726 FAM138A SLC35E2 FAM41C PERM1 TTLL10-AS1 LOC102724312 WASH7P KLHL17 LINC01128 UBE2J2 MMP23B SDF4 LOC100287934 SKI MXRA8 LOC100288069 NADK FAAP20 LOC100129534 B3GALT6 LOC100130417 SCNN1D ACAP3 MIR200A LOC107984841 GABRD MIR429 LOC100134822 SLC35E2B MTND1P23 NOC2L LOC100132287 TNFRSF18 MIR6859-1 LOC102725121 TMEM88B LINC01342 MIR200B PUSL1 LOC284600 MIB2 CPSF3L SAMD11 LOC107985728 CDK11B LOC105378591
nsv950451 Chr.1:899421 - 1094520 on Build GRCh38 Deletion NOC2L C1orf159 HES4 PERM1 LOC100288175 LOC284600 ISG15 LOC100130417 KLHL17 SAMD11 LOC107985728 LOC105378948 PLEKHN1 AGRN RNF223
nsv509035 Chr.1:891406 - 985724 on Build GRCh38 Insertion NOC2L PERM1 SAMD11 LOC107985728 PLEKHN1 LOC284600 LOC100130417 KLHL17
dgv9n54 Chr.1:925754 - 952840 on Build GRCh38 Loss NOC2L SAMD11
nsv428334 Chr.1:874371 - 1220569 on Build GRCh38 Gain C1orf159 HES4 LOC100288175 LOC100130417 MIR200A MIR429 AGRN FAM41C TTLL10 NOC2L TNFRSF18 PERM1 TTLL10-AS1 LINC01342 MIR200B LOC284600 ISG15 KLHL17 TNFRSF4 SDF4 SAMD11 LOC107985728 LOC105378948 PLEKHN1 RNF223
dgv5n100 Chr.1:585989 - 1114424 on Build GRCh38 Gain C1orf159 HES4 LOC105378947 MTND2P28 LOC100288069 OR4F16 LOC100288175 LOC100130417 LOC107984841 LINC00115 LOC100133331 AGRN MTND1P23 FAM41C NOC2L MIR6723 PERM1 FAM87B LOC101928626 LOC284600 ISG15 KLHL17 LINC01128 SAMD11 LOC100287934 LOC107985728 LOC105378948 PLEKHN1 RNF223
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss C1orf159 HES4 TAS1R3 FAM132A CPTP ANKRD65 LOC100288175 MIR6727 ATAD3C AURKAIP1 MIR6726 MIR6808 LOC107985729 SLC35E2 FAM41C TTLL10 CCNL2 ATAD3A DVL1 PERM1 FAM87B TTLL10-AS1 LOC148413 LOC102724312 KLHL17 LINC01128 UBE2J2 TNFRSF4 MMP23B VWA1 SDF4 MMP23A LOC100287934 LOC105378948 PLEKHN1 MXRA8 LOC100288069 NADK B3GALT6 LOC100130417 SCNN1D SSU72 ACAP3 MIR200A LINC00115 MIR429 SLC35E2B AGRN ATAD3B NOC2L TNFRSF18 TMEM88B LINC01342 MIR200B CDK11A PUSL1 LOC284600 ISG15 TMEM240 MIB2 CPSF3L SAMD11 LOC107985728 MRPL20 CDK11B FNDC10 RNF223
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss C1orf159 HES4 FAM132A B3GALT6 LOC100288175 LOC100130417 MIR200A LINC00115 MIR429 AGRN FAM41C TTLL10 NOC2L TNFRSF18 PERM1 FAM87B TTLL10-AS1 LINC01342 MIR200B LOC284600 ISG15 KLHL17 LINC01128 UBE2J2 TNFRSF4 SDF4 SAMD11 LOC107985728 LOC105378948 PLEKHN1 RNF223
nsv1013524 Chr.1:939510 - 1161955 on Build GRCh38 Gain NOC2L C1orf159 HES4 PERM1 LINC01342 LOC100288175 ISG15 KLHL17 SAMD11 LOC105378948 PLEKHN1 AGRN RNF223
nsv544895 Chr.1:917392 - 1054900 on Build GRCh38 Loss NOC2L HES4 PERM1 SAMD11 LOC107985728 PLEKHN1 AGRN LOC284600 ISG15 LOC100130417 KLHL17
nsv1160644 Chr.1:939652 - 1220663 on Build GRCh38 Deletion NOC2L TNFRSF18 C1orf159 HES4 PERM1 TTLL10-AS1 LINC01342 MIR200B LOC100288175 ISG15 KLHL17 TNFRSF4 SDF4 MIR200A SAMD11 MIR429 LOC105378948 PLEKHN1 AGRN RNF223 TTLL10
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain C1orf159 HES4 TAS1R3 FAM132A CPTP ANKRD65 MXRA8 B3GALT6 LOC100288175 MIR6727 LOC100130417 SCNN1D ACAP3 MIR200A AURKAIP1 MIR429 MIR6726 MIR6808 AGRN TTLL10 CCNL2 NOC2L TNFRSF18 DVL1 PERM1 TMEM88B TTLL10-AS1 LINC01342 MIR200B LOC148413 PUSL1 LOC284600 ISG15 KLHL17 UBE2J2 TNFRSF4 CPSF3L SDF4 SAMD11 LOC107985728 MRPL20 LOC105378948 PLEKHN1 RNF223
nsv832980 Chr.1:848279 - 1007037 on Build GRCh38 Gain NOC2L HES4 PERM1 SAMD11 LOC107985728 PLEKHN1 LOC284600 FAM41C LOC100130417 KLHL17 LINC01128

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of B cell apoptotic process
transcription, DNA-templated
apoptotic process
chromatin assembly
nucleolus to nucleoplasm transport
cellular response to UV
negative regulation of histone acetylation
ribosomal large subunit biogenesis
regulation of signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway
chromatin binding
transcription corepressor activity
protein binding
nucleosome binding
histone binding
poly(A) RNA binding
repressing transcription factor binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline