This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03363726_cn
See other GNB1 CNV Assays ›
Gene Symbol
GNB1
Assay Reference Genome
Location

Chr.1:1803102 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03363726_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2782

Gene Name:

G protein subunit beta 1

Gene Aliases:

MRD42

Location:

Chr.1:1785285-1891117 on Build GRCh38

Assay Gene Location:

Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GNB1 NM_001282538.1 NP_001269467.1
NM_001282539.1 NP_001269468.1
NM_002074.4 NP_002065.1
XM_017001059.1 XP_016856548.1
XM_017001060.1 XP_016856549.1
XM_017001061.1 XP_016856550.1
AF070603.1 AAC28655.1
AF501882.1 AAM15918.1
AK122949.1
AK312614.1
BC004186.2 AAH04186.1
BC005888.2 AAH05888.1
BC008991.1 AAH08991.1
BC114618.1 AAI14619.1
BT007305.1 AAP35969.1
CR456784.1 CAG33065.1
M36430.1 AAA63265.1
X04526.1 CAA28207.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion NADK MMP23A CDK11A SLC35E2B SLC35E2 MMP23B CDK11B GNB1 MIB2 FNDC10
nsv834091 Chr.1:1701359 - 1880638 on Build GRCh38 Loss NADK MMP23A CDK11A SLC35E2 GNB1
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication NADK MMP23A CDK11A SLC35E2B SSU72 SLC35E2 MMP23B CDK11B GNB1 MIB2 FNDC10
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC100287934 CDK11A MTND2P28 LOC729737 B3GALT6 LOC102724312 LOC105378948 MIR200A LOC107985729 UBE2J2 LOC284600 MXRA8 NADK ACAP3 SSU72 LOC100132287 LOC100506504 MTND1P23 CFAP74 MMP23B CDK11B ISG15 MIB2 LOC100129534 FNDC10 SCNN1D MIR200B AURKAIP1 TMEM52 RNF223 MIR6859-2 TNFRSF18 LINC01128 LOC107985728 CPSF3L LOC148413 LOC102725121 LOC100288069 LOC105378947 PERM1 CPTP FAM132A MIR6723 LINC01342 ATAD3C FAM87B SDF4 DDX11L1 PRKCZ TMEM88B TTLL10 C1orf159 TMEM240 DVL1 LOC101928626 OR4F5 MMP23A LOC107984841 ANKRD65 KLHL17 LOC100133331 LOC100130417 LINC00115 MIR6859-1 LOC105378592 OR4F29 MIR1302-2 SAMD11 ATAD3B SKI CALML6 LOC105378949 MRPL20 LOC100288175 MIR6808 TNFRSF4 MIR6727 SLC35E2 ATAD3A LOC105378589 FAM41C FAM138A AGRN MORN1 SLC35E2B TTLL10-AS1 PUSL1 MIR6726 MIR429 HES4 GNB1 WASH7P VWA1 CCNL2 LOC105378591 GABRD TAS1R3 LOC100134822 PLEKHN1 FAAP20 OR4F16 NOC2L
nsv1160926 Chr.1:1772699 - 1905488 on Build GRCh38 Duplication NADK LOC105378949 GNB1
dgv10n100 Chr.1:1779975 - 1884542 on Build GRCh38 Gain NADK GNB1
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss MMP23A CDK11A ANKRD65 TMEM52 SLC35E2 ATAD3A LOC105378589 LOC102724312 SLC35E2B LOC107985729 LOC148413 NADK SSU72 ATAD3B GNB1 VWA1 CCNL2 ATAD3C CALML6 LOC105378591 CFAP74 LOC105378949 PRKCZ GABRD TMEM88B MMP23B CDK11B MRPL20 TMEM240 MIB2 FNDC10

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

protein folding
signal transduction
adenylate cyclase-activating dopamine receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
G-protein coupled acetylcholine receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
Ras protein signal transduction
cell proliferation
cardiac muscle cell apoptotic process
rhodopsin mediated signaling pathway
platelet activation
sensory perception of taste
retina development in camera-type eye
cellular response to glucagon stimulus
cellular response to prostaglandin E stimulus
cellular response to hypoxia
cellular response to catecholamine stimulus
GTPase activity
signal transducer activity
protein binding
spectrin binding
type 1 angiotensin receptor binding
protein complex binding
GTPase binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline