This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03365771_cn
See other GNB1 CNV Assays ›
Gene Symbol
GNB1
Assay Reference Genome
Location

Chr.1:1796937 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03365771_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2782

Gene Name:

G protein subunit beta 1

Gene Aliases:

MRD42

Location:

Chr.1:1785285-1891117 on Build GRCh38

Assay Gene Location:

Within Intron 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GNB1 NM_001282538.1 NP_001269467.1
NM_001282539.1 NP_001269468.1
NM_002074.4 NP_002065.1
XM_017001059.1 XP_016856548.1
XM_017001060.1 XP_016856549.1
XM_017001061.1 XP_016856550.1
AF070603.1 AAC28655.1
AF501882.1 AAM15918.1
AK122949.1
AK312614.1
BC004186.2 AAH04186.1
BC005888.2 AAH05888.1
BC008991.1 AAH08991.1
BC114618.1 AAI14619.1
BT007305.1 AAP35969.1
CR456784.1 CAG33065.1
M36430.1 AAA63265.1
X04526.1 CAA28207.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion MIB2 SLC35E2 MMP23B SLC35E2B NADK FNDC10 MMP23A CDK11B GNB1 CDK11A
nsv834091 Chr.1:1701359 - 1880638 on Build GRCh38 Loss SLC35E2 NADK MMP23A GNB1 CDK11A
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication SSU72 MIB2 SLC35E2 MMP23B SLC35E2B NADK FNDC10 MMP23A CDK11B GNB1 CDK11A
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss MIR6723 PLEKHN1 FAM132A SCNN1D LOC100129534 WASH7P ANKRD65 MMP23B SLC35E2B DVL1 LOC100133331 AURKAIP1 HES4 TMEM52 LOC100288175 LOC100288069 TTLL10 LOC105378589 CALML6 FAM41C KLHL17 GNB1 SSU72 ACAP3 CPTP MXRA8 MTND1P23 ATAD3B MRPL20 LOC105378947 ATAD3C TNFRSF18 LOC100132287 MORN1 LOC105378948 FAAP20 C1orf159 MIR200B SLC35E2 LOC102725121 GABRD ISG15 LOC100287934 AGRN LOC100134822 NOC2L CFAP74 TNFRSF4 NADK LOC107985729 CDK11B LOC107985728 UBE2J2 LOC100506504 MIR6726 MIR6727 B3GALT6 MIR429 SAMD11 LOC105378591 FNDC10 MIR200A OR4F16 OR4F29 VWA1 TMEM88B LINC01128 LOC729737 CPSF3L TAS1R3 MIR6859-1 TMEM240 MIR6859-2 LOC105378949 LOC105378592 MIR6808 PUSL1 LOC148413 CCNL2 CDK11A PERM1 LINC01342 LOC107984841 LINC00115 OR4F5 RNF223 DDX11L1 MMP23A TTLL10-AS1 FAM87B FAM138A MIB2 MIR1302-2 SDF4 PRKCZ LOC101928626 LOC284600 LOC102724312 ATAD3A LOC100130417 MTND2P28 SKI
nsv1160926 Chr.1:1772699 - 1905488 on Build GRCh38 Duplication LOC105378949 NADK GNB1
dgv10n100 Chr.1:1779975 - 1884542 on Build GRCh38 Gain NADK GNB1
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss LOC105378591 FNDC10 ATAD3C ANKRD65 SLC35E2 MMP23B SLC35E2B VWA1 GABRD TMEM88B MMP23A TMEM52 CFAP74 LOC105378589 MIB2 NADK CALML6 LOC107985729 CDK11B TMEM240 PRKCZ GNB1 SSU72 LOC105378949 LOC102724312 LOC148413 ATAD3B ATAD3A CCNL2 MRPL20 CDK11A

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

protein folding
signal transduction
adenylate cyclase-activating dopamine receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
G-protein coupled acetylcholine receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
Ras protein signal transduction
cell proliferation
cardiac muscle cell apoptotic process
rhodopsin mediated signaling pathway
platelet activation
sensory perception of taste
retina development in camera-type eye
cellular response to glucagon stimulus
cellular response to prostaglandin E stimulus
cellular response to hypoxia
cellular response to catecholamine stimulus
GTPase activity
signal transducer activity
protein binding
spectrin binding
type 1 angiotensin receptor binding
protein complex binding
GTPase binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline