This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs03366504_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1258914 on build GRCh38
Cytoband
1p36.33
Assay ID Hs03366504_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Intron 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1
BQ881730.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss UBE2J2 MIR6726 SCNN1D ACAP3
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication PUSL1 LOC105378948 LINC01342 CPTP TTLL10-AS1 FAM132A TTLL10 CPSF3L TNFRSF18 SDF4 MIR200B MIR200A TAS1R3 MIR6726 TNFRSF4 MIR429 RNF223 UBE2J2 LOC100288175 C1orf159 AGRN B3GALT6 SCNN1D MIR6727 ACAP3
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss FNDC10 LOC102724312 LOC105378948 LINC01342 FAM87B PERM1 TTLL10-AS1 TTLL10 HES4 VWA1 SLC35E2B MXRA8 MIR200B MIB2 NOC2L KLHL17 TAS1R3 MMP23B LOC148413 MIR6808 CDK11B MMP23A MIR6726 LOC107985728 MIR429 LOC100288069 UBE2J2 TMEM240 MRPL20 LOC100288175 LOC100130417 AGRN SAMD11 SCNN1D MIR6727 TMEM88B ISG15 PUSL1 LOC100287934 CPTP SLC35E2 SSU72 FAM132A CPSF3L CCNL2 TNFRSF18 SDF4 ATAD3B LINC01128 MIR200A FAM41C AURKAIP1 NADK LINC00115 LOC107985729 ATAD3C DVL1 TNFRSF4 LOC284600 RNF223 ATAD3A C1orf159 PLEKHN1 B3GALT6 ANKRD65 ACAP3 CDK11A
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss UBE2J2 MIR6726 SDF4 B3GALT6 SCNN1D FAM132A ACAP3
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss MIR6723 PERM1 OR4F16 GABRD HES4 VWA1 LOC102725121 SLC35E2B MXRA8 MIR200B MIB2 LOC100506504 NOC2L TAS1R3 MMP23B MIR6808 FAAP20 LOC107985728 GNB1 UBE2J2 TMEM240 LOC100130417 LOC100132287 AGRN TMEM88B PUSL1 CPTP LOC729737 SLC35E2 SSU72 FAM132A CPSF3L CCNL2 TNFRSF18 SDF4 ATAD3B MORN1 MIR200A FAM41C LINC00115 TNFRSF4 RNF223 FAM138A PLEKHN1 MIR6859-1 ACAP3 MTND2P28 FNDC10 LOC102724312 LOC105378948 LINC01342 FAM87B TTLL10-AS1 TTLL10 LOC107984841 LOC105378591 KLHL17 LOC148413 CDK11B MMP23A MIR6726 LOC101928626 OR4F29 LOC105378589 OR4F5 PRKCZ MIR429 LOC100288069 MRPL20 LOC100288175 SAMD11 SCNN1D SKI MIR6727 MTND1P23 ISG15 CFAP74 LOC105378947 LOC100287934 LOC100134822 MIR1302-2 LOC105378949 LOC105378592 DDX11L1 LINC01128 TMEM52 LOC100133331 AURKAIP1 NADK LOC100129534 LOC107985729 ATAD3C DVL1 CALML6 LOC284600 WASH7P ATAD3A C1orf159 B3GALT6 ANKRD65 MIR6859-2 CDK11A
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss LOC105378948 LINC01342 FAM87B PERM1 TTLL10-AS1 FAM132A TTLL10 HES4 TNFRSF18 SDF4 MIR200B LINC01128 MIR200A NOC2L KLHL17 FAM41C LOC107985728 LINC00115 TNFRSF4 LOC284600 MIR429 RNF223 UBE2J2 LOC100288175 C1orf159 LOC100130417 AGRN PLEKHN1 SAMD11 B3GALT6 ISG15
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion PUSL1 FNDC10 LOC102724312 LINC01342 CPTP TTLL10-AS1 SSU72 FAM132A TTLL10 CPSF3L VWA1 CCNL2 TNFRSF18 SDF4 MXRA8 MIR200B ATAD3B MIB2 MIR200A TAS1R3 MMP23B AURKAIP1 LOC148413 MIR6808 CDK11B MIR6726 LOC107985729 ATAD3C DVL1 TNFRSF4 MIR429 UBE2J2 ATAD3A TMEM240 MRPL20 C1orf159 B3GALT6 SCNN1D ANKRD65 MIR6727 ACAP3 TMEM88B
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain PUSL1 LOC105378948 LINC01342 CPTP PERM1 TTLL10-AS1 FAM132A TTLL10 CPSF3L HES4 CCNL2 TNFRSF18 SDF4 MXRA8 MIR200B MIR200A NOC2L KLHL17 TAS1R3 AURKAIP1 LOC148413 MIR6808 MIR6726 LOC107985728 DVL1 TNFRSF4 LOC284600 MIR429 RNF223 UBE2J2 MRPL20 LOC100288175 C1orf159 LOC100130417 AGRN PLEKHN1 SAMD11 B3GALT6 SCNN1D ANKRD65 MIR6727 ACAP3 TMEM88B ISG15
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain LOC105378948 LINC01342 TTLL10-AS1 TNFRSF4 FAM132A TTLL10 MIR429 RNF223 UBE2J2 C1orf159 TNFRSF18 SDF4 MIR200B B3GALT6 SCNN1D MIR200A
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss LINC01342 MIR6726 TTLL10-AS1 TNFRSF4 FAM132A TTLL10 MIR429 RNF223 UBE2J2 C1orf159 TNFRSF18 SDF4 MIR200B B3GALT6 SCNN1D MIR200A ACAP3
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain PUSL1 MIR6726 TTLL10-AS1 TNFRSF4 FAM132A TTLL10 MIR429 CPSF3L UBE2J2 TNFRSF18 SDF4 MIR200B B3GALT6 SCNN1D MIR200A MIR6727 ACAP3

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline